Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18628059-18628565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTD1
Entrez Gene ID
92675
Gene Name
D-tyrosyl-tRNA deacylase 1
Gene Aliases
C20orf88, DUE-B, DUEB, HARS2, pqn-68
UniGene
Hs.659442
Gene Chromosome Location
Chr.20: 18587893-18763917 on Build GRCh38
Transcripts
NM_001318043.1 NM_080820.5 XM_005260872.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6004365 c.374G>A p.G125D Chr.20: 18628130-18628130 on Build GRCh38
3544440 c.409A>C p.N137H Chr.20: 18628165-18628165 on Build GRCh38
4096917 c.456T>C p.T152T Chr.20: 18628212-18628212 on Build GRCh38
4646634 c.469C>A p.P157T Chr.20: 18628225-18628225 on Build GRCh38
3910925 c.440C>T p.S147L Chr.20: 18628196-18628196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141540138 Chr.20: 18628179-18628179 on Build GRCh38
rs891724584 Chr.20: 18628407-18628407 on Build GRCh38
rs34294812 Chr.20: 18628488-18628489 on Build GRCh38
rs969841935 Chr.20: 18628341-18628341 on Build GRCh38
rs2295574 Chr.20: 18628372-18628372 on Build GRCh38
rs142618746 Chr.20: 18628521-18628521 on Build GRCh38
rs1032916849 Chr.20: 18628230-18628230 on Build GRCh38
rs766017552 Chr.20: 18628237-18628237 on Build GRCh38
rs969517173 Chr.20: 18628100-18628100 on Build GRCh38
rs73900461 Chr.20: 18628406-18628406 on Build GRCh38
rs749370547 Chr.20: 18628197-18628197 on Build GRCh38
rs555884164 Chr.20: 18628298-18628298 on Build GRCh38
rs918477061 Chr.20: 18628383-18628383 on Build GRCh38
rs3818220 Chr.20: 18628301-18628301 on Build GRCh38
rs776160314 Chr.20: 18628253-18628253 on Build GRCh38
rs767120102 Chr.20: 18628087-18628087 on Build GRCh38
rs746109514 Chr.20: 18628173-18628173 on Build GRCh38
rs773380232 Chr.20: 18628218-18628218 on Build GRCh38
rs78934288 Chr.20: 18628262-18628262 on Build GRCh38
rs942435323 Chr.20: 18628268-18628268 on Build GRCh38
rs752714953 Chr.20: 18628261-18628261 on Build GRCh38
rs368781399 Chr.20: 18628104-18628104 on Build GRCh38
rs112846864 Chr.20: 18628516-18628516 on Build GRCh38
rs752976817 Chr.20: 18628269-18628269 on Build GRCh38
rs984327975 Chr.20: 18628433-18628433 on Build GRCh38
rs751296630 Chr.20: 18628267-18628267 on Build GRCh38
rs3818221 Chr.20: 18628293-18628293 on Build GRCh38
rs1023816568 Chr.20: 18628337-18628337 on Build GRCh38
rs759024818 Chr.20: 18628257-18628257 on Build GRCh38
rs1048636612 Chr.20: 18628085-18628085 on Build GRCh38
rs911060374 Chr.20: 18628440-18628440 on Build GRCh38
rs758822457 Chr.20: 18628159-18628159 on Build GRCh38
rs375053755 Chr.20: 18628271-18628271 on Build GRCh38
rs780336372 Chr.20: 18628196-18628196 on Build GRCh38
rs146240497 Chr.20: 18628091-18628091 on Build GRCh38
rs893954348 Chr.20: 18628326-18628326 on Build GRCh38
rs535348765 Chr.20: 18628309-18628309 on Build GRCh38
rs769322465 Chr.20: 18628198-18628198 on Build GRCh38
rs200436210 Chr.20: 18628131-18628131 on Build GRCh38
rs562696895 Chr.20: 18628517-18628517 on Build GRCh38
rs1000014960 Chr.20: 18628389-18628389 on Build GRCh38
rs760379955 Chr.20: 18628233-18628233 on Build GRCh38
rs1040852055 Chr.20: 18628507-18628507 on Build GRCh38
rs1003585581 Chr.20: 18628366-18628366 on Build GRCh38
rs942427066 Chr.20: 18628488-18628488 on Build GRCh38
rs754978568 Chr.20: 18628145-18628145 on Build GRCh38
rs531863942 Chr.20: 18628451-18628451 on Build GRCh38
rs1000555965 Chr.20: 18628208-18628208 on Build GRCh38
rs552269007 Chr.20: 18628206-18628206 on Build GRCh38
rs910008813 Chr.20: 18628531-18628531 on Build GRCh38
rs1011466852 Chr.20: 18628328-18628328 on Build GRCh38
rs999773546 Chr.20: 18628300-18628300 on Build GRCh38
rs77499464 Chr.20: 18628373-18628373 on Build GRCh38
rs780389306 Chr.20: 18628273-18628273 on Build GRCh38
rs764607012 Chr.20: 18628259-18628259 on Build GRCh38
rs1023947955 Chr.20: 18628285-18628285 on Build GRCh38
rs545578155 Chr.20: 18628382-18628382 on Build GRCh38
rs780781020 Chr.20: 18628308-18628308 on Build GRCh38
rs375449236 Chr.20: 18628099-18628099 on Build GRCh38
rs777394975 Chr.20: 18628169-18628169 on Build GRCh38
rs910114707 Chr.20: 18628364-18628364 on Build GRCh38
rs772516651 Chr.20: 18628207-18628207 on Build GRCh38
rs1037544024 Chr.20: 18628113-18628113 on Build GRCh38
rs370279612 Chr.20: 18628153-18628153 on Build GRCh38
rs576920896 Chr.20: 18628425-18628425 on Build GRCh38

View Full Product Details