Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23748929-23749426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST1
Entrez Gene ID
1469
Gene Name
cystatin SN
Gene Aliases
-
UniGene
Hs.123114
Gene Chromosome Location
Chr.20: 23747553-23750937 on Build GRCh38
Transcripts
NM_001898.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7363104 c.236G>T p.G79V Chr.20: 23749122-23749122 on Build GRCh38
7363102 c.244A>G p.N82D Chr.20: 23749114-23749114 on Build GRCh38
5068036 c.241_242insGG p.V81fs*2 Chr.20: 23749116-23749117 on Build GRCh38
7363103 c.236G>C p.G79A Chr.20: 23749122-23749122 on Build GRCh38
3389637 c.232G>A p.V78I Chr.20: 23749126-23749126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773480726 Chr.20: 23748973-23748973 on Build GRCh38
rs779205556 Chr.20: 23749086-23749086 on Build GRCh38
rs370189313 Chr.20: 23749090-23749090 on Build GRCh38
rs534368202 Chr.20: 23749120-23749120 on Build GRCh38
rs201505177 Chr.20: 23749099-23749099 on Build GRCh38
rs765258832 Chr.20: 23749338-23749338 on Build GRCh38
rs746147492 Chr.20: 23749102-23749102 on Build GRCh38
rs756782667 Chr.20: 23749062-23749062 on Build GRCh38
rs538836090 Chr.20: 23749371-23749371 on Build GRCh38
rs200524765 Chr.20: 23749156-23749156 on Build GRCh38
rs781501457 Chr.20: 23749100-23749100 on Build GRCh38
rs766239558 Chr.20: 23749333-23749333 on Build GRCh38
rs753225892 Chr.20: 23749087-23749087 on Build GRCh38
rs768428419 Chr.20: 23749118-23749118 on Build GRCh38
rs748198710 Chr.20: 23748966-23748966 on Build GRCh38
rs112974717 Chr.20: 23748993-23748993 on Build GRCh38
rs751028209 Chr.20: 23749061-23749061 on Build GRCh38
rs762833357 Chr.20: 23749109-23749109 on Build GRCh38
rs148178833 Chr.20: 23749387-23749387 on Build GRCh38
rs919564675 Chr.20: 23749300-23749300 on Build GRCh38
rs968027409 Chr.20: 23749214-23749214 on Build GRCh38
rs556178746 Chr.20: 23749130-23749130 on Build GRCh38
rs1020872102 Chr.20: 23749172-23749172 on Build GRCh38
rs375967648 Chr.20: 23749150-23749150 on Build GRCh38
rs940229740 Chr.20: 23749276-23749276 on Build GRCh38
rs1019514872 Chr.20: 23749159-23749159 on Build GRCh38
rs763163192 Chr.20: 23749146-23749146 on Build GRCh38
rs747204459 Chr.20: 23749022-23749022 on Build GRCh38
rs780073308 Chr.20: 23749163-23749163 on Build GRCh38
rs778730371 Chr.20: 23749173-23749173 on Build GRCh38
rs772181608 Chr.20: 23749015-23749015 on Build GRCh38
rs1057124611 Chr.20: 23748953-23748953 on Build GRCh38
rs531311199 Chr.20: 23749302-23749302 on Build GRCh38
rs903838471 Chr.20: 23749372-23749372 on Build GRCh38
rs757412216 Chr.20: 23749123-23749123 on Build GRCh38
rs7354713 Chr.20: 23749000-23749000 on Build GRCh38
rs768501392 Chr.20: 23749132-23749132 on Build GRCh38
rs565470782 Chr.20: 23749256-23749256 on Build GRCh38
rs761798325 Chr.20: 23749134-23749134 on Build GRCh38
rs566536535 Chr.20: 23748949-23748949 on Build GRCh38
rs146690838 Chr.20: 23749121-23749121 on Build GRCh38
rs751198098 Chr.20: 23749001-23749001 on Build GRCh38
rs529861770 Chr.20: 23749218-23749218 on Build GRCh38
rs767097742 Chr.20: 23749066-23749066 on Build GRCh38
rs748048090 Chr.20: 23749175-23749175 on Build GRCh38
rs143819315 Chr.20: 23749303-23749303 on Build GRCh38
rs755348321 Chr.20: 23749012-23749012 on Build GRCh38
rs1041315221 Chr.20: 23749334-23749334 on Build GRCh38
rs1033709220 Chr.20: 23749389-23749389 on Build GRCh38
rs979455366 Chr.20: 23749225-23749225 on Build GRCh38
rs781037716 Chr.20: 23749117-23749117 on Build GRCh38
rs992762827 Chr.20: 23749261-23749261 on Build GRCh38
rs376987841 Chr.20: 23748983-23748983 on Build GRCh38
rs772050680 Chr.20: 23748972-23748972 on Build GRCh38
rs3188313 Chr.20: 23749126-23749126 on Build GRCh38
rs1033741788 Chr.20: 23749235-23749235 on Build GRCh38
rs561205427 Chr.20: 23749180-23749180 on Build GRCh38
rs143185509 Chr.20: 23749369-23749369 on Build GRCh38
rs772749942 Chr.20: 23749138-23749138 on Build GRCh38
rs889633760 Chr.20: 23749058-23749058 on Build GRCh38
rs748978249 Chr.20: 23749127-23749127 on Build GRCh38
rs117082322 Chr.20: 23749280-23749280 on Build GRCh38
rs139812159 Chr.20: 23749033-23749033 on Build GRCh38
rs753546545 Chr.20: 23749145-23749145 on Build GRCh38
rs373733417 Chr.20: 23749170-23749170 on Build GRCh38
rs116626492 Chr.20: 23749270-23749270 on Build GRCh38
rs552250628 Chr.20: 23749029-23749029 on Build GRCh38
rs1008117728 Chr.20: 23749070-23749070 on Build GRCh38
rs555199581 Chr.20: 23749108-23749108 on Build GRCh38
rs540466158 Chr.20: 23749228-23749228 on Build GRCh38
rs749605831 Chr.20: 23749011-23749011 on Build GRCh38
rs752593595 Chr.20: 23748994-23748994 on Build GRCh38
rs771428973 Chr.20: 23749148-23749149 on Build GRCh38
rs746209786 Chr.20: 23749123-23749124 on Build GRCh38
rs763625180 Chr.20: 23749057-23749057 on Build GRCh38
rs370802976 Chr.20: 23749168-23749168 on Build GRCh38
rs553989967 Chr.20: 23749402-23749402 on Build GRCh38
rs758870694 Chr.20: 23749088-23749088 on Build GRCh38
rs3188314 Chr.20: 23749085-23749085 on Build GRCh38
rs750079881 Chr.20: 23749072-23749072 on Build GRCh38
rs889562509 Chr.20: 23749327-23749327 on Build GRCh38
rs368289519 Chr.20: 23748990-23748990 on Build GRCh38
rs972990170 Chr.20: 23749299-23749299 on Build GRCh38
rs769607319 Chr.20: 23749103-23749103 on Build GRCh38
rs183935304 Chr.20: 23749373-23749373 on Build GRCh38
rs527236830 Chr.20: 23749374-23749374 on Build GRCh38
rs376106415 Chr.20: 23749157-23749157 on Build GRCh38
rs760263887 Chr.20: 23749140-23749140 on Build GRCh38
rs372538698 Chr.20: 23749119-23749119 on Build GRCh38
rs959763446 Chr.20: 23749396-23749396 on Build GRCh38
rs756924682 Chr.20: 23749116-23749117 on Build GRCh38
rs372641078 Chr.20: 23749007-23749007 on Build GRCh38
rs577447988 Chr.20: 23749298-23749298 on Build GRCh38
rs766256150 Chr.20: 23748988-23748988 on Build GRCh38
rs778379691 Chr.20: 23749089-23749089 on Build GRCh38
rs771957958 Chr.20: 23749179-23749179 on Build GRCh38
rs375466801 Chr.20: 23749155-23749155 on Build GRCh38
rs771242039 Chr.20: 23749030-23749030 on Build GRCh38
rs781468557 Chr.20: 23749125-23749125 on Build GRCh38
rs375902412 Chr.20: 23749037-23749037 on Build GRCh38
rs112232806 Chr.20: 23749122-23749122 on Build GRCh38
rs891628914 Chr.20: 23749336-23749336 on Build GRCh38
rs765902215 Chr.20: 23749143-23749143 on Build GRCh38
rs372192719 Chr.20: 23748965-23748965 on Build GRCh38
rs762843755 Chr.20: 23749041-23749041 on Build GRCh38
rs544717292 Chr.20: 23749348-23749348 on Build GRCh38
rs543969659 Chr.20: 23749158-23749158 on Build GRCh38
rs879142715 Chr.20: 23749220-23749220 on Build GRCh38
rs190915622 Chr.20: 23749285-23749285 on Build GRCh38
rs771551065 Chr.20: 23749137-23749137 on Build GRCh38

View Full Product Details