Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23750661-23751163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST1
Entrez Gene ID
1469
Gene Name
cystatin SN
Gene Aliases
-
UniGene
Hs.123114
Gene Chromosome Location
Chr.20: 23747553-23750937 on Build GRCh38
Transcripts
NM_001898.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5454818 c.72G>T p.K24N Chr.20: 23750795-23750795 on Build GRCh38
1737433 c.7C>A p.Q3K Chr.20: 23750860-23750860 on Build GRCh38
253705 c.112G>A p.D38N Chr.20: 23750755-23750755 on Build GRCh38
5707370 c.103T>C p.Y35H Chr.20: 23750764-23750764 on Build GRCh38
7423907 c.37G>A p.A13T Chr.20: 23750830-23750830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565527258 Chr.20: 23750909-23750909 on Build GRCh38
rs776620783 Chr.20: 23750859-23750859 on Build GRCh38
rs141813234 Chr.20: 23750828-23750828 on Build GRCh38
rs767388872 Chr.20: 23750810-23750810 on Build GRCh38
rs771421684 Chr.20: 23750693-23750693 on Build GRCh38
rs577584309 Chr.20: 23750948-23750948 on Build GRCh38
rs574300031 Chr.20: 23750963-23750963 on Build GRCh38
rs199758963 Chr.20: 23750822-23750822 on Build GRCh38
rs530782485 Chr.20: 23750886-23750886 on Build GRCh38
rs774372228 Chr.20: 23750793-23750793 on Build GRCh38
rs2070855 Chr.20: 23751028-23751028 on Build GRCh38
rs13045804 Chr.20: 23751016-23751016 on Build GRCh38
rs6076122 Chr.20: 23750857-23750857 on Build GRCh38
rs768554764 Chr.20: 23750825-23750825 on Build GRCh38
rs151261857 Chr.20: 23751069-23751069 on Build GRCh38
rs201376709 Chr.20: 23750890-23750890 on Build GRCh38
rs749815196 Chr.20: 23750847-23750847 on Build GRCh38
rs756160302 Chr.20: 23750781-23750781 on Build GRCh38
rs370685433 Chr.20: 23750978-23750978 on Build GRCh38
rs780592963 Chr.20: 23750754-23750755 on Build GRCh38
rs773666920 Chr.20: 23750690-23750690 on Build GRCh38
rs770829389 Chr.20: 23750905-23750905 on Build GRCh38
rs775498598 Chr.20: 23750912-23750912 on Build GRCh38
rs3188296 Chr.20: 23750702-23750702 on Build GRCh38
rs749007247 Chr.20: 23750686-23750686 on Build GRCh38
rs879190888 Chr.20: 23750761-23750762 on Build GRCh38
rs781590256 Chr.20: 23750798-23750798 on Build GRCh38
rs900341340 Chr.20: 23750975-23750975 on Build GRCh38
rs200307238 Chr.20: 23750782-23750782 on Build GRCh38
rs150586879 Chr.20: 23750833-23750833 on Build GRCh38
rs149367943 Chr.20: 23750779-23750779 on Build GRCh38
rs571670080 Chr.20: 23750881-23750881 on Build GRCh38
rs13045477 Chr.20: 23750789-23750789 on Build GRCh38
rs758030895 Chr.20: 23750830-23750830 on Build GRCh38
rs770919061 Chr.20: 23750858-23750858 on Build GRCh38
rs566215865 Chr.20: 23750717-23750717 on Build GRCh38
rs368640809 Chr.20: 23750774-23750774 on Build GRCh38
rs1039240472 Chr.20: 23750968-23750968 on Build GRCh38
rs777437512 Chr.20: 23750831-23750831 on Build GRCh38
rs754461118 Chr.20: 23750788-23750788 on Build GRCh38
rs781114268 Chr.20: 23750733-23750733 on Build GRCh38
rs778138934 Chr.20: 23750899-23750899 on Build GRCh38
rs529213045 Chr.20: 23751034-23751034 on Build GRCh38
rs750450795 Chr.20: 23750746-23750746 on Build GRCh38
rs140613927 Chr.20: 23750738-23750738 on Build GRCh38
rs747825121 Chr.20: 23750790-23750790 on Build GRCh38
rs1047502032 Chr.20: 23750945-23750945 on Build GRCh38
rs757630639 Chr.20: 23750797-23750797 on Build GRCh38
rs1023518465 Chr.20: 23751086-23751086 on Build GRCh38
rs561328965 Chr.20: 23751030-23751030 on Build GRCh38
rs573542764 Chr.20: 23751114-23751114 on Build GRCh38
rs754507565 Chr.20: 23750824-23750824 on Build GRCh38
rs760570692 Chr.20: 23750815-23750815 on Build GRCh38
rs950356287 Chr.20: 23751142-23751142 on Build GRCh38
rs748119641 Chr.20: 23750896-23750896 on Build GRCh38
rs77188058 Chr.20: 23750748-23750748 on Build GRCh38
rs540721922 Chr.20: 23750805-23750805 on Build GRCh38
rs777019391 Chr.20: 23750700-23750700 on Build GRCh38
rs548823200 Chr.20: 23751041-23751041 on Build GRCh38
rs778452912 Chr.20: 23750758-23750758 on Build GRCh38
rs559999556 Chr.20: 23751002-23751002 on Build GRCh38
rs534420407 Chr.20: 23750914-23750914 on Build GRCh38
rs759443369 Chr.20: 23750861-23750861 on Build GRCh38
rs531019169 Chr.20: 23751027-23751027 on Build GRCh38
rs774332779 Chr.20: 23750852-23750852 on Build GRCh38
rs775986521 Chr.20: 23750804-23750804 on Build GRCh38
rs386813407 Chr.20: 23751067-23751069 on Build GRCh38
rs536277016 Chr.20: 23751053-23751053 on Build GRCh38
rs565562046 Chr.20: 23751045-23751045 on Build GRCh38
rs569786302 Chr.20: 23750731-23750731 on Build GRCh38
rs748247973 Chr.20: 23750833-23750833 on Build GRCh38
rs572086468 Chr.20: 23750730-23750730 on Build GRCh38
rs763666467 Chr.20: 23750878-23750878 on Build GRCh38
rs386813406 Chr.20: 23751002-23751003 on Build GRCh38
rs111818589 Chr.20: 23751083-23751083 on Build GRCh38
rs756454450 Chr.20: 23750885-23750885 on Build GRCh38
rs772252501 Chr.20: 23750856-23750860 on Build GRCh38
rs146601403 Chr.20: 23750769-23750769 on Build GRCh38
rs113896553 Chr.20: 23750952-23750952 on Build GRCh38
rs6083250 Chr.20: 23751084-23751084 on Build GRCh38
rs80263900 Chr.20: 23750801-23750801 on Build GRCh38
rs3188297 Chr.20: 23750689-23750689 on Build GRCh38
rs144729819 Chr.20: 23751067-23751067 on Build GRCh38
rs763214720 Chr.20: 23750716-23750716 on Build GRCh38
rs6083249 Chr.20: 23751082-23751082 on Build GRCh38
rs762459197 Chr.20: 23750877-23750877 on Build GRCh38
rs765370716 Chr.20: 23750708-23750708 on Build GRCh38
rs555186339 Chr.20: 23750898-23750898 on Build GRCh38
rs144543419 Chr.20: 23750840-23750840 on Build GRCh38
rs769846587 Chr.20: 23750765-23750765 on Build GRCh38
rs528650630 Chr.20: 23751003-23751003 on Build GRCh38
rs545539848 Chr.20: 23751004-23751004 on Build GRCh38
rs201315915 Chr.20: 23750866-23750866 on Build GRCh38
rs3188301 Chr.20: 23750681-23750681 on Build GRCh38
rs748290646 Chr.20: 23750855-23750855 on Build GRCh38
rs556897543 Chr.20: 23750953-23750953 on Build GRCh38
rs891106928 Chr.20: 23751013-23751013 on Build GRCh38
rs557370944 Chr.20: 23750814-23750814 on Build GRCh38
rs982075732 Chr.20: 23751134-23751134 on Build GRCh38
rs777121328 Chr.20: 23750761-23750761 on Build GRCh38
rs749699113 Chr.20: 23750756-23750757 on Build GRCh38
rs6076121 Chr.20: 23750821-23750821 on Build GRCh38
rs140676884 Chr.20: 23750990-23750990 on Build GRCh38
rs372905911 Chr.20: 23750786-23750786 on Build GRCh38
rs143142026 Chr.20: 23750851-23750851 on Build GRCh38
rs71333777 Chr.20: 23751082-23751084 on Build GRCh38
rs548281880 Chr.20: 23750691-23750691 on Build GRCh38
rs151291851 Chr.20: 23750685-23750685 on Build GRCh38
rs769573688 Chr.20: 23750910-23750910 on Build GRCh38
rs756498779 Chr.20: 23750741-23750742 on Build GRCh38
rs528575607 Chr.20: 23750724-23750724 on Build GRCh38
rs143815991 Chr.20: 23750759-23750759 on Build GRCh38
rs747294673 Chr.20: 23750903-23750903 on Build GRCh38
rs761661785 Chr.20: 23750776-23750776 on Build GRCh38
rs746347123 Chr.20: 23750800-23750800 on Build GRCh38
rs6076123 Chr.20: 23750923-23750923 on Build GRCh38
rs556724843 Chr.20: 23751098-23751098 on Build GRCh38
rs2070856 Chr.20: 23750775-23750775 on Build GRCh38
rs550064441 Chr.20: 23750887-23750887 on Build GRCh38
rs1141013 Chr.20: 23750875-23750875 on Build GRCh38
rs117238940 Chr.20: 23750752-23750752 on Build GRCh38
rs752907397 Chr.20: 23750715-23750715 on Build GRCh38
rs774617467 Chr.20: 23750807-23750807 on Build GRCh38
rs769155130 Chr.20: 23750775-23750775 on Build GRCh38
rs553672708 Chr.20: 23751130-23751130 on Build GRCh38
rs374004963 Chr.20: 23750880-23750880 on Build GRCh38
rs542879965 Chr.20: 23750846-23750846 on Build GRCh38
rs760095816 Chr.20: 23750763-23750763 on Build GRCh38
rs772372352 Chr.20: 23750856-23750856 on Build GRCh38
rs569886383 Chr.20: 23750889-23750889 on Build GRCh38
rs764777005 Chr.20: 23750863-23750863 on Build GRCh38
rs368695583 Chr.20: 23751116-23751116 on Build GRCh38
rs368203290 Chr.20: 23750707-23750707 on Build GRCh38
rs753833000 Chr.20: 23750823-23750823 on Build GRCh38

View Full Product Details