Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25441581-25442107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINS1
Entrez Gene ID
9837
Gene Name
GINS complex subunit 1
Gene Aliases
PSF1
UniGene
Hs.658464
Gene Chromosome Location
Chr.20: 25407470-25448568 on Build GRCh38
Transcripts
NM_021067.4 NR_134574.1 XM_017028162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208186 c.472G>T p.E158* Chr.20: 25441726-25441726 on Build GRCh38
5057210 c.491G>A p.G164D Chr.20: 25441745-25441745 on Build GRCh38
311453 c.507delA p.N172fs*14 Chr.20: 25441761-25441761 on Build GRCh38
6671036 c.452G>A p.R151Q Chr.20: 25441706-25441706 on Build GRCh38
5531702 c.502C>T p.L168L Chr.20: 25441756-25441756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006691916 Chr.20: 25441931-25441931 on Build GRCh38
rs762561743 Chr.20: 25441705-25441705 on Build GRCh38
rs372460196 Chr.20: 25441787-25441787 on Build GRCh38
rs746790334 Chr.20: 25441671-25441671 on Build GRCh38
rs985800076 Chr.20: 25441626-25441626 on Build GRCh38
rs779677003 Chr.20: 25441872-25441872 on Build GRCh38
rs973422914 Chr.20: 25442018-25442018 on Build GRCh38
rs1039983035 Chr.20: 25441963-25441963 on Build GRCh38
rs543112005 Chr.20: 25441736-25441736 on Build GRCh38
rs746803614 Chr.20: 25441758-25441758 on Build GRCh38
rs537344888 Chr.20: 25441814-25441814 on Build GRCh38
rs748871676 Chr.20: 25442030-25442030 on Build GRCh38
rs1042018969 Chr.20: 25441856-25441856 on Build GRCh38
rs962658022 Chr.20: 25441940-25441940 on Build GRCh38
rs755911974 Chr.20: 25441760-25441761 on Build GRCh38
rs954018149 Chr.20: 25441925-25441925 on Build GRCh38
rs113727746 Chr.20: 25441919-25441919 on Build GRCh38
rs3764694 Chr.20: 25441847-25441847 on Build GRCh38
rs775127073 Chr.20: 25441695-25441695 on Build GRCh38
rs6083865 Chr.20: 25441756-25441756 on Build GRCh38
rs1014767660 Chr.20: 25442065-25442065 on Build GRCh38
rs773994647 Chr.20: 25441827-25441827 on Build GRCh38
rs758590981 Chr.20: 25441748-25441748 on Build GRCh38
rs752802883 Chr.20: 25441740-25441740 on Build GRCh38
rs35367807 Chr.20: 25442004-25442005 on Build GRCh38
rs769363143 Chr.20: 25441688-25441688 on Build GRCh38
rs145931317 Chr.20: 25441770-25441770 on Build GRCh38
rs982209087 Chr.20: 25442078-25442078 on Build GRCh38
rs745858629 Chr.20: 25441775-25441775 on Build GRCh38
rs781279371 Chr.20: 25441773-25441773 on Build GRCh38
rs531677406 Chr.20: 25442061-25442061 on Build GRCh38
rs769830918 Chr.20: 25441779-25441779 on Build GRCh38
rs61170040 Chr.20: 25442036-25442036 on Build GRCh38
rs900200838 Chr.20: 25441970-25441970 on Build GRCh38
rs572625827 Chr.20: 25442049-25442049 on Build GRCh38
rs894022863 Chr.20: 25441798-25441798 on Build GRCh38
rs996106194 Chr.20: 25441972-25441972 on Build GRCh38
rs1028008568 Chr.20: 25442029-25442029 on Build GRCh38
rs376610445 Chr.20: 25441709-25441709 on Build GRCh38
rs778204667 Chr.20: 25441682-25441683 on Build GRCh38
rs770807594 Chr.20: 25441676-25441676 on Build GRCh38
rs566209853 Chr.20: 25441683-25441683 on Build GRCh38
rs973422242 Chr.20: 25441707-25441707 on Build GRCh38
rs754273692 Chr.20: 25441731-25441731 on Build GRCh38
rs909763670 Chr.20: 25441632-25441632 on Build GRCh38
rs370853877 Chr.20: 25441735-25441735 on Build GRCh38
rs199881509 Chr.20: 25441682-25441682 on Build GRCh38
rs567233253 Chr.20: 25441871-25441871 on Build GRCh38
rs567569896 Chr.20: 25441973-25441973 on Build GRCh38
rs750783029 Chr.20: 25441711-25441711 on Build GRCh38
rs773826684 Chr.20: 25441906-25441906 on Build GRCh38
rs750227570 Chr.20: 25441761-25441761 on Build GRCh38
rs761508160 Chr.20: 25442032-25442032 on Build GRCh38
rs1017300979 Chr.20: 25441933-25441933 on Build GRCh38
rs944464535 Chr.20: 25441912-25441912 on Build GRCh38
rs528270299 Chr.20: 25441722-25441722 on Build GRCh38
rs73904473 Chr.20: 25441897-25441897 on Build GRCh38

View Full Product Details