Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33178005-33178502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA2
Entrez Gene ID
140683
Gene Name
BPI fold containing family A member 2
Gene Aliases
C20orf70, PSP, SPLUNC2, bA49G10.1
UniGene
Hs.125960
Gene Chromosome Location
Chr.20: 33161848-33181417 on Build GRCh38
Transcripts
NM_001319164.1 NM_080574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752161 c.617G>T p.S206I Chr.20: 33178200-33178200 on Build GRCh38
4665963 c.588C>T p.F196F Chr.20: 33178171-33178171 on Build GRCh38
1246770 c.642G>A p.K214K Chr.20: 33178225-33178225 on Build GRCh38
4752160 c.602T>A p.I201N Chr.20: 33178185-33178185 on Build GRCh38
4653271 c.570C>T p.S190S Chr.20: 33178153-33178153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773913502 Chr.20: 33178126-33178126 on Build GRCh38
rs562811373 Chr.20: 33178191-33178191 on Build GRCh38
rs753090984 Chr.20: 33178160-33178160 on Build GRCh38
rs556126086 Chr.20: 33178428-33178428 on Build GRCh38
rs577910391 Chr.20: 33178062-33178062 on Build GRCh38
rs1001676557 Chr.20: 33178073-33178073 on Build GRCh38
rs567702815 Chr.20: 33178394-33178394 on Build GRCh38
rs753666052 Chr.20: 33178269-33178269 on Build GRCh38
rs760053685 Chr.20: 33178153-33178153 on Build GRCh38
rs756008861 Chr.20: 33178063-33178063 on Build GRCh38
rs765909063 Chr.20: 33178158-33178158 on Build GRCh38
rs186551834 Chr.20: 33178121-33178121 on Build GRCh38
rs1045817464 Chr.20: 33178342-33178342 on Build GRCh38
rs755360221 Chr.20: 33178135-33178136 on Build GRCh38
rs750572385 Chr.20: 33178181-33178181 on Build GRCh38
rs907443835 Chr.20: 33178347-33178347 on Build GRCh38
rs528886764 Chr.20: 33178171-33178171 on Build GRCh38
rs201394079 Chr.20: 33178216-33178216 on Build GRCh38
rs8125148 Chr.20: 33178084-33178084 on Build GRCh38
rs779319559 Chr.20: 33178162-33178164 on Build GRCh38
rs756035208 Chr.20: 33178184-33178184 on Build GRCh38
rs117298016 Chr.20: 33178325-33178325 on Build GRCh38
rs1002218758 Chr.20: 33178354-33178354 on Build GRCh38
rs988253772 Chr.20: 33178038-33178038 on Build GRCh38
rs539973676 Chr.20: 33178152-33178152 on Build GRCh38
rs370964727 Chr.20: 33178142-33178142 on Build GRCh38
rs967329609 Chr.20: 33178159-33178159 on Build GRCh38
rs958106050 Chr.20: 33178377-33178377 on Build GRCh38
rs907450806 Chr.20: 33178072-33178072 on Build GRCh38
rs1011647294 Chr.20: 33178115-33178115 on Build GRCh38
rs767083982 Chr.20: 33178132-33178132 on Build GRCh38
rs1030784847 Chr.20: 33178448-33178448 on Build GRCh38
rs374784792 Chr.20: 33178205-33178205 on Build GRCh38
rs756229519 Chr.20: 33178240-33178240 on Build GRCh38
rs778625003 Chr.20: 33178303-33178303 on Build GRCh38
rs893277405 Chr.20: 33178393-33178393 on Build GRCh38
rs941074069 Chr.20: 33178264-33178264 on Build GRCh38
rs1321426 Chr.20: 33178398-33178398 on Build GRCh38
rs761553728 Chr.20: 33178129-33178129 on Build GRCh38
rs963729469 Chr.20: 33178463-33178463 on Build GRCh38
rs1038096240 Chr.20: 33178295-33178295 on Build GRCh38
rs955296851 Chr.20: 33178460-33178460 on Build GRCh38
rs373151340 Chr.20: 33178192-33178192 on Build GRCh38
rs777951400 Chr.20: 33178415-33178415 on Build GRCh38
rs775822241 Chr.20: 33178221-33178221 on Build GRCh38
rs933585758 Chr.20: 33178068-33178068 on Build GRCh38
rs760317544 Chr.20: 33178214-33178214 on Build GRCh38
rs958218923 Chr.20: 33178037-33178037 on Build GRCh38
rs750482356 Chr.20: 33178239-33178239 on Build GRCh38
rs763268966 Chr.20: 33178165-33178165 on Build GRCh38
rs561787624 Chr.20: 33178169-33178169 on Build GRCh38
rs772844929 Chr.20: 33178209-33178209 on Build GRCh38
rs369127446 Chr.20: 33178228-33178228 on Build GRCh38
rs762021343 Chr.20: 33178235-33178235 on Build GRCh38
rs34084783 Chr.20: 33178086-33178086 on Build GRCh38
rs768044154 Chr.20: 33178315-33178315 on Build GRCh38
rs918357553 Chr.20: 33178297-33178297 on Build GRCh38
rs549216722 Chr.20: 33178390-33178390 on Build GRCh38
rs751959026 Chr.20: 33178233-33178233 on Build GRCh38
rs780210872 Chr.20: 33178267-33178267 on Build GRCh38
rs145037130 Chr.20: 33178172-33178172 on Build GRCh38
rs896851583 Chr.20: 33178033-33178033 on Build GRCh38
rs745748781 Chr.20: 33178097-33178097 on Build GRCh38
rs143314379 Chr.20: 33178231-33178231 on Build GRCh38
rs748521842 Chr.20: 33178279-33178279 on Build GRCh38
rs769731903 Chr.20: 33178101-33178101 on Build GRCh38
rs1055954720 Chr.20: 33178372-33178372 on Build GRCh38
rs8125150 Chr.20: 33178095-33178095 on Build GRCh38
rs748022397 Chr.20: 33178280-33178280 on Build GRCh38
rs985808261 Chr.20: 33178464-33178464 on Build GRCh38
rs549553112 Chr.20: 33178369-33178369 on Build GRCh38
rs754953342 Chr.20: 33178272-33178272 on Build GRCh38
rs778504156 Chr.20: 33178273-33178273 on Build GRCh38
rs967445797 Chr.20: 33178405-33178405 on Build GRCh38

View Full Product Details