Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38926234-38926722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83D
Entrez Gene ID
81610
Gene Name
family with sequence similarity 83 member D
Gene Aliases
C20orf129, CHICA, dJ616B8.3
UniGene
Hs.726442
Gene Chromosome Location
Chr.20: 38926312-38953060 on Build GRCh38
Transcripts
NM_030919.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776852 c.106G>C p.E36Q Chr.20: 38926458-38926458 on Build GRCh38
4777094 c.106G>A p.E36K Chr.20: 38926458-38926458 on Build GRCh38
4777095 c.277G>A p.D93N Chr.20: 38926629-38926629 on Build GRCh38
6859452 c.327_328insGCGGCG p.A116_E117insAA Chr.20: 38926679-38926680 on Build GRCh38
7163734 c.18G>C p.L6F Chr.20: 38926370-38926370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772376318 Chr.20: 38926488-38926488 on Build GRCh38
rs770492819 Chr.20: 38926322-38926322 on Build GRCh38
rs758739865 Chr.20: 38926366-38926366 on Build GRCh38
rs931576818 Chr.20: 38926680-38926688 on Build GRCh38
rs766046202 Chr.20: 38926456-38926456 on Build GRCh38
rs746079486 Chr.20: 38926477-38926477 on Build GRCh38
rs778401144 Chr.20: 38926641-38926641 on Build GRCh38
rs984244548 Chr.20: 38926675-38926675 on Build GRCh38
rs556508798 Chr.20: 38926383-38926383 on Build GRCh38
rs774015167 Chr.20: 38926332-38926332 on Build GRCh38
rs567727695 Chr.20: 38926638-38926638 on Build GRCh38
rs778940985 Chr.20: 38926345-38926345 on Build GRCh38
rs748088372 Chr.20: 38926432-38926432 on Build GRCh38
rs758038932 Chr.20: 38926489-38926490 on Build GRCh38
rs200524612 Chr.20: 38926457-38926457 on Build GRCh38
rs779226814 Chr.20: 38926406-38926406 on Build GRCh38
rs777287733 Chr.20: 38926679-38926680 on Build GRCh38
rs953741959 Chr.20: 38926256-38926256 on Build GRCh38
rs761888701 Chr.20: 38926357-38926357 on Build GRCh38
rs973113180 Chr.20: 38926517-38926517 on Build GRCh38
rs746751854 Chr.20: 38926390-38926390 on Build GRCh38
rs752180279 Chr.20: 38926372-38926372 on Build GRCh38
rs748094923 Chr.20: 38926461-38926461 on Build GRCh38
rs371952180 Chr.20: 38926422-38926422 on Build GRCh38
rs773329651 Chr.20: 38926441-38926441 on Build GRCh38
rs777814644 Chr.20: 38926601-38926601 on Build GRCh38
rs747426389 Chr.20: 38926629-38926629 on Build GRCh38
rs371733722 Chr.20: 38926443-38926443 on Build GRCh38
rs866860978 Chr.20: 38926587-38926587 on Build GRCh38
rs768396194 Chr.20: 38926429-38926429 on Build GRCh38
rs751975889 Chr.20: 38926404-38926404 on Build GRCh38
rs778477780 Chr.20: 38926416-38926416 on Build GRCh38
rs779808014 Chr.20: 38926386-38926386 on Build GRCh38
rs573681061 Chr.20: 38926449-38926449 on Build GRCh38
rs912057993 Chr.20: 38926298-38926298 on Build GRCh38
rs536381265 Chr.20: 38926652-38926652 on Build GRCh38
rs777166362 Chr.20: 38926640-38926640 on Build GRCh38
rs755195621 Chr.20: 38926315-38926315 on Build GRCh38
rs542789388 Chr.20: 38926452-38926452 on Build GRCh38
rs1016695822 Chr.20: 38926704-38926704 on Build GRCh38
rs745883040 Chr.20: 38926606-38926606 on Build GRCh38
rs749400749 Chr.20: 38926469-38926469 on Build GRCh38
rs775538014 Chr.20: 38926628-38926628 on Build GRCh38
rs771875218 Chr.20: 38926337-38926337 on Build GRCh38
rs1054546893 Chr.20: 38926579-38926579 on Build GRCh38
rs761824141 Chr.20: 38926399-38926399 on Build GRCh38
rs369962897 Chr.20: 38926442-38926442 on Build GRCh38
rs752565827 Chr.20: 38926578-38926578 on Build GRCh38
rs760027242 Chr.20: 38926446-38926446 on Build GRCh38
rs748747959 Chr.20: 38926318-38926318 on Build GRCh38
rs764449159 Chr.20: 38926451-38926451 on Build GRCh38
rs961782687 Chr.20: 38926674-38926674 on Build GRCh38
rs368203632 Chr.20: 38926349-38926349 on Build GRCh38
rs752268662 Chr.20: 38926669-38926669 on Build GRCh38
rs760879047 Chr.20: 38926496-38926496 on Build GRCh38
rs771111401 Chr.20: 38926470-38926470 on Build GRCh38
rs765413052 Chr.20: 38926358-38926358 on Build GRCh38
rs781044152 Chr.20: 38926431-38926431 on Build GRCh38
rs745671732 Chr.20: 38926336-38926336 on Build GRCh38
rs1007535806 Chr.20: 38926593-38926593 on Build GRCh38
rs895266632 Chr.20: 38926272-38926272 on Build GRCh38
rs987126176 Chr.20: 38926657-38926657 on Build GRCh38
rs559144194 Chr.20: 38926585-38926585 on Build GRCh38
rs749206418 Chr.20: 38926604-38926604 on Build GRCh38
rs778358748 Chr.20: 38926365-38926365 on Build GRCh38
rs952249518 Chr.20: 38926648-38926648 on Build GRCh38
rs747667137 Chr.20: 38926443-38926444 on Build GRCh38
rs767219479 Chr.20: 38926668-38926668 on Build GRCh38
rs757475574 Chr.20: 38926605-38926605 on Build GRCh38
rs6028207 Chr.20: 38926347-38926347 on Build GRCh38
rs775472518 Chr.20: 38926680-38926685 on Build GRCh38
rs970953264 Chr.20: 38926294-38926294 on Build GRCh38
rs760558087 Chr.20: 38926678-38926678 on Build GRCh38
rs112253251 Chr.20: 38926436-38926436 on Build GRCh38
rs768587132 Chr.20: 38926391-38926391 on Build GRCh38
rs758056434 Chr.20: 38926419-38926419 on Build GRCh38
rs1031351803 Chr.20: 38926340-38926340 on Build GRCh38
rs763233937 Chr.20: 38926403-38926403 on Build GRCh38
rs758906450 Chr.20: 38926666-38926666 on Build GRCh38
rs777436878 Chr.20: 38926552-38926557 on Build GRCh38
rs577717767 Chr.20: 38926439-38926439 on Build GRCh38
rs919462045 Chr.20: 38926447-38926447 on Build GRCh38
rs766822255 Chr.20: 38926370-38926370 on Build GRCh38
rs966191661 Chr.20: 38926329-38926329 on Build GRCh38
rs777419039 Chr.20: 38926311-38926311 on Build GRCh38
rs80348623 Chr.20: 38926426-38926426 on Build GRCh38
rs149393382 Chr.20: 38926417-38926417 on Build GRCh38
rs762502120 Chr.20: 38926518-38926518 on Build GRCh38
rs527698652 Chr.20: 38926546-38926546 on Build GRCh38
rs781760146 Chr.20: 38926360-38926360 on Build GRCh38
rs551264776 Chr.20: 38926505-38926505 on Build GRCh38
rs775442870 Chr.20: 38926342-38926342 on Build GRCh38
rs753225581 Chr.20: 38926412-38926412 on Build GRCh38
rs115100890 Chr.20: 38926255-38926255 on Build GRCh38
rs201544468 Chr.20: 38926385-38926385 on Build GRCh38
rs1024830900 Chr.20: 38926275-38926275 on Build GRCh38
rs769896529 Chr.20: 38926400-38926400 on Build GRCh38
rs771352489 Chr.20: 38926592-38926592 on Build GRCh38
rs755554711 Chr.20: 38926373-38926373 on Build GRCh38
rs973345415 Chr.20: 38926676-38926676 on Build GRCh38
rs765847709 Chr.20: 38926540-38926540 on Build GRCh38
rs547544811 Chr.20: 38926614-38926614 on Build GRCh38
rs199696070 Chr.20: 38926512-38926512 on Build GRCh38
rs929768186 Chr.20: 38926338-38926338 on Build GRCh38
rs765932046 Chr.20: 38926647-38926647 on Build GRCh38
rs773347296 Chr.20: 38926402-38926402 on Build GRCh38
rs745498446 Chr.20: 38926384-38926384 on Build GRCh38
rs534813259 Chr.20: 38926682-38926683 on Build GRCh38
rs936080000 Chr.20: 38926569-38926569 on Build GRCh38
rs750863345 Chr.20: 38926392-38926392 on Build GRCh38
rs757868001 Chr.20: 38926454-38926454 on Build GRCh38
rs754613428 Chr.20: 38926552-38926552 on Build GRCh38
rs775934002 Chr.20: 38926493-38926493 on Build GRCh38
rs756093327 Chr.20: 38926464-38926464 on Build GRCh38
rs903049079 Chr.20: 38926568-38926568 on Build GRCh38
rs767021627 Chr.20: 38926565-38926565 on Build GRCh38
rs570408132 Chr.20: 38926680-38926682 on Build GRCh38
rs755815110 Chr.20: 38926588-38926588 on Build GRCh38
rs763792256 Chr.20: 38926688-38926688 on Build GRCh38
rs532282158 Chr.20: 38926405-38926405 on Build GRCh38
rs776957965 Chr.20: 38926513-38926513 on Build GRCh38
rs773865796 Chr.20: 38926655-38926655 on Build GRCh38
rs781735724 Chr.20: 38926382-38926382 on Build GRCh38
rs986431857 Chr.20: 38926260-38926260 on Build GRCh38
rs3752290 Chr.20: 38926473-38926473 on Build GRCh38
rs1012333683 Chr.20: 38926274-38926274 on Build GRCh38
rs375053183 Chr.20: 38926450-38926450 on Build GRCh38
rs756753911 Chr.20: 38926415-38926415 on Build GRCh38
rs777529947 Chr.20: 38926468-38926468 on Build GRCh38
rs753880108 Chr.20: 38926694-38926694 on Build GRCh38
rs747363365 Chr.20: 38926590-38926591 on Build GRCh38

View Full Product Details