Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45409717-45410254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBNDD2
Entrez Gene ID
55861
Gene Name
dysbindin domain containing 2
Gene Aliases
C20orf35, CK1BP, HSMNP1
UniGene
Hs.730643
Gene Chromosome Location
Chr.20: 45405993-45410610 on Build GRCh38
Transcripts
NM_001048221.2 NM_001048222.2 NM_001048223.2 NM_001048224.2 NM_001048225.2 NM_001048226.2 NM_001197139.1 NM_001197140.1 NM_018478.3 NR_003189.2

Gene Symbol
SYS1-DBNDD2
Entrez Gene ID
767557
Gene Name
SYS1-DBNDD2 readthrough (NMD candidate)
Gene Aliases
C20orf169-DBNDD2
UniGene
Hs.730643
Gene Chromosome Location
Chr.20: 45363169-45410610 on Build GRCh38
Transcripts
NM_001048221.2 NM_001048222.2 NM_001048223.2 NM_001048224.2 NM_001048225.2 NM_001048226.2 NM_001197139.1 NM_001197140.1 NM_018478.3 NR_003189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5080370 c.638_640delCCT p.S219delS Chr.20: 45409998-45410000 on Build GRCh38
5468683 c.574A>G p.M192V Chr.20: 45409934-45409934 on Build GRCh38
6332624 c.711G>A p.T237T Chr.20: 45410071-45410071 on Build GRCh38
1578382 c.655_657delTCC p.S219delS Chr.20: 45410015-45410017 on Build GRCh38
1412063 c.606G>A p.P202P Chr.20: 45409966-45409966 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs968526455 Chr.20: 45410199-45410199 on Build GRCh38
rs368669359 Chr.20: 45410028-45410028 on Build GRCh38
rs765712045 Chr.20: 45409935-45409935 on Build GRCh38
rs34250114 Chr.20: 45410003-45410004 on Build GRCh38
rs1127497 Chr.20: 45409934-45409934 on Build GRCh38
rs773129936 Chr.20: 45410200-45410200 on Build GRCh38
rs921353862 Chr.20: 45410126-45410126 on Build GRCh38
rs558880559 Chr.20: 45409744-45409744 on Build GRCh38
rs530082713 Chr.20: 45410169-45410169 on Build GRCh38
rs375779944 Chr.20: 45410040-45410040 on Build GRCh38
rs778502357 Chr.20: 45410101-45410101 on Build GRCh38
rs768829262 Chr.20: 45410071-45410071 on Build GRCh38
rs1013796328 Chr.20: 45409863-45409863 on Build GRCh38
rs745305925 Chr.20: 45410104-45410104 on Build GRCh38
rs747260940 Chr.20: 45410023-45410023 on Build GRCh38
rs764828259 Chr.20: 45410086-45410086 on Build GRCh38
rs370188558 Chr.20: 45409963-45409963 on Build GRCh38
rs759223689 Chr.20: 45410096-45410096 on Build GRCh38
rs199981378 Chr.20: 45410109-45410109 on Build GRCh38
rs980464662 Chr.20: 45409987-45409987 on Build GRCh38
rs775427994 Chr.20: 45410018-45410018 on Build GRCh38
rs538139554 Chr.20: 45409880-45409880 on Build GRCh38
rs932692344 Chr.20: 45410146-45410146 on Build GRCh38
rs745908202 Chr.20: 45409797-45409797 on Build GRCh38
rs776970750 Chr.20: 45410025-45410025 on Build GRCh38
rs771844039 Chr.20: 45409998-45409998 on Build GRCh38
rs969086257 Chr.20: 45409981-45409981 on Build GRCh38
rs372468073 Chr.20: 45410118-45410118 on Build GRCh38
rs1043412769 Chr.20: 45409898-45409898 on Build GRCh38
rs765808639 Chr.20: 45409998-45410006 on Build GRCh38
rs775517761 Chr.20: 45410021-45410021 on Build GRCh38
rs1041182560 Chr.20: 45409821-45409821 on Build GRCh38
rs765388012 Chr.20: 45410043-45410043 on Build GRCh38
rs6130819 Chr.20: 45409804-45409804 on Build GRCh38
rs1031454036 Chr.20: 45410216-45410216 on Build GRCh38
rs943835016 Chr.20: 45409818-45409818 on Build GRCh38
rs184711810 Chr.20: 45409886-45409886 on Build GRCh38
rs373454001 Chr.20: 45409965-45409965 on Build GRCh38
rs753562292 Chr.20: 45410081-45410081 on Build GRCh38
rs1053329491 Chr.20: 45410225-45410225 on Build GRCh38
rs754034228 Chr.20: 45409915-45409915 on Build GRCh38
rs6104147 Chr.20: 45410033-45410033 on Build GRCh38
rs370820232 Chr.20: 45409977-45409977 on Build GRCh38
rs753850586 Chr.20: 45409998-45410000 on Build GRCh38
rs754939513 Chr.20: 45410000-45410001 on Build GRCh38
rs949515647 Chr.20: 45409998-45410003 on Build GRCh38
rs745843780 Chr.20: 45409980-45409980 on Build GRCh38
rs751979023 Chr.20: 45410069-45410069 on Build GRCh38
rs539986667 Chr.20: 45410017-45410017 on Build GRCh38
rs11541738 Chr.20: 45410022-45410022 on Build GRCh38
rs113971375 Chr.20: 45409930-45409930 on Build GRCh38
rs377074151 Chr.20: 45409956-45409956 on Build GRCh38
rs367788273 Chr.20: 45410170-45410170 on Build GRCh38
rs201200240 Chr.20: 45410059-45410059 on Build GRCh38
rs536966139 Chr.20: 45409994-45409994 on Build GRCh38
rs931190334 Chr.20: 45409753-45409753 on Build GRCh38
rs571065417 Chr.20: 45409754-45409754 on Build GRCh38
rs768539476 Chr.20: 45410160-45410160 on Build GRCh38
rs935559107 Chr.20: 45409764-45409764 on Build GRCh38
rs11541735 Chr.20: 45410091-45410091 on Build GRCh38
rs2743273 Chr.20: 45410020-45410020 on Build GRCh38
rs943867666 Chr.20: 45410063-45410063 on Build GRCh38
rs934658222 Chr.20: 45410212-45410212 on Build GRCh38
rs932689991 Chr.20: 45409889-45409889 on Build GRCh38
rs753446087 Chr.20: 45409997-45409998 on Build GRCh38
rs773115877 Chr.20: 45410056-45410056 on Build GRCh38
rs952411581 Chr.20: 45409999-45409999 on Build GRCh38
rs374150638 Chr.20: 45409979-45409979 on Build GRCh38
rs1052705896 Chr.20: 45409775-45409775 on Build GRCh38
rs1045174034 Chr.20: 45409874-45409874 on Build GRCh38
rs113063391 Chr.20: 45410105-45410105 on Build GRCh38
rs201185742 Chr.20: 45410070-45410070 on Build GRCh38
rs368600687 Chr.20: 45410186-45410187 on Build GRCh38
rs1006269923 Chr.20: 45410036-45410036 on Build GRCh38
rs748849184 Chr.20: 45409976-45409976 on Build GRCh38
rs112151458 Chr.20: 45409931-45409931 on Build GRCh38
rs758789226 Chr.20: 45409946-45409946 on Build GRCh38
rs766869038 Chr.20: 45410061-45410061 on Build GRCh38
rs562823941 Chr.20: 45410127-45410127 on Build GRCh38
rs897810573 Chr.20: 45410064-45410064 on Build GRCh38
rs374582383 Chr.20: 45410085-45410085 on Build GRCh38
rs746846826 Chr.20: 45410133-45410133 on Build GRCh38
rs1016651014 Chr.20: 45409895-45409895 on Build GRCh38
rs760961546 Chr.20: 45410011-45410019 on Build GRCh38
rs535857781 Chr.20: 45409942-45409942 on Build GRCh38
rs376399077 Chr.20: 45409966-45409966 on Build GRCh38
rs1037796739 Chr.20: 45410042-45410042 on Build GRCh38

View Full Product Details