Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46371320-46371820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO2
Entrez Gene ID
63916
Gene Name
engulfment and cell motility 2
Gene Aliases
CED-12, CED12, Ced-12A, ELMO-2
UniGene
Hs.210469
Gene Chromosome Location
Chr.20: 46366045-46407051 on Build GRCh38
Transcripts
NM_001318253.1 NM_133171.4 NM_182764.2 XM_005260496.3 XM_005260498.3 XM_005260499.3 XM_005260500.3 XM_005260501.4 XM_006723854.3 XM_017028009.1 XM_017028010.1 XM_017028011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680794 c.1652A>G p.E551G Chr.20: 46371620-46371620 on Build GRCh38
3701633 c.1801A>G p.I601V Chr.20: 46371352-46371352 on Build GRCh38
5726588 c.1770G>A p.G590G Chr.20: 46371383-46371383 on Build GRCh38
7186577 c.1684C>A p.R562R Chr.20: 46371588-46371588 on Build GRCh38
3294517 c.1572G>A p.P524P Chr.20: 46371814-46371814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779413222 Chr.20: 46371421-46371421 on Build GRCh38
rs949109192 Chr.20: 46371494-46371494 on Build GRCh38
rs774341686 Chr.20: 46371546-46371546 on Build GRCh38
rs961394374 Chr.20: 46371753-46371753 on Build GRCh38
rs899038794 Chr.20: 46371576-46371576 on Build GRCh38
rs752380720 Chr.20: 46371393-46371393 on Build GRCh38
rs755062291 Chr.20: 46371652-46371652 on Build GRCh38
rs546819827 Chr.20: 46371778-46371778 on Build GRCh38
rs754184468 Chr.20: 46371397-46371397 on Build GRCh38
rs763747637 Chr.20: 46371567-46371567 on Build GRCh38
rs781290331 Chr.20: 46371505-46371505 on Build GRCh38
rs764520008 Chr.20: 46371396-46371396 on Build GRCh38
rs775583783 Chr.20: 46371694-46371694 on Build GRCh38
rs372963438 Chr.20: 46371587-46371587 on Build GRCh38
rs778618407 Chr.20: 46371680-46371680 on Build GRCh38
rs570435889 Chr.20: 46371699-46371699 on Build GRCh38
rs954941889 Chr.20: 46371614-46371614 on Build GRCh38
rs972450025 Chr.20: 46371760-46371760 on Build GRCh38
rs757981591 Chr.20: 46371719-46371719 on Build GRCh38
rs757578934 Chr.20: 46371411-46371411 on Build GRCh38
rs1016697465 Chr.20: 46371723-46371723 on Build GRCh38
rs766811805 Chr.20: 46371781-46371781 on Build GRCh38
rs762231531 Chr.20: 46371773-46371774 on Build GRCh38
rs149080790 Chr.20: 46371687-46371687 on Build GRCh38
rs369324348 Chr.20: 46371549-46371549 on Build GRCh38
rs768828164 Chr.20: 46371498-46371498 on Build GRCh38
rs767962684 Chr.20: 46371790-46371791 on Build GRCh38
rs752937635 Chr.20: 46371795-46371795 on Build GRCh38
rs138931316 Chr.20: 46371385-46371386 on Build GRCh38
rs767551212 Chr.20: 46371788-46371788 on Build GRCh38
rs143362697 Chr.20: 46371726-46371727 on Build GRCh38
rs750963456 Chr.20: 46371456-46371456 on Build GRCh38
rs773794464 Chr.20: 46371791-46371791 on Build GRCh38
rs757057713 Chr.20: 46371679-46371679 on Build GRCh38
rs2297052 Chr.20: 46371783-46371783 on Build GRCh38
rs199553318 Chr.20: 46371718-46371718 on Build GRCh38
rs758944976 Chr.20: 46371604-46371604 on Build GRCh38
rs749053010 Chr.20: 46371533-46371533 on Build GRCh38
rs746456458 Chr.20: 46371706-46371706 on Build GRCh38
rs760241662 Chr.20: 46371696-46371696 on Build GRCh38
rs753440055 Chr.20: 46371575-46371575 on Build GRCh38
rs533065163 Chr.20: 46371392-46371392 on Build GRCh38
rs780571140 Chr.20: 46371605-46371605 on Build GRCh38
rs147623211 Chr.20: 46371493-46371493 on Build GRCh38
rs745766574 Chr.20: 46371685-46371685 on Build GRCh38
rs1008961827 Chr.20: 46371636-46371636 on Build GRCh38
rs913674833 Chr.20: 46371707-46371707 on Build GRCh38
rs762689956 Chr.20: 46371344-46371344 on Build GRCh38
rs142132720 Chr.20: 46371649-46371649 on Build GRCh38
rs926280664 Chr.20: 46371732-46371732 on Build GRCh38
rs985623713 Chr.20: 46371399-46371399 on Build GRCh38
rs781407932 Chr.20: 46371667-46371667 on Build GRCh38
rs780157170 Chr.20: 46371490-46371490 on Build GRCh38
rs370879690 Chr.20: 46371779-46371779 on Build GRCh38
rs775154225 Chr.20: 46371563-46371563 on Build GRCh38
rs748385528 Chr.20: 46371674-46371674 on Build GRCh38
rs748841493 Chr.20: 46371710-46371712 on Build GRCh38
rs778560733 Chr.20: 46371666-46371666 on Build GRCh38
rs370328364 Chr.20: 46371566-46371566 on Build GRCh38
rs756717915 Chr.20: 46371400-46371400 on Build GRCh38
rs759601387 Chr.20: 46371547-46371547 on Build GRCh38
rs569684760 Chr.20: 46371537-46371537 on Build GRCh38
rs752065089 Chr.20: 46371782-46371782 on Build GRCh38
rs765582782 Chr.20: 46371588-46371588 on Build GRCh38
rs760450272 Chr.20: 46371394-46371394 on Build GRCh38
rs758459502 Chr.20: 46371471-46371471 on Build GRCh38
rs774615848 Chr.20: 46371733-46371733 on Build GRCh38
rs761421278 Chr.20: 46371730-46371730 on Build GRCh38
rs374134295 Chr.20: 46371362-46371362 on Build GRCh38
rs548986105 Chr.20: 46371560-46371560 on Build GRCh38
rs750919846 Chr.20: 46371590-46371590 on Build GRCh38
rs770851300 Chr.20: 46371358-46371358 on Build GRCh38
rs201627182 Chr.20: 46371562-46371562 on Build GRCh38

View Full Product Details