Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46371664-46372160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO2
Entrez Gene ID
63916
Gene Name
engulfment and cell motility 2
Gene Aliases
CED-12, CED12, Ced-12A, ELMO-2
UniGene
Hs.210469
Gene Chromosome Location
Chr.20: 46366045-46407051 on Build GRCh38
Transcripts
NM_001318253.1 NM_133171.4 NM_182764.2 XM_005260496.3 XM_005260498.3 XM_005260499.3 XM_005260500.3 XM_005260501.4 XM_006723854.3 XM_017028009.1 XM_017028010.1 XM_017028011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4099001 c.1532G>A p.R511H Chr.20: 46371854-46371854 on Build GRCh38
724377 c.1499G>T p.R500L Chr.20: 46371887-46371887 on Build GRCh38
6378397 c.1535A>T p.Q512L Chr.20: 46371851-46371851 on Build GRCh38
6441079 c.1540G>A p.E514K Chr.20: 46371846-46371846 on Build GRCh38
3294517 c.1572G>A p.P524P Chr.20: 46371814-46371814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565489343 Chr.20: 46372086-46372086 on Build GRCh38
rs747312721 Chr.20: 46371921-46371921 on Build GRCh38
rs778085258 Chr.20: 46371811-46371811 on Build GRCh38
rs961394374 Chr.20: 46371753-46371753 on Build GRCh38
rs896931179 Chr.20: 46372131-46372131 on Build GRCh38
rs756238676 Chr.20: 46371913-46371913 on Build GRCh38
rs546819827 Chr.20: 46371778-46371778 on Build GRCh38
rs769284976 Chr.20: 46371871-46371871 on Build GRCh38
rs752677688 Chr.20: 46371905-46371905 on Build GRCh38
rs775583783 Chr.20: 46371694-46371694 on Build GRCh38
rs767601980 Chr.20: 46371898-46371898 on Build GRCh38
rs1005401997 Chr.20: 46372100-46372100 on Build GRCh38
rs570435889 Chr.20: 46371699-46371699 on Build GRCh38
rs776049108 Chr.20: 46371862-46371862 on Build GRCh38
rs772866712 Chr.20: 46371872-46371872 on Build GRCh38
rs748257472 Chr.20: 46372072-46372072 on Build GRCh38
rs747557564 Chr.20: 46371957-46371957 on Build GRCh38
rs760009580 Chr.20: 46371887-46371887 on Build GRCh38
rs777234620 Chr.20: 46372051-46372051 on Build GRCh38
rs972450025 Chr.20: 46371760-46371760 on Build GRCh38
rs774822992 Chr.20: 46371879-46371879 on Build GRCh38
rs748994312 Chr.20: 46371990-46371990 on Build GRCh38
rs757981591 Chr.20: 46371719-46371719 on Build GRCh38
rs151220651 Chr.20: 46372039-46372039 on Build GRCh38
rs1016697465 Chr.20: 46371723-46371723 on Build GRCh38
rs575255605 Chr.20: 46371953-46371953 on Build GRCh38
rs766811805 Chr.20: 46371781-46371781 on Build GRCh38
rs370937421 Chr.20: 46371919-46371919 on Build GRCh38
rs762231531 Chr.20: 46371773-46371774 on Build GRCh38
rs149080790 Chr.20: 46371687-46371687 on Build GRCh38
rs767962684 Chr.20: 46371790-46371791 on Build GRCh38
rs752937635 Chr.20: 46371795-46371795 on Build GRCh38
rs917414681 Chr.20: 46372098-46372101 on Build GRCh38
rs767551212 Chr.20: 46371788-46371788 on Build GRCh38
rs1017648369 Chr.20: 46371972-46371972 on Build GRCh38
rs766793261 Chr.20: 46371878-46371878 on Build GRCh38
rs971550865 Chr.20: 46372107-46372107 on Build GRCh38
rs143362697 Chr.20: 46371726-46371727 on Build GRCh38
rs773794464 Chr.20: 46371791-46371791 on Build GRCh38
rs985676110 Chr.20: 46371810-46371810 on Build GRCh38
rs540426658 Chr.20: 46372084-46372084 on Build GRCh38
rs2297052 Chr.20: 46371783-46371783 on Build GRCh38
rs771259016 Chr.20: 46371994-46371994 on Build GRCh38
rs768190847 Chr.20: 46371860-46371860 on Build GRCh38
rs199553318 Chr.20: 46371718-46371718 on Build GRCh38
rs746456458 Chr.20: 46371706-46371706 on Build GRCh38
rs760241662 Chr.20: 46371696-46371696 on Build GRCh38
rs774737378 Chr.20: 46371995-46371995 on Build GRCh38
rs747728081 Chr.20: 46371864-46371864 on Build GRCh38
rs964718959 Chr.20: 46371974-46371976 on Build GRCh38
rs376458042 Chr.20: 46371816-46371816 on Build GRCh38
rs745766574 Chr.20: 46371685-46371685 on Build GRCh38
rs913674833 Chr.20: 46371707-46371707 on Build GRCh38
rs577781881 Chr.20: 46372091-46372091 on Build GRCh38
rs759975632 Chr.20: 46371996-46371996 on Build GRCh38
rs1017160295 Chr.20: 46372113-46372113 on Build GRCh38
rs189560449 Chr.20: 46372055-46372055 on Build GRCh38
rs185032247 Chr.20: 46372028-46372028 on Build GRCh38
rs926280664 Chr.20: 46371732-46371732 on Build GRCh38
rs895029663 Chr.20: 46372097-46372097 on Build GRCh38
rs370879690 Chr.20: 46371779-46371779 on Build GRCh38
rs375449065 Chr.20: 46371940-46371940 on Build GRCh38
rs993357068 Chr.20: 46371823-46371823 on Build GRCh38
rs748841493 Chr.20: 46371710-46371712 on Build GRCh38
rs563402926 Chr.20: 46372116-46372116 on Build GRCh38
rs763234588 Chr.20: 46371877-46371877 on Build GRCh38
rs201222160 Chr.20: 46371942-46371942 on Build GRCh38
rs756327149 Chr.20: 46371801-46371801 on Build GRCh38
rs574674005 Chr.20: 46371916-46371916 on Build GRCh38
rs545149143 Chr.20: 46372093-46372093 on Build GRCh38
rs747002155 Chr.20: 46371850-46371850 on Build GRCh38
rs982310032 Chr.20: 46372118-46372118 on Build GRCh38
rs752065089 Chr.20: 46371782-46371782 on Build GRCh38
rs538756608 Chr.20: 46371820-46371820 on Build GRCh38
rs994313779 Chr.20: 46372133-46372133 on Build GRCh38
rs774615848 Chr.20: 46371733-46371733 on Build GRCh38
rs780136305 Chr.20: 46371847-46371847 on Build GRCh38
rs761421278 Chr.20: 46371730-46371730 on Build GRCh38
rs745607349 Chr.20: 46371815-46371815 on Build GRCh38
rs769349670 Chr.20: 46371973-46371973 on Build GRCh38
rs1033251803 Chr.20: 46372062-46372062 on Build GRCh38

View Full Product Details