Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46894524-46895062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA2
Entrez Gene ID
2139
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
EAB1
UniGene
Hs.472877
Gene Chromosome Location
Chr.20: 46894624-47188844 on Build GRCh38
Transcripts
NM_005244.4 NM_172110.3 XM_005260327.2 XM_017027721.1

SNP Detail

SNP ID Chromosome Location
rs945244382 Chr.20: 46894845-46894845 on Build GRCh38
rs575347855 Chr.20: 46894878-46894878 on Build GRCh38
rs957491871 Chr.20: 46894819-46894819 on Build GRCh38
rs13043501 Chr.20: 46894668-46894668 on Build GRCh38
rs112705259 Chr.20: 46894932-46894932 on Build GRCh38
rs983553847 Chr.20: 46894581-46894581 on Build GRCh38
rs564698072 Chr.20: 46894929-46894929 on Build GRCh38
rs556717726 Chr.20: 46894698-46894698 on Build GRCh38
rs922080250 Chr.20: 46894636-46894636 on Build GRCh38
rs190766608 Chr.20: 46894656-46894656 on Build GRCh38
rs1004544865 Chr.20: 46894899-46894899 on Build GRCh38
rs894857952 Chr.20: 46894882-46894882 on Build GRCh38
rs1042318500 Chr.20: 46894847-46894847 on Build GRCh38
rs974906109 Chr.20: 46894799-46894799 on Build GRCh38
rs932000671 Chr.20: 46894615-46894615 on Build GRCh38
rs957647329 Chr.20: 46894938-46894941 on Build GRCh38
rs1040376285 Chr.20: 46894609-46894609 on Build GRCh38
rs1027333915 Chr.20: 46894755-46894755 on Build GRCh38
rs1031598586 Chr.20: 46894641-46894641 on Build GRCh38
rs529837200 Chr.20: 46895007-46895007 on Build GRCh38
rs533914123 Chr.20: 46894582-46894582 on Build GRCh38
rs994549991 Chr.20: 46894545-46894545 on Build GRCh38
rs1020824121 Chr.20: 46894964-46894964 on Build GRCh38
rs904102903 Chr.20: 46894628-46894628 on Build GRCh38
rs1054875462 Chr.20: 46894718-46894718 on Build GRCh38
rs550561620 Chr.20: 46894969-46894969 on Build GRCh38
rs572903570 Chr.20: 46894849-46894849 on Build GRCh38
rs13043776 Chr.20: 46894669-46894669 on Build GRCh38
rs11700141 Chr.20: 46895000-46895000 on Build GRCh38
rs540250020 Chr.20: 46894874-46894874 on Build GRCh38
rs902133125 Chr.20: 46894918-46894918 on Build GRCh38
rs542784017 Chr.20: 46894735-46894735 on Build GRCh38
rs997105975 Chr.20: 46895010-46895010 on Build GRCh38
rs927851186 Chr.20: 46894971-46894971 on Build GRCh38
rs879913925 Chr.20: 46894668-46894669 on Build GRCh38
rs951025146 Chr.20: 46894566-46894566 on Build GRCh38
rs560962767 Chr.20: 46894767-46894767 on Build GRCh38
rs939261242 Chr.20: 46894972-46894972 on Build GRCh38
rs886113785 Chr.20: 46894892-46894892 on Build GRCh38
rs1026864406 Chr.20: 46895039-46895039 on Build GRCh38
rs969017652 Chr.20: 46895019-46895019 on Build GRCh38
rs966178882 Chr.20: 46894791-46894791 on Build GRCh38
rs996272586 Chr.20: 46894925-46894925 on Build GRCh38
rs1042208548 Chr.20: 46894686-46894686 on Build GRCh38
rs36023289 Chr.20: 46894668-46894669 on Build GRCh38
rs566577160 Chr.20: 46894565-46894565 on Build GRCh38
rs940770058 Chr.20: 46894589-46894589 on Build GRCh38
rs952748578 Chr.20: 46895040-46895040 on Build GRCh38
rs906374354 Chr.20: 46894697-46894697 on Build GRCh38
rs892191393 Chr.20: 46894646-46894646 on Build GRCh38
rs945333840 Chr.20: 46894679-46894679 on Build GRCh38
rs1005093493 Chr.20: 46894777-46894777 on Build GRCh38
rs921998958 Chr.20: 46894801-46894801 on Build GRCh38
rs1019035970 Chr.20: 46894784-46894784 on Build GRCh38
rs1050695154 Chr.20: 46894625-46894625 on Build GRCh38
rs765746486 Chr.20: 46894612-46894612 on Build GRCh38
rs990535046 Chr.20: 46894824-46894824 on Build GRCh38
rs939148095 Chr.20: 46894857-46894857 on Build GRCh38
rs990260861 Chr.20: 46894940-46894940 on Build GRCh38
rs113583272 Chr.20: 46895023-46895023 on Build GRCh38
rs1043785514 Chr.20: 46894786-46894786 on Build GRCh38
rs1019137985 Chr.20: 46894908-46894908 on Build GRCh38
rs1001005849 Chr.20: 46894637-46894637 on Build GRCh38
rs913236694 Chr.20: 46894831-46894831 on Build GRCh38
rs112479792 Chr.20: 46894853-46894853 on Build GRCh38
rs532440232 Chr.20: 46894933-46894933 on Build GRCh38
rs1049165323 Chr.20: 46894883-46894883 on Build GRCh38
rs915116582 Chr.20: 46894674-46894674 on Build GRCh38
rs574894302 Chr.20: 46894734-46894734 on Build GRCh38

View Full Product Details