Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58155376-58155867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf85
Entrez Gene ID
128602
Gene Name
chromosome 20 open reading frame 85
Gene Aliases
LLC1, bA196N14.1
UniGene
Hs.43977
Gene Chromosome Location
Chr.20: 58150927-58161130 on Build GRCh38
Transcripts
NM_178456.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1028435 c.151-2A>C p.? Chr.20: 58155466-58155466 on Build GRCh38
4293454 c.209G>A p.R70H Chr.20: 58155526-58155526 on Build GRCh38
227396 c.181C>T p.P61S Chr.20: 58155498-58155498 on Build GRCh38
6539472 c.223C>G p.P75A Chr.20: 58155540-58155540 on Build GRCh38
4099750 c.225G>A p.P75P Chr.20: 58155542-58155542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139141770 Chr.20: 58155563-58155563 on Build GRCh38
rs767638835 Chr.20: 58155418-58155418 on Build GRCh38
rs769281185 Chr.20: 58155579-58155579 on Build GRCh38
rs914041639 Chr.20: 58155719-58155719 on Build GRCh38
rs796802518 Chr.20: 58155688-58155688 on Build GRCh38
rs754786080 Chr.20: 58155490-58155490 on Build GRCh38
rs763938254 Chr.20: 58155537-58155537 on Build GRCh38
rs573067979 Chr.20: 58155798-58155798 on Build GRCh38
rs545870632 Chr.20: 58155727-58155727 on Build GRCh38
rs762736386 Chr.20: 58155675-58155675 on Build GRCh38
rs761997914 Chr.20: 58155447-58155447 on Build GRCh38
rs557839246 Chr.20: 58155526-58155526 on Build GRCh38
rs568065621 Chr.20: 58155542-58155542 on Build GRCh38
rs141491612 Chr.20: 58155538-58155538 on Build GRCh38
rs917259953 Chr.20: 58155846-58155846 on Build GRCh38
rs778745684 Chr.20: 58155564-58155565 on Build GRCh38
rs931762153 Chr.20: 58155776-58155776 on Build GRCh38
rs184379210 Chr.20: 58155578-58155578 on Build GRCh38
rs750463707 Chr.20: 58155453-58155453 on Build GRCh38
rs983682511 Chr.20: 58155842-58155842 on Build GRCh38
rs775964835 Chr.20: 58155433-58155433 on Build GRCh38
rs763088623 Chr.20: 58155607-58155607 on Build GRCh38
rs777411777 Chr.20: 58155483-58155483 on Build GRCh38
rs144357163 Chr.20: 58155468-58155468 on Build GRCh38
rs765523837 Chr.20: 58155513-58155513 on Build GRCh38
rs922998532 Chr.20: 58155775-58155775 on Build GRCh38
rs748885829 Chr.20: 58155485-58155485 on Build GRCh38
rs1027812104 Chr.20: 58155807-58155807 on Build GRCh38
rs969466682 Chr.20: 58155667-58155667 on Build GRCh38
rs752555419 Chr.20: 58155476-58155476 on Build GRCh38
rs969350484 Chr.20: 58155411-58155411 on Build GRCh38
rs995440444 Chr.20: 58155743-58155743 on Build GRCh38
rs893743898 Chr.20: 58155617-58155617 on Build GRCh38
rs758360857 Chr.20: 58155593-58155594 on Build GRCh38
rs1053607091 Chr.20: 58155608-58155608 on Build GRCh38
rs770862872 Chr.20: 58155610-58155610 on Build GRCh38
rs752670167 Chr.20: 58155584-58155585 on Build GRCh38
rs535081654 Chr.20: 58155600-58155600 on Build GRCh38
rs773388585 Chr.20: 58155525-58155525 on Build GRCh38
rs550982937 Chr.20: 58155438-58155438 on Build GRCh38
rs745341135 Chr.20: 58155494-58155494 on Build GRCh38
rs200941765 Chr.20: 58155509-58155509 on Build GRCh38
rs958510986 Chr.20: 58155690-58155690 on Build GRCh38
rs750653709 Chr.20: 58155480-58155482 on Build GRCh38
rs773263421 Chr.20: 58155601-58155601 on Build GRCh38
rs564223024 Chr.20: 58155765-58155765 on Build GRCh38
rs35471857 Chr.20: 58155624-58155625 on Build GRCh38
rs370521824 Chr.20: 58155435-58155435 on Build GRCh38
rs777122137 Chr.20: 58155512-58155512 on Build GRCh38
rs540468313 Chr.20: 58155825-58155825 on Build GRCh38
rs760676033 Chr.20: 58155620-58155620 on Build GRCh38
rs765463981 Chr.20: 58155448-58155448 on Build GRCh38
rs778409751 Chr.20: 58155551-58155551 on Build GRCh38
rs1050489964 Chr.20: 58155815-58155815 on Build GRCh38
rs755821115 Chr.20: 58155478-58155478 on Build GRCh38
rs373651344 Chr.20: 58155466-58155466 on Build GRCh38
rs142992497 Chr.20: 58155619-58155619 on Build GRCh38
rs753726968 Chr.20: 58155666-58155666 on Build GRCh38
rs1008610408 Chr.20: 58155683-58155683 on Build GRCh38
rs756893215 Chr.20: 58155489-58155489 on Build GRCh38
rs780623678 Chr.20: 58155569-58155569 on Build GRCh38
rs759897050 Chr.20: 58155595-58155595 on Build GRCh38
rs769083376 Chr.20: 58155510-58155510 on Build GRCh38
rs988415670 Chr.20: 58155718-58155718 on Build GRCh38
rs778464611 Chr.20: 58155491-58155491 on Build GRCh38
rs148318638 Chr.20: 58155532-58155532 on Build GRCh38
rs529257096 Chr.20: 58155434-58155434 on Build GRCh38
rs929558898 Chr.20: 58155503-58155503 on Build GRCh38
rs532382153 Chr.20: 58155487-58155487 on Build GRCh38
rs887453119 Chr.20: 58155837-58155837 on Build GRCh38
rs772686114 Chr.20: 58155498-58155498 on Build GRCh38
rs561813900 Chr.20: 58155834-58155834 on Build GRCh38
rs575144100 Chr.20: 58155650-58155650 on Build GRCh38
rs550650793 Chr.20: 58155832-58155832 on Build GRCh38
rs757806142 Chr.20: 58155564-58155564 on Build GRCh38
rs568385765 Chr.20: 58155397-58155397 on Build GRCh38

View Full Product Details