Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59324249-59324748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDN3
Entrez Gene ID
1908
Gene Name
endothelin 3
Gene Aliases
ET-3, ET3, HSCR4, PPET3, WS4B
UniGene
Hs.1408
Gene Chromosome Location
Chr.20: 59300415-59325992 on Build GRCh38
Transcripts
NM_001302455.1 NM_001302456.1 NM_207032.2 NM_207033.2 NM_207034.2 XM_005260312.4 XM_005260313.4 XM_006723734.3 XM_011528655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3707887 c.585C>T p.D195D Chr.20: 59324369-59324369 on Build GRCh38
1483796 c.633C>T p.H211H Chr.20: 59324375-59324375 on Build GRCh38
5576611 c.666C>T p.A222A Chr.20: 59324408-59324408 on Build GRCh38
1592679 c.589-6A>G p.? Chr.20: 59324325-59324325 on Build GRCh38
5223950 c.591C>T p.H197H Chr.20: 59324375-59324375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953126688 Chr.20: 59324599-59324599 on Build GRCh38
rs144250756 Chr.20: 59324431-59324431 on Build GRCh38
rs374488792 Chr.20: 59324487-59324487 on Build GRCh38
rs377287357 Chr.20: 59324289-59324289 on Build GRCh38
rs892710743 Chr.20: 59324699-59324699 on Build GRCh38
rs545057062 Chr.20: 59324287-59324287 on Build GRCh38
rs561456608 Chr.20: 59324312-59324312 on Build GRCh38
rs567494206 Chr.20: 59324502-59324502 on Build GRCh38
rs746504168 Chr.20: 59324700-59324700 on Build GRCh38
rs11570352 Chr.20: 59324605-59324605 on Build GRCh38
rs779040805 Chr.20: 59324337-59324337 on Build GRCh38
rs370508486 Chr.20: 59324411-59324411 on Build GRCh38
rs376137509 Chr.20: 59324459-59324459 on Build GRCh38
rs886056876 Chr.20: 59324597-59324598 on Build GRCh38
rs1056166329 Chr.20: 59324687-59324687 on Build GRCh38
rs372569173 Chr.20: 59324601-59324601 on Build GRCh38
rs374180061 Chr.20: 59324314-59324314 on Build GRCh38
rs925245673 Chr.20: 59324629-59324629 on Build GRCh38
rs463415 Chr.20: 59324517-59324517 on Build GRCh38
rs3026576 Chr.20: 59324611-59324611 on Build GRCh38
rs768584962 Chr.20: 59324277-59324277 on Build GRCh38
rs745960429 Chr.20: 59324342-59324342 on Build GRCh38
rs571072081 Chr.20: 59324537-59324537 on Build GRCh38
rs138451394 Chr.20: 59324460-59324460 on Build GRCh38
rs756038575 Chr.20: 59324441-59324441 on Build GRCh38
rs1053285924 Chr.20: 59324596-59324596 on Build GRCh38
rs754204521 Chr.20: 59324417-59324417 on Build GRCh38
rs970107700 Chr.20: 59324545-59324545 on Build GRCh38
rs377130048 Chr.20: 59324604-59324605 on Build GRCh38
rs979253286 Chr.20: 59324551-59324551 on Build GRCh38
rs370288393 Chr.20: 59324477-59324477 on Build GRCh38
rs1004679269 Chr.20: 59324466-59324466 on Build GRCh38
rs764620452 Chr.20: 59324415-59324415 on Build GRCh38
rs749112686 Chr.20: 59324449-59324449 on Build GRCh38
rs201352056 Chr.20: 59324406-59324406 on Build GRCh38
rs886056879 Chr.20: 59324607-59324608 on Build GRCh38
rs549116987 Chr.20: 59324439-59324439 on Build GRCh38
rs896308632 Chr.20: 59324688-59324688 on Build GRCh38
rs200182949 Chr.20: 59324296-59324297 on Build GRCh38
rs553876011 Chr.20: 59324552-59324552 on Build GRCh38
rs748005608 Chr.20: 59324393-59324393 on Build GRCh38
rs774182574 Chr.20: 59324453-59324453 on Build GRCh38
rs909168391 Chr.20: 59324361-59324361 on Build GRCh38
rs199781865 Chr.20: 59324321-59324321 on Build GRCh38
rs972033394 Chr.20: 59324604-59324604 on Build GRCh38
rs542985516 Chr.20: 59324311-59324311 on Build GRCh38
rs941911524 Chr.20: 59324389-59324389 on Build GRCh38
rs11570350 Chr.20: 59324291-59324291 on Build GRCh38
rs1030078945 Chr.20: 59324594-59324594 on Build GRCh38
rs548702985 Chr.20: 59324664-59324664 on Build GRCh38
rs760163643 Chr.20: 59324473-59324473 on Build GRCh38
rs748824746 Chr.20: 59324327-59324327 on Build GRCh38
rs527379112 Chr.20: 59324638-59324638 on Build GRCh38
rs757499758 Chr.20: 59324332-59324332 on Build GRCh38
rs886056878 Chr.20: 59324598-59324599 on Build GRCh38
rs1051847553 Chr.20: 59324718-59324718 on Build GRCh38
rs11570351 Chr.20: 59324412-59324412 on Build GRCh38
rs886056881 Chr.20: 59324604-59324604 on Build GRCh38
rs1045052631 Chr.20: 59324479-59324479 on Build GRCh38
rs751148274 Chr.20: 59324300-59324300 on Build GRCh38
rs998077490 Chr.20: 59324555-59324555 on Build GRCh38
rs762674210 Chr.20: 59324294-59324294 on Build GRCh38
rs886056875 Chr.20: 59324555-59324555 on Build GRCh38
rs532030990 Chr.20: 59324397-59324397 on Build GRCh38
rs754876648 Chr.20: 59324600-59324600 on Build GRCh38
rs71321536 Chr.20: 59324602-59324602 on Build GRCh38
rs576673862 Chr.20: 59324606-59324606 on Build GRCh38
rs555946641 Chr.20: 59324524-59324524 on Build GRCh38
rs750694380 Chr.20: 59324495-59324495 on Build GRCh38
rs752653775 Chr.20: 59324599-59324600 on Build GRCh38
rs1012117157 Chr.20: 59324691-59324691 on Build GRCh38
rs767213063 Chr.20: 59324444-59324444 on Build GRCh38
rs997731228 Chr.20: 59324706-59324706 on Build GRCh38
rs758459588 Chr.20: 59324345-59324345 on Build GRCh38
rs11570354 Chr.20: 59324636-59324636 on Build GRCh38
rs926092698 Chr.20: 59324276-59324276 on Build GRCh38
rs774891807 Chr.20: 59324469-59324469 on Build GRCh38
rs372958987 Chr.20: 59324430-59324430 on Build GRCh38
rs756538873 Chr.20: 59324313-59324313 on Build GRCh38
rs918012816 Chr.20: 59324610-59324610 on Build GRCh38
rs572322014 Chr.20: 59324298-59324298 on Build GRCh38
rs11570353 Chr.20: 59324624-59324624 on Build GRCh38
rs750823872 Chr.20: 59324494-59324494 on Build GRCh38
rs11475273 Chr.20: 59324599-59324599 on Build GRCh38
rs773949944 Chr.20: 59324297-59324297 on Build GRCh38
rs544095029 Chr.20: 59324607-59324607 on Build GRCh38
rs137985872 Chr.20: 59324405-59324405 on Build GRCh38
rs777109038 Chr.20: 59324485-59324485 on Build GRCh38
rs757559436 Chr.20: 59324427-59324427 on Build GRCh38
rs565501608 Chr.20: 59324608-59324608 on Build GRCh38
rs751597991 Chr.20: 59324506-59324506 on Build GRCh38
rs761573030 Chr.20: 59324369-59324371 on Build GRCh38
rs368688196 Chr.20: 59324488-59324488 on Build GRCh38
rs200741552 Chr.20: 59324419-59324419 on Build GRCh38
rs538768879 Chr.20: 59324540-59324540 on Build GRCh38
rs770841292 Chr.20: 59324452-59324452 on Build GRCh38
rs145582774 Chr.20: 59324341-59324341 on Build GRCh38
rs777702187 Chr.20: 59324445-59324445 on Build GRCh38
rs765500226 Chr.20: 59324429-59324429 on Build GRCh38
rs772906477 Chr.20: 59324394-59324394 on Build GRCh38
rs781257967 Chr.20: 59324372-59324372 on Build GRCh38
rs886056880 Chr.20: 59324603-59324603 on Build GRCh38
rs963636691 Chr.20: 59324603-59324603 on Build GRCh38
rs886056877 Chr.20: 59324598-59324598 on Build GRCh38
rs200872063 Chr.20: 59324503-59324503 on Build GRCh38
rs3026575 Chr.20: 59324630-59324630 on Build GRCh38
rs761106361 Chr.20: 59324413-59324413 on Build GRCh38
rs397866060 Chr.20: 59324608-59324608 on Build GRCh38
rs776373684 Chr.20: 59324285-59324285 on Build GRCh38

View Full Product Details