Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59984547-59985045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH26
Entrez Gene ID
60437
Gene Name
cadherin 26
Gene Aliases
VR20
UniGene
Hs.729046
Gene Chromosome Location
Chr.20: 59957764-60035629 on Build GRCh38
Transcripts
NM_177980.2 XM_011528969.2 XM_017027994.1 XM_017027995.1 XM_017027996.1 XM_017027997.1 XM_017027998.1 XM_017027999.1 XM_017028000.1 XM_017028001.1 XM_017028002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6160353 c.711C>T p.T237T Chr.20: 59985003-59985003 on Build GRCh38
4099920 c.622T>C p.S208P Chr.20: 59984719-59984719 on Build GRCh38
192812 c.682C>T p.R228* Chr.20: 59984779-59984779 on Build GRCh38
4977216 c.544C>A p.Q182K Chr.20: 59984641-59984641 on Build GRCh38
1028666 c.623C>A p.S208Y Chr.20: 59984720-59984720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150599622 Chr.20: 59984631-59984631 on Build GRCh38
rs1007472835 Chr.20: 59984732-59984732 on Build GRCh38
rs745666352 Chr.20: 59984635-59984635 on Build GRCh38
rs762497452 Chr.20: 59984768-59984768 on Build GRCh38
rs147941031 Chr.20: 59984753-59984753 on Build GRCh38
rs537999388 Chr.20: 59984730-59984730 on Build GRCh38
rs748377796 Chr.20: 59984712-59984712 on Build GRCh38
rs770075229 Chr.20: 59984725-59984725 on Build GRCh38
rs757351536 Chr.20: 59985019-59985019 on Build GRCh38
rs150010707 Chr.20: 59984797-59984797 on Build GRCh38
rs561008703 Chr.20: 59984888-59984888 on Build GRCh38
rs141052091 Chr.20: 59984704-59984704 on Build GRCh38
rs988886389 Chr.20: 59984893-59984893 on Build GRCh38
rs749375933 Chr.20: 59984823-59984823 on Build GRCh38
rs775177946 Chr.20: 59984614-59984614 on Build GRCh38
rs751913103 Chr.20: 59984692-59984692 on Build GRCh38
rs141839224 Chr.20: 59984609-59984609 on Build GRCh38
rs760516934 Chr.20: 59984735-59984735 on Build GRCh38
rs760430859 Chr.20: 59984628-59984628 on Build GRCh38
rs367645805 Chr.20: 59984811-59984811 on Build GRCh38
rs1001402526 Chr.20: 59984620-59984620 on Build GRCh38
rs925875112 Chr.20: 59984868-59984868 on Build GRCh38
rs915565610 Chr.20: 59984832-59984832 on Build GRCh38
rs997994144 Chr.20: 59984932-59984932 on Build GRCh38
rs761559005 Chr.20: 59984960-59984960 on Build GRCh38
rs776430278 Chr.20: 59984856-59984856 on Build GRCh38
rs867689552 Chr.20: 59984734-59984735 on Build GRCh38
rs773183532 Chr.20: 59984848-59984848 on Build GRCh38
rs969908232 Chr.20: 59984831-59984831 on Build GRCh38
rs201340865 Chr.20: 59985003-59985003 on Build GRCh38
rs578030495 Chr.20: 59984762-59984762 on Build GRCh38
rs764655668 Chr.20: 59984657-59984657 on Build GRCh38
rs186140944 Chr.20: 59984663-59984663 on Build GRCh38
rs190219512 Chr.20: 59985002-59985002 on Build GRCh38
rs867833872 Chr.20: 59984785-59984785 on Build GRCh38
rs781694705 Chr.20: 59984792-59984792 on Build GRCh38
rs762835716 Chr.20: 59984803-59984803 on Build GRCh38
rs766848205 Chr.20: 59984779-59984779 on Build GRCh38
rs918964707 Chr.20: 59984574-59984574 on Build GRCh38
rs368776859 Chr.20: 59984961-59984961 on Build GRCh38
rs545316943 Chr.20: 59984770-59984770 on Build GRCh38
rs780572708 Chr.20: 59984672-59984672 on Build GRCh38
rs578227845 Chr.20: 59984577-59984577 on Build GRCh38
rs144417269 Chr.20: 59984982-59984982 on Build GRCh38
rs557944532 Chr.20: 59984667-59984667 on Build GRCh38
rs768519489 Chr.20: 59984850-59984850 on Build GRCh38
rs770515588 Chr.20: 59984607-59984607 on Build GRCh38
rs1053008908 Chr.20: 59984929-59984929 on Build GRCh38
rs531813212 Chr.20: 59984908-59984908 on Build GRCh38
rs768090380 Chr.20: 59984999-59984999 on Build GRCh38
rs557419371 Chr.20: 59984575-59984575 on Build GRCh38
rs756434386 Chr.20: 59984810-59984810 on Build GRCh38
rs936950521 Chr.20: 59984903-59984903 on Build GRCh38
rs565753502 Chr.20: 59985004-59985004 on Build GRCh38
rs762689603 Chr.20: 59984658-59984658 on Build GRCh38
rs35720809 Chr.20: 59984780-59984780 on Build GRCh38
rs771029295 Chr.20: 59984842-59984842 on Build GRCh38
rs762432945 Chr.20: 59984979-59984979 on Build GRCh38
rs960401402 Chr.20: 59984809-59984809 on Build GRCh38
rs892656528 Chr.20: 59984930-59984930 on Build GRCh38
rs6100681 Chr.20: 59984805-59984805 on Build GRCh38
rs1042706327 Chr.20: 59984865-59984865 on Build GRCh38
rs1041195048 Chr.20: 59984648-59984648 on Build GRCh38
rs542370493 Chr.20: 59984876-59984876 on Build GRCh38
rs116411308 Chr.20: 59984990-59984990 on Build GRCh38
rs755359539 Chr.20: 59984823-59984824 on Build GRCh38
rs386815753 Chr.20: 59984575-59984577 on Build GRCh38
rs749936794 Chr.20: 59984764-59984764 on Build GRCh38
rs761466507 Chr.20: 59984761-59984761 on Build GRCh38
rs779918020 Chr.20: 59984844-59984844 on Build GRCh38
rs1474909 Chr.20: 59984599-59984599 on Build GRCh38
rs1057173677 Chr.20: 59984576-59984576 on Build GRCh38
rs777189094 Chr.20: 59984573-59984573 on Build GRCh38
rs3761265 Chr.20: 59984947-59984947 on Build GRCh38
rs777730285 Chr.20: 59984728-59984728 on Build GRCh38
rs899051033 Chr.20: 59985012-59985012 on Build GRCh38
rs769374963 Chr.20: 59984964-59984964 on Build GRCh38
rs754201068 Chr.20: 59984824-59984824 on Build GRCh38
rs144821881 Chr.20: 59984752-59984752 on Build GRCh38
rs755376152 Chr.20: 59984698-59984698 on Build GRCh38
rs767589540 Chr.20: 59984837-59984837 on Build GRCh38
rs758854327 Chr.20: 59984668-59984668 on Build GRCh38
rs755461972 Chr.20: 59984789-59984789 on Build GRCh38
rs763785040 Chr.20: 59984630-59984630 on Build GRCh38

View Full Product Details