Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78891088-78891598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRL4
Entrez Gene ID
64123
Gene Name
adhesion G protein-coupled receptor L4
Gene Aliases
ELTD1, ETL, KPG_003
UniGene
Hs.132314
Gene Chromosome Location
Chr.1: 78889764-79006810 on Build GRCh38
Transcripts
NM_022159.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7099512 c.377C>A p.S126Y Chr.1: 78891531-78891531 on Build GRCh38
1344453 c.1987T>G p.F663V Chr.1: 78891547-78891547 on Build GRCh38
3492610 c.351C>T p.F117F Chr.1: 78891557-78891557 on Build GRCh38
4815617 c.380G>C p.R127T Chr.1: 78891528-78891528 on Build GRCh38
395862 c.1968G>T p.Q656H Chr.1: 78891566-78891566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772066842 Chr.1: 78891488-78891488 on Build GRCh38
rs541284919 Chr.1: 78891264-78891264 on Build GRCh38
rs751196124 Chr.1: 78891218-78891218 on Build GRCh38
rs773280974 Chr.1: 78891493-78891493 on Build GRCh38
rs896732476 Chr.1: 78891296-78891296 on Build GRCh38
rs755609733 Chr.1: 78891550-78891550 on Build GRCh38
rs756751896 Chr.1: 78891223-78891223 on Build GRCh38
rs923856375 Chr.1: 78891416-78891416 on Build GRCh38
rs895005606 Chr.1: 78891551-78891551 on Build GRCh38
rs762885058 Chr.1: 78891518-78891518 on Build GRCh38
rs767601963 Chr.1: 78891176-78891176 on Build GRCh38
rs776766899 Chr.1: 78891256-78891256 on Build GRCh38
rs12025343 Chr.1: 78891374-78891374 on Build GRCh38
rs749763189 Chr.1: 78891563-78891563 on Build GRCh38
rs545309183 Chr.1: 78891395-78891395 on Build GRCh38
rs770040543 Chr.1: 78891549-78891549 on Build GRCh38
rs773815939 Chr.1: 78891548-78891549 on Build GRCh38
rs780709172 Chr.1: 78891225-78891225 on Build GRCh38
rs964717377 Chr.1: 78891383-78891383 on Build GRCh38
rs947134421 Chr.1: 78891159-78891159 on Build GRCh38
rs747051803 Chr.1: 78891133-78891134 on Build GRCh38
rs955992965 Chr.1: 78891444-78891445 on Build GRCh38
rs748577253 Chr.1: 78891516-78891516 on Build GRCh38
rs777620699 Chr.1: 78891185-78891185 on Build GRCh38
rs17102364 Chr.1: 78891254-78891254 on Build GRCh38
rs201074858 Chr.1: 78891508-78891508 on Build GRCh38
rs768934690 Chr.1: 78891150-78891150 on Build GRCh38
rs770077981 Chr.1: 78891133-78891133 on Build GRCh38
rs1047898526 Chr.1: 78891335-78891335 on Build GRCh38
rs780585372 Chr.1: 78891554-78891554 on Build GRCh38
rs766724256 Chr.1: 78891350-78891350 on Build GRCh38
rs371284509 Chr.1: 78891138-78891138 on Build GRCh38
rs578258210 Chr.1: 78891315-78891315 on Build GRCh38
rs368003305 Chr.1: 78891112-78891112 on Build GRCh38
rs765711623 Chr.1: 78891405-78891405 on Build GRCh38
rs763945454 Chr.1: 78891519-78891519 on Build GRCh38
rs761846480 Chr.1: 78891165-78891165 on Build GRCh38
rs756877278 Chr.1: 78891523-78891523 on Build GRCh38
rs775552842 Chr.1: 78891513-78891513 on Build GRCh38
rs748328276 Chr.1: 78891576-78891576 on Build GRCh38
rs751198452 Chr.1: 78891521-78891521 on Build GRCh38
rs756548124 Chr.1: 78891233-78891233 on Build GRCh38
rs944016375 Chr.1: 78891408-78891408 on Build GRCh38
rs117322873 Chr.1: 78891409-78891409 on Build GRCh38
rs765133355 Chr.1: 78891181-78891181 on Build GRCh38
rs750017791 Chr.1: 78891532-78891532 on Build GRCh38
rs759465075 Chr.1: 78891180-78891180 on Build GRCh38
rs554373790 Chr.1: 78891177-78891177 on Build GRCh38
rs968522372 Chr.1: 78891500-78891500 on Build GRCh38
rs539155792 Chr.1: 78891307-78891307 on Build GRCh38
rs748360221 Chr.1: 78891484-78891484 on Build GRCh38
rs143948092 Chr.1: 78891226-78891226 on Build GRCh38
rs934531375 Chr.1: 78891118-78891118 on Build GRCh38
rs577581447 Chr.1: 78891184-78891184 on Build GRCh38
rs558462927 Chr.1: 78891341-78891341 on Build GRCh38
rs755366746 Chr.1: 78891564-78891564 on Build GRCh38
rs974233005 Chr.1: 78891322-78891322 on Build GRCh38
rs988666224 Chr.1: 78891140-78891140 on Build GRCh38
rs759976634 Chr.1: 78891402-78891402 on Build GRCh38
rs372767071 Chr.1: 78891479-78891479 on Build GRCh38
rs780536142 Chr.1: 78891238-78891238 on Build GRCh38
rs545398491 Chr.1: 78891340-78891340 on Build GRCh38
rs746374532 Chr.1: 78891229-78891229 on Build GRCh38
rs561230591 Chr.1: 78891465-78891465 on Build GRCh38
rs759392236 Chr.1: 78891498-78891498 on Build GRCh38
rs1017623496 Chr.1: 78891371-78891371 on Build GRCh38
rs766942019 Chr.1: 78891530-78891530 on Build GRCh38
rs938348930 Chr.1: 78891251-78891251 on Build GRCh38
rs779288246 Chr.1: 78891566-78891566 on Build GRCh38
rs936625570 Chr.1: 78891419-78891419 on Build GRCh38
rs201808653 Chr.1: 78891141-78891141 on Build GRCh38
rs768983718 Chr.1: 78891257-78891257 on Build GRCh38
rs774541181 Chr.1: 78891163-78891163 on Build GRCh38
rs914427081 Chr.1: 78891480-78891480 on Build GRCh38

View Full Product Details