Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86441314-86441811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA2
Entrez Gene ID
9635
Gene Name
chloride channel accessory 2
Gene Aliases
CACC, CACC3, CLCRG2, CaCC-3
UniGene
Hs.241551
Gene Chromosome Location
Chr.1: 86424086-86456558 on Build GRCh38
Transcripts
NM_006536.5 XM_011542448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
682845 c.1444A>T p.R482* Chr.1: 86441499-86441499 on Build GRCh38
5041708 c.1391A>T p.K464M Chr.1: 86441446-86441446 on Build GRCh38
465030 c.1423A>G p.S475G Chr.1: 86441478-86441478 on Build GRCh38
7263631 c.1389_1390delAA p.K464fs*11 Chr.1: 86441444-86441445 on Build GRCh38
4168386 c.1429A>C p.I477L Chr.1: 86441484-86441484 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765787586 Chr.1: 86441503-86441503 on Build GRCh38
rs1010074003 Chr.1: 86441666-86441666 on Build GRCh38
rs768959628 Chr.1: 86441485-86441485 on Build GRCh38
rs752431432 Chr.1: 86441529-86441529 on Build GRCh38
rs764893223 Chr.1: 86441511-86441511 on Build GRCh38
rs184189407 Chr.1: 86441542-86441542 on Build GRCh38
rs996604801 Chr.1: 86441613-86441613 on Build GRCh38
rs775695100 Chr.1: 86441469-86441469 on Build GRCh38
rs749414235 Chr.1: 86441543-86441543 on Build GRCh38
rs757266293 Chr.1: 86441539-86441539 on Build GRCh38
rs150380012 Chr.1: 86441418-86441418 on Build GRCh38
rs368041893 Chr.1: 86441508-86441508 on Build GRCh38
rs774812585 Chr.1: 86441486-86441486 on Build GRCh38
rs888301194 Chr.1: 86441684-86441684 on Build GRCh38
rs72951856 Chr.1: 86441356-86441356 on Build GRCh38
rs922161923 Chr.1: 86441455-86441455 on Build GRCh38
rs746695614 Chr.1: 86441778-86441778 on Build GRCh38
rs192143040 Chr.1: 86441376-86441376 on Build GRCh38
rs979038527 Chr.1: 86441764-86441764 on Build GRCh38
rs748451774 Chr.1: 86441567-86441567 on Build GRCh38
rs17129169 Chr.1: 86441714-86441714 on Build GRCh38
rs899777902 Chr.1: 86441596-86441596 on Build GRCh38
rs571469714 Chr.1: 86441590-86441590 on Build GRCh38
rs768074158 Chr.1: 86441492-86441492 on Build GRCh38
rs138070441 Chr.1: 86441466-86441466 on Build GRCh38
rs763061061 Chr.1: 86441474-86441474 on Build GRCh38
rs574335752 Chr.1: 86441510-86441510 on Build GRCh38
rs1008127480 Chr.1: 86441342-86441342 on Build GRCh38
rs758193376 Chr.1: 86441530-86441530 on Build GRCh38
rs372970570 Chr.1: 86441406-86441406 on Build GRCh38
rs55913596 Chr.1: 86441708-86441708 on Build GRCh38
rs771948029 Chr.1: 86441433-86441433 on Build GRCh38
rs34373018 Chr.1: 86441401-86441402 on Build GRCh38
rs952810059 Chr.1: 86441400-86441400 on Build GRCh38
rs777701119 Chr.1: 86441458-86441458 on Build GRCh38
rs970312385 Chr.1: 86441715-86441715 on Build GRCh38
rs371299688 Chr.1: 86441557-86441557 on Build GRCh38
rs1026760206 Chr.1: 86441614-86441614 on Build GRCh38
rs755219834 Chr.1: 86441564-86441564 on Build GRCh38
rs761306120 Chr.1: 86441499-86441499 on Build GRCh38
rs267598742 Chr.1: 86441498-86441498 on Build GRCh38
rs201243498 Chr.1: 86441490-86441490 on Build GRCh38
rs1045108119 Chr.1: 86441559-86441559 on Build GRCh38
rs1016125132 Chr.1: 86441692-86441692 on Build GRCh38
rs764238169 Chr.1: 86441550-86441550 on Build GRCh38
rs56398778 Chr.1: 86441579-86441579 on Build GRCh38
rs996637239 Chr.1: 86441377-86441377 on Build GRCh38
rs529671681 Chr.1: 86441463-86441463 on Build GRCh38
rs549739374 Chr.1: 86441693-86441693 on Build GRCh38
rs149530117 Chr.1: 86441690-86441690 on Build GRCh38
rs962486719 Chr.1: 86441785-86441785 on Build GRCh38
rs778650447 Chr.1: 86441393-86441393 on Build GRCh38
rs537265526 Chr.1: 86441633-86441633 on Build GRCh38
rs1027002960 Chr.1: 86441398-86441398 on Build GRCh38
rs1052636917 Chr.1: 86441547-86441547 on Build GRCh38
rs746905925 Chr.1: 86441537-86441537 on Build GRCh38
rs375454289 Chr.1: 86441496-86441496 on Build GRCh38
rs370308309 Chr.1: 86441468-86441468 on Build GRCh38
rs760779161 Chr.1: 86441456-86441456 on Build GRCh38
rs960353609 Chr.1: 86441462-86441462 on Build GRCh38
rs777611253 Chr.1: 86441534-86441534 on Build GRCh38
rs987677225 Chr.1: 86441734-86441734 on Build GRCh38

View Full Product Details