Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62474919-62475408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA5
Entrez Gene ID
140628
Gene Name
GATA binding protein 5
Gene Aliases
GATAS, bB379O24.1
UniGene
Hs.352250
Gene Chromosome Location
Chr.20: 62463497-62475970 on Build GRCh38
Transcripts
NM_080473.4 XM_006723699.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6575125 c.370G>A p.E124K Chr.20: 62475152-62475152 on Build GRCh38
4168635 c.286G>A p.A96T Chr.20: 62475236-62475236 on Build GRCh38
5766615 c.378C>T p.F126F Chr.20: 62475144-62475144 on Build GRCh38
6575126 c.225C>T p.F75F Chr.20: 62475297-62475297 on Build GRCh38
6558972 c.437T>A p.V146E Chr.20: 62475085-62475085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113823160 Chr.20: 62475235-62475235 on Build GRCh38
rs1034727380 Chr.20: 62475279-62475279 on Build GRCh38
rs782029132 Chr.20: 62475141-62475141 on Build GRCh38
rs899188199 Chr.20: 62475261-62475261 on Build GRCh38
rs782144157 Chr.20: 62474950-62474950 on Build GRCh38
rs782299640 Chr.20: 62475069-62475069 on Build GRCh38
rs572247741 Chr.20: 62475290-62475290 on Build GRCh38
rs782667889 Chr.20: 62475331-62475331 on Build GRCh38
rs111545528 Chr.20: 62475074-62475074 on Build GRCh38
rs782693475 Chr.20: 62475140-62475140 on Build GRCh38
rs781793264 Chr.20: 62475313-62475313 on Build GRCh38
rs7267800 Chr.20: 62474941-62474941 on Build GRCh38
rs781995328 Chr.20: 62475367-62475367 on Build GRCh38
rs111821441 Chr.20: 62474948-62474948 on Build GRCh38
rs6061248 Chr.20: 62475048-62475048 on Build GRCh38
rs141950357 Chr.20: 62475026-62475026 on Build GRCh38
rs1014889761 Chr.20: 62475000-62475000 on Build GRCh38
rs869025434 Chr.20: 62475379-62475379 on Build GRCh38
rs1024163550 Chr.20: 62475296-62475296 on Build GRCh38
rs975107982 Chr.20: 62475335-62475335 on Build GRCh38
rs782352129 Chr.20: 62475127-62475127 on Build GRCh38
rs977365389 Chr.20: 62475321-62475321 on Build GRCh38
rs112433432 Chr.20: 62475045-62475045 on Build GRCh38
rs111633561 Chr.20: 62475030-62475030 on Build GRCh38
rs782804041 Chr.20: 62474983-62474984 on Build GRCh38
rs781801892 Chr.20: 62475014-62475014 on Build GRCh38
rs782403666 Chr.20: 62475341-62475341 on Build GRCh38
rs539125153 Chr.20: 62475216-62475216 on Build GRCh38
rs782337853 Chr.20: 62475258-62475258 on Build GRCh38
rs995352294 Chr.20: 62475019-62475019 on Build GRCh38
rs1039893924 Chr.20: 62475177-62475177 on Build GRCh38
rs554758872 Chr.20: 62475345-62475345 on Build GRCh38
rs782221467 Chr.20: 62475050-62475050 on Build GRCh38
rs782726884 Chr.20: 62474985-62474985 on Build GRCh38
rs967186417 Chr.20: 62475311-62475311 on Build GRCh38
rs782160947 Chr.20: 62475327-62475327 on Build GRCh38
rs111554140 Chr.20: 62475098-62475098 on Build GRCh38
rs782503555 Chr.20: 62475044-62475044 on Build GRCh38
rs890593605 Chr.20: 62475231-62475231 on Build GRCh38
rs373706342 Chr.20: 62475132-62475132 on Build GRCh38
rs921019435 Chr.20: 62475206-62475206 on Build GRCh38
rs782095551 Chr.20: 62474975-62474975 on Build GRCh38
rs782791728 Chr.20: 62474959-62474959 on Build GRCh38
rs111252974 Chr.20: 62475359-62475359 on Build GRCh38
rs782058415 Chr.20: 62475137-62475137 on Build GRCh38
rs782141667 Chr.20: 62475144-62475144 on Build GRCh38
rs935667661 Chr.20: 62475136-62475136 on Build GRCh38
rs894573638 Chr.20: 62475282-62475282 on Build GRCh38
rs782240636 Chr.20: 62475105-62475105 on Build GRCh38
rs781981404 Chr.20: 62474961-62474961 on Build GRCh38
rs782571269 Chr.20: 62475053-62475053 on Build GRCh38
rs781841004 Chr.20: 62475214-62475214 on Build GRCh38
rs994848200 Chr.20: 62475274-62475274 on Build GRCh38
rs933702892 Chr.20: 62475212-62475212 on Build GRCh38
rs1036595030 Chr.20: 62475237-62475237 on Build GRCh38
rs6142775 Chr.20: 62475323-62475323 on Build GRCh38
rs782427595 Chr.20: 62475017-62475017 on Build GRCh38

View Full Product Details