Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62475173-62475701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA5
Entrez Gene ID
140628
Gene Name
GATA binding protein 5
Gene Aliases
GATAS, bB379O24.1
UniGene
Hs.352250
Gene Chromosome Location
Chr.20: 62463497-62475970 on Build GRCh38
Transcripts
NM_080473.4 XM_006723699.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4168635 c.286G>A p.A96T Chr.20: 62475236-62475236 on Build GRCh38
6575126 c.225C>T p.F75F Chr.20: 62475297-62475297 on Build GRCh38
5730265 c.86_93delGCTCTCCG p.G29fs*19 Chr.20: 62475429-62475436 on Build GRCh38
6387374 c.322G>A p.A108T Chr.20: 62475200-62475200 on Build GRCh38
3749728 c.199A>C p.T67P Chr.20: 62475323-62475323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782556189 Chr.20: 62475456-62475456 on Build GRCh38
rs894458923 Chr.20: 62475469-62475469 on Build GRCh38
rs782570688 Chr.20: 62475525-62475525 on Build GRCh38
rs930857802 Chr.20: 62475442-62475442 on Build GRCh38
rs1034727380 Chr.20: 62475279-62475279 on Build GRCh38
rs113823160 Chr.20: 62475235-62475235 on Build GRCh38
rs529753490 Chr.20: 62475614-62475614 on Build GRCh38
rs782470530 Chr.20: 62475516-62475516 on Build GRCh38
rs782050292 Chr.20: 62475487-62475487 on Build GRCh38
rs534892601 Chr.20: 62475653-62475653 on Build GRCh38
rs782320503 Chr.20: 62475444-62475444 on Build GRCh38
rs930931298 Chr.20: 62475624-62475624 on Build GRCh38
rs781869793 Chr.20: 62475392-62475392 on Build GRCh38
rs782399317 Chr.20: 62475470-62475470 on Build GRCh38
rs899188199 Chr.20: 62475261-62475261 on Build GRCh38
rs113068438 Chr.20: 62475514-62475514 on Build GRCh38
rs782254054 Chr.20: 62475548-62475548 on Build GRCh38
rs782108635 Chr.20: 62475473-62475473 on Build GRCh38
rs572247741 Chr.20: 62475290-62475290 on Build GRCh38
rs782819196 Chr.20: 62475492-62475492 on Build GRCh38
rs782667889 Chr.20: 62475331-62475331 on Build GRCh38
rs781942087 Chr.20: 62475484-62475484 on Build GRCh38
rs781793264 Chr.20: 62475313-62475313 on Build GRCh38
rs564720227 Chr.20: 62475496-62475496 on Build GRCh38
rs782607630 Chr.20: 62475575-62475575 on Build GRCh38
rs781995328 Chr.20: 62475367-62475367 on Build GRCh38
rs986539596 Chr.20: 62475564-62475564 on Build GRCh38
rs112701160 Chr.20: 62475439-62475439 on Build GRCh38
rs986487322 Chr.20: 62475399-62475399 on Build GRCh38
rs869025434 Chr.20: 62475379-62475379 on Build GRCh38
rs1045889469 Chr.20: 62475673-62475673 on Build GRCh38
rs1024163550 Chr.20: 62475296-62475296 on Build GRCh38
rs730880111 Chr.20: 62475418-62475418 on Build GRCh38
rs975107982 Chr.20: 62475335-62475335 on Build GRCh38
rs977365389 Chr.20: 62475321-62475321 on Build GRCh38
rs78235297 Chr.20: 62475569-62475569 on Build GRCh38
rs782349038 Chr.20: 62475474-62475474 on Build GRCh38
rs955021509 Chr.20: 62475545-62475545 on Build GRCh38
rs1056432005 Chr.20: 62475640-62475640 on Build GRCh38
rs933755273 Chr.20: 62475391-62475391 on Build GRCh38
rs939735663 Chr.20: 62475570-62475570 on Build GRCh38
rs782403666 Chr.20: 62475341-62475341 on Build GRCh38
rs539125153 Chr.20: 62475216-62475216 on Build GRCh38
rs940848073 Chr.20: 62475411-62475411 on Build GRCh38
rs957317148 Chr.20: 62475523-62475523 on Build GRCh38
rs782337853 Chr.20: 62475258-62475258 on Build GRCh38
rs554758872 Chr.20: 62475345-62475345 on Build GRCh38
rs967186417 Chr.20: 62475311-62475311 on Build GRCh38
rs200383755 Chr.20: 62475466-62475466 on Build GRCh38
rs782160947 Chr.20: 62475327-62475327 on Build GRCh38
rs781990084 Chr.20: 62475472-62475472 on Build GRCh38
rs890593605 Chr.20: 62475231-62475231 on Build GRCh38
rs781786147 Chr.20: 62475530-62475530 on Build GRCh38
rs972267570 Chr.20: 62475597-62475597 on Build GRCh38
rs782225800 Chr.20: 62475540-62475540 on Build GRCh38
rs768370922 Chr.20: 62475675-62475675 on Build GRCh38
rs782762414 Chr.20: 62475505-62475505 on Build GRCh38
rs921019435 Chr.20: 62475206-62475206 on Build GRCh38
rs950024887 Chr.20: 62475663-62475663 on Build GRCh38
rs782535897 Chr.20: 62475534-62475534 on Build GRCh38
rs782504520 Chr.20: 62475405-62475405 on Build GRCh38
rs111252974 Chr.20: 62475359-62475359 on Build GRCh38
rs903053351 Chr.20: 62475511-62475511 on Build GRCh38
rs532016630 Chr.20: 62475497-62475497 on Build GRCh38
rs782621711 Chr.20: 62475437-62475437 on Build GRCh38
rs894573638 Chr.20: 62475282-62475282 on Build GRCh38
rs781841004 Chr.20: 62475214-62475214 on Build GRCh38
rs894509480 Chr.20: 62475644-62475644 on Build GRCh38
rs1036595030 Chr.20: 62475237-62475237 on Build GRCh38
rs933702892 Chr.20: 62475212-62475212 on Build GRCh38
rs994848200 Chr.20: 62475274-62475274 on Build GRCh38
rs6142775 Chr.20: 62475323-62475323 on Build GRCh38

View Full Product Details