Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63238224-63238717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC7
Entrez Gene ID
79444
Gene Name
baculoviral IAP repeat containing 7
Gene Aliases
KIAP, LIVIN, ML-IAP, MLIAP, RNF50
UniGene
Hs.256126
Gene Chromosome Location
Chr.20: 63235883-63240507 on Build GRCh38
Transcripts
NM_022161.3 NM_139317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285444 c.465C>T p.L155L Chr.20: 63238411-63238411 on Build GRCh38
30441 c.491A>G p.H164R Chr.20: 63238437-63238437 on Build GRCh38
5858314 c.541G>A p.E181K Chr.20: 63238578-63238578 on Build GRCh38
3758732 c.528C>T p.S176S Chr.20: 63238474-63238474 on Build GRCh38
5025124 c.467G>A p.R156Q Chr.20: 63238413-63238413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773299388 Chr.20: 63238403-63238403 on Build GRCh38
rs747602542 Chr.20: 63238248-63238248 on Build GRCh38
rs774145246 Chr.20: 63238323-63238323 on Build GRCh38
rs772884973 Chr.20: 63238281-63238281 on Build GRCh38
rs541193453 Chr.20: 63238276-63238276 on Build GRCh38
rs6090325 Chr.20: 63238336-63238336 on Build GRCh38
rs1028256660 Chr.20: 63238366-63238366 on Build GRCh38
rs768567221 Chr.20: 63238444-63238444 on Build GRCh38
rs61737392 Chr.20: 63238437-63238437 on Build GRCh38
rs944748744 Chr.20: 63238261-63238261 on Build GRCh38
rs539371735 Chr.20: 63238505-63238505 on Build GRCh38
rs768121481 Chr.20: 63238416-63238416 on Build GRCh38
rs552295732 Chr.20: 63238412-63238412 on Build GRCh38
rs375365000 Chr.20: 63238497-63238497 on Build GRCh38
rs764547508 Chr.20: 63238570-63238570 on Build GRCh38
rs779148609 Chr.20: 63238634-63238634 on Build GRCh38
rs568867607 Chr.20: 63238500-63238500 on Build GRCh38
rs756960663 Chr.20: 63238431-63238431 on Build GRCh38
rs769086136 Chr.20: 63238649-63238649 on Build GRCh38
rs939540240 Chr.20: 63238304-63238304 on Build GRCh38
rs6011693 Chr.20: 63238597-63238597 on Build GRCh38
rs765533405 Chr.20: 63238287-63238287 on Build GRCh38
rs766054062 Chr.20: 63238589-63238589 on Build GRCh38
rs756606378 Chr.20: 63238417-63238417 on Build GRCh38
rs374758660 Chr.20: 63238687-63238687 on Build GRCh38
rs371308264 Chr.20: 63238303-63238303 on Build GRCh38
rs775750104 Chr.20: 63238643-63238643 on Build GRCh38
rs73316017 Chr.20: 63238639-63238639 on Build GRCh38
rs372159109 Chr.20: 63238439-63238439 on Build GRCh38
rs199795769 Chr.20: 63238413-63238413 on Build GRCh38
rs767384081 Chr.20: 63238274-63238274 on Build GRCh38
rs965848770 Chr.20: 63238614-63238614 on Build GRCh38
rs755558237 Chr.20: 63238307-63238307 on Build GRCh38
rs770504777 Chr.20: 63238317-63238317 on Build GRCh38
rs989197876 Chr.20: 63238585-63238585 on Build GRCh38
rs753412654 Chr.20: 63238483-63238483 on Build GRCh38
rs759784654 Chr.20: 63238473-63238473 on Build GRCh38
rs753330289 Chr.20: 63238427-63238427 on Build GRCh38
rs772279823 Chr.20: 63238545-63238545 on Build GRCh38
rs756051104 Chr.20: 63238613-63238613 on Build GRCh38
rs995718826 Chr.20: 63238640-63238640 on Build GRCh38
rs753372481 Chr.20: 63238605-63238606 on Build GRCh38
rs761999334 Chr.20: 63238376-63238376 on Build GRCh38
rs201279207 Chr.20: 63238504-63238504 on Build GRCh38
rs6010879 Chr.20: 63238293-63238293 on Build GRCh38
rs774534364 Chr.20: 63238516-63238516 on Build GRCh38
rs763268084 Chr.20: 63238404-63238404 on Build GRCh38
rs183504802 Chr.20: 63238250-63238250 on Build GRCh38
rs894481112 Chr.20: 63238478-63238478 on Build GRCh38
rs956356857 Chr.20: 63238339-63238339 on Build GRCh38
rs751355860 Chr.20: 63238592-63238592 on Build GRCh38
rs761793165 Chr.20: 63238279-63238279 on Build GRCh38
rs749158370 Chr.20: 63238258-63238258 on Build GRCh38
rs771321879 Chr.20: 63238455-63238455 on Build GRCh38
rs199946289 Chr.20: 63238558-63238558 on Build GRCh38
rs764755969 Chr.20: 63238575-63238575 on Build GRCh38
rs765252456 Chr.20: 63238381-63238381 on Build GRCh38
rs147960898 Chr.20: 63238586-63238586 on Build GRCh38
rs759474257 Chr.20: 63238269-63238269 on Build GRCh38
rs202043692 Chr.20: 63238581-63238581 on Build GRCh38
rs577122373 Chr.20: 63238263-63238263 on Build GRCh38
rs202147805 Chr.20: 63238648-63238648 on Build GRCh38
rs761100275 Chr.20: 63238477-63238477 on Build GRCh38
rs530112822 Chr.20: 63238297-63238297 on Build GRCh38
rs774695783 Chr.20: 63238466-63238466 on Build GRCh38
rs755643687 Chr.20: 63238449-63238453 on Build GRCh38
rs146445887 Chr.20: 63238425-63238425 on Build GRCh38
rs778583357 Chr.20: 63238493-63238493 on Build GRCh38
rs745930356 Chr.20: 63238636-63238636 on Build GRCh38
rs746946810 Chr.20: 63238443-63238443 on Build GRCh38
rs772062466 Chr.20: 63238330-63238330 on Build GRCh38
rs914112625 Chr.20: 63238529-63238529 on Build GRCh38
rs369077805 Chr.20: 63238631-63238631 on Build GRCh38
rs750096360 Chr.20: 63238496-63238496 on Build GRCh38
rs79055277 Chr.20: 63238604-63238604 on Build GRCh38
rs200627723 Chr.20: 63238435-63238435 on Build GRCh38
rs188716182 Chr.20: 63238305-63238305 on Build GRCh38
rs377228781 Chr.20: 63238650-63238650 on Build GRCh38
rs775257810 Chr.20: 63238359-63238359 on Build GRCh38
rs6090326 Chr.20: 63238447-63238447 on Build GRCh38
rs752080915 Chr.20: 63238573-63238573 on Build GRCh38
rs1044199004 Chr.20: 63238310-63238310 on Build GRCh38
rs75064873 Chr.20: 63238309-63238309 on Build GRCh38
rs1008135042 Chr.20: 63238389-63238389 on Build GRCh38
rs759845051 Chr.20: 63238525-63238525 on Build GRCh38
rs764649174 Chr.20: 63238480-63238480 on Build GRCh38
rs557352737 Chr.20: 63238546-63238546 on Build GRCh38
rs556406992 Chr.20: 63238268-63238268 on Build GRCh38
rs6122405 Chr.20: 63238574-63238574 on Build GRCh38
rs777290172 Chr.20: 63238308-63238308 on Build GRCh38
rs6122106 Chr.20: 63238361-63238361 on Build GRCh38
rs747537721 Chr.20: 63238246-63238246 on Build GRCh38
rs763123962 Chr.20: 63238454-63238454 on Build GRCh38
rs112863575 Chr.20: 63238441-63238441 on Build GRCh38
rs149839516 Chr.20: 63238583-63238583 on Build GRCh38
rs751918819 Chr.20: 63238300-63238300 on Build GRCh38
rs755498636 Chr.20: 63238247-63238247 on Build GRCh38
rs553149604 Chr.20: 63238587-63238587 on Build GRCh38
rs2273487 Chr.20: 63238474-63238474 on Build GRCh38
rs867474913 Chr.20: 63238356-63238356 on Build GRCh38
rs779853651 Chr.20: 63238586-63238586 on Build GRCh38
rs766699014 Chr.20: 63238407-63238407 on Build GRCh38
rs6010878 Chr.20: 63238255-63238255 on Build GRCh38
rs762309805 Chr.20: 63238651-63238651 on Build GRCh38
rs781088554 Chr.20: 63238601-63238601 on Build GRCh38
rs766877625 Chr.20: 63238298-63238298 on Build GRCh38
rs979863831 Chr.20: 63238694-63238694 on Build GRCh38
rs756728148 Chr.20: 63238488-63238488 on Build GRCh38
rs745648768 Chr.20: 63238326-63238326 on Build GRCh38

View Full Product Details