Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32602175-32602680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf59
Entrez Gene ID
56683
Gene Name
chromosome 21 open reading frame 59
Gene Aliases
C21orf48, CILD26, FBB18, Kur
UniGene
Hs.5811
Gene Chromosome Location
Chr.21: 32601674-32612608 on Build GRCh38
Transcripts
NM_021254.2 NR_036552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7097813 c.700A>C p.I234L Chr.21: 32602334-32602334 on Build GRCh38
7147154 c.726G>C p.L242L Chr.21: 32602308-32602308 on Build GRCh38
237076 c.693G>A p.E231E Chr.21: 32602341-32602341 on Build GRCh38
6280852 c.721C>A p.Q241K Chr.21: 32602313-32602313 on Build GRCh38
4934320 c.697A>G p.I233V Chr.21: 32602337-32602337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011337350 Chr.21: 32602415-32602415 on Build GRCh38
rs780236404 Chr.21: 32602304-32602304 on Build GRCh38
rs755214255 Chr.21: 32602391-32602391 on Build GRCh38
rs149526723 Chr.21: 32602305-32602305 on Build GRCh38
rs554890649 Chr.21: 32602424-32602424 on Build GRCh38
rs181212182 Chr.21: 32602447-32602447 on Build GRCh38
rs184197655 Chr.21: 32602448-32602448 on Build GRCh38
rs745936315 Chr.21: 32602219-32602219 on Build GRCh38
rs373822170 Chr.21: 32602224-32602224 on Build GRCh38
rs759139045 Chr.21: 32602263-32602263 on Build GRCh38
rs139494282 Chr.21: 32602320-32602320 on Build GRCh38
rs764480518 Chr.21: 32602235-32602235 on Build GRCh38
rs766192360 Chr.21: 32602581-32602581 on Build GRCh38
rs149703412 Chr.21: 32602333-32602333 on Build GRCh38
rs895381551 Chr.21: 32602433-32602433 on Build GRCh38
rs570924113 Chr.21: 32602642-32602642 on Build GRCh38
rs772477321 Chr.21: 32602223-32602223 on Build GRCh38
rs559133435 Chr.21: 32602500-32602500 on Build GRCh38
rs956102006 Chr.21: 32602521-32602521 on Build GRCh38
rs1031083532 Chr.21: 32602503-32602503 on Build GRCh38
rs754209600 Chr.21: 32602347-32602347 on Build GRCh38
rs780919228 Chr.21: 32602322-32602322 on Build GRCh38
rs188996393 Chr.21: 32602612-32602612 on Build GRCh38
rs762213272 Chr.21: 32602352-32602352 on Build GRCh38
rs963090427 Chr.21: 32602613-32602613 on Build GRCh38
rs1029912582 Chr.21: 32602491-32602491 on Build GRCh38
rs1018475106 Chr.21: 32602582-32602582 on Build GRCh38
rs771932871 Chr.21: 32602394-32602394 on Build GRCh38
rs955533207 Chr.21: 32602212-32602212 on Build GRCh38
rs111702591 Chr.21: 32602236-32602236 on Build GRCh38
rs576336322 Chr.21: 32602437-32602437 on Build GRCh38
rs767192965 Chr.21: 32602346-32602346 on Build GRCh38
rs761560756 Chr.21: 32602300-32602300 on Build GRCh38
rs769669889 Chr.21: 32602327-32602327 on Build GRCh38
rs138091636 Chr.21: 32602266-32602266 on Build GRCh38
rs766060470 Chr.21: 32602259-32602259 on Build GRCh38
rs750995181 Chr.21: 32602361-32602361 on Build GRCh38
rs754162158 Chr.21: 32602281-32602281 on Build GRCh38
rs144798054 Chr.21: 32602495-32602495 on Build GRCh38
rs974205868 Chr.21: 32602637-32602637 on Build GRCh38
rs377554956 Chr.21: 32602275-32602275 on Build GRCh38
rs892864092 Chr.21: 32602398-32602398 on Build GRCh38
rs1007045986 Chr.21: 32602519-32602519 on Build GRCh38
rs989970436 Chr.21: 32602531-32602531 on Build GRCh38
rs747963630 Chr.21: 32602326-32602326 on Build GRCh38
rs563611454 Chr.21: 32602568-32602568 on Build GRCh38
rs370327238 Chr.21: 32602337-32602337 on Build GRCh38
rs751242770 Chr.21: 32602260-32602260 on Build GRCh38
rs552310570 Chr.21: 32602638-32602638 on Build GRCh38
rs542143068 Chr.21: 32602532-32602532 on Build GRCh38
rs781456818 Chr.21: 32602397-32602397 on Build GRCh38
rs1027220406 Chr.21: 32602431-32602431 on Build GRCh38
rs758517253 Chr.21: 32602302-32602302 on Build GRCh38
rs777160491 Chr.21: 32602236-32602236 on Build GRCh38
rs576823465 Chr.21: 32602492-32602492 on Build GRCh38
rs568437344 Chr.21: 32602345-32602345 on Build GRCh38
rs914485251 Chr.21: 32602335-32602335 on Build GRCh38
rs1037211382 Chr.21: 32602388-32602388 on Build GRCh38
rs528623742 Chr.21: 32602644-32602644 on Build GRCh38
rs773792172 Chr.21: 32602341-32602341 on Build GRCh38
rs111325880 Chr.21: 32602271-32602271 on Build GRCh38
rs970843465 Chr.21: 32602488-32602488 on Build GRCh38
rs529675973 Chr.21: 32602513-32602513 on Build GRCh38
rs1037220149 Chr.21: 32602626-32602626 on Build GRCh38
rs552275739 Chr.21: 32602228-32602228 on Build GRCh38
rs62214719 Chr.21: 32602211-32602211 on Build GRCh38
rs374530868 Chr.21: 32602389-32602389 on Build GRCh38
rs760617356 Chr.21: 32602508-32602508 on Build GRCh38
rs770529829 Chr.21: 32602343-32602354 on Build GRCh38
rs202094637 Chr.21: 32602299-32602299 on Build GRCh38
rs370677451 Chr.21: 32602330-32602330 on Build GRCh38
rs999745374 Chr.21: 32602461-32602461 on Build GRCh38
rs758921317 Chr.21: 32602364-32602364 on Build GRCh38
rs778861616 Chr.21: 32602198-32602198 on Build GRCh38
rs374753522 Chr.21: 32602360-32602360 on Build GRCh38
rs995680562 Chr.21: 32602392-32602392 on Build GRCh38
rs912199616 Chr.21: 32602553-32602553 on Build GRCh38
rs758342832 Chr.21: 32602222-32602223 on Build GRCh38
rs985139265 Chr.21: 32602537-32602537 on Build GRCh38
rs768339606 Chr.21: 32602258-32602258 on Build GRCh38
rs746477690 Chr.21: 32602248-32602249 on Build GRCh38
rs943713348 Chr.21: 32602577-32602577 on Build GRCh38
rs768598720 Chr.21: 32602227-32602227 on Build GRCh38
rs75615777 Chr.21: 32602382-32602382 on Build GRCh38

View Full Product Details