Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33765743-33766269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITSN1
Entrez Gene ID
6453
Gene Name
intersectin 1
Gene Aliases
ITSN, SH3D1A, SH3P17
UniGene
Hs.160324
Gene Chromosome Location
Chr.21: 33642400-33899861 on Build GRCh38
Transcripts
NM_001001132.1 NM_003024.2 XM_005261026.2 XM_005261027.1 XM_005261031.1 XM_011529691.1 XM_011529692.1 XM_011529693.2 XM_017028428.1 XM_017028429.1 XM_017028430.1 XM_017028431.1 XM_017028432.1 XM_017028433.1 XM_017028434.1 XM_017028435.1 XM_017028436.1 XM_017028437.1 XM_017028438.1 XM_017028439.1 XM_017028440.1 XM_017028441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7095782 c.859G>A p.D287N Chr.21: 33765945-33765945 on Build GRCh38
7095783 c.859G>A p.D287N Chr.21: 33765945-33765945 on Build GRCh38
3550430 c.794T>C p.L265P Chr.21: 33765880-33765880 on Build GRCh38
4002007 c.926+10A>G p.? Chr.21: 33766022-33766022 on Build GRCh38
4295802 c.835T>A p.F279I Chr.21: 33765921-33765921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367655089 Chr.21: 33765990-33765990 on Build GRCh38
rs926375004 Chr.21: 33766236-33766236 on Build GRCh38
rs746761496 Chr.21: 33765835-33765835 on Build GRCh38
rs750462238 Chr.21: 33765959-33765959 on Build GRCh38
rs758421806 Chr.21: 33765969-33765969 on Build GRCh38
rs747441208 Chr.21: 33766047-33766047 on Build GRCh38
rs1009386195 Chr.21: 33765785-33765785 on Build GRCh38
rs1016033971 Chr.21: 33766020-33766020 on Build GRCh38
rs926288590 Chr.21: 33765995-33765995 on Build GRCh38
rs765466442 Chr.21: 33765934-33765934 on Build GRCh38
rs1022417095 Chr.21: 33765786-33765786 on Build GRCh38
rs115018555 Chr.21: 33766075-33766075 on Build GRCh38
rs754984360 Chr.21: 33765989-33765989 on Build GRCh38
rs371591679 Chr.21: 33766138-33766138 on Build GRCh38
rs974850965 Chr.21: 33766221-33766221 on Build GRCh38
rs372747157 Chr.21: 33765863-33765863 on Build GRCh38
rs151199077 Chr.21: 33766059-33766059 on Build GRCh38
rs1056810089 Chr.21: 33766042-33766042 on Build GRCh38
rs770331415 Chr.21: 33766027-33766027 on Build GRCh38
rs1039410639 Chr.21: 33765831-33765831 on Build GRCh38
rs566751220 Chr.21: 33766163-33766163 on Build GRCh38
rs755791159 Chr.21: 33766012-33766012 on Build GRCh38
rs577365732 Chr.21: 33765871-33765872 on Build GRCh38
rs766538916 Chr.21: 33765972-33765972 on Build GRCh38
rs563181072 Chr.21: 33765770-33765770 on Build GRCh38
rs988756055 Chr.21: 33766088-33766088 on Build GRCh38
rs751448156 Chr.21: 33765981-33765981 on Build GRCh38
rs367792826 Chr.21: 33765853-33765853 on Build GRCh38
rs1028332854 Chr.21: 33765859-33765859 on Build GRCh38
rs773975634 Chr.21: 33766043-33766043 on Build GRCh38
rs764370155 Chr.21: 33766158-33766158 on Build GRCh38
rs913104697 Chr.21: 33765778-33765778 on Build GRCh38
rs182582846 Chr.21: 33766215-33766215 on Build GRCh38
rs747981470 Chr.21: 33765991-33765991 on Build GRCh38
rs987474121 Chr.21: 33765868-33765868 on Build GRCh38
rs201600778 Chr.21: 33765826-33765826 on Build GRCh38
rs531410475 Chr.21: 33766044-33766044 on Build GRCh38
rs370368854 Chr.21: 33765847-33765847 on Build GRCh38
rs772559655 Chr.21: 33765877-33765877 on Build GRCh38
rs920377572 Chr.21: 33765956-33765956 on Build GRCh38
rs754239354 Chr.21: 33765910-33765910 on Build GRCh38
rs761998738 Chr.21: 33765932-33765932 on Build GRCh38
rs766175904 Chr.21: 33765813-33765813 on Build GRCh38
rs974250742 Chr.21: 33766197-33766197 on Build GRCh38
rs760689150 Chr.21: 33765810-33765810 on Build GRCh38
rs147001871 Chr.21: 33766103-33766103 on Build GRCh38
rs147654153 Chr.21: 33765870-33765870 on Build GRCh38
rs930392753 Chr.21: 33765962-33765962 on Build GRCh38
rs201087456 Chr.21: 33765902-33765902 on Build GRCh38
rs970166141 Chr.21: 33766222-33766222 on Build GRCh38
rs1006326896 Chr.21: 33765947-33765947 on Build GRCh38
rs996944169 Chr.21: 33766136-33766136 on Build GRCh38
rs769253885 Chr.21: 33766048-33766048 on Build GRCh38
rs776868658 Chr.21: 33766119-33766120 on Build GRCh38
rs2248815 Chr.21: 33766022-33766022 on Build GRCh38
rs559874721 Chr.21: 33765865-33765865 on Build GRCh38
rs751770583 Chr.21: 33766188-33766188 on Build GRCh38
rs761300104 Chr.21: 33765898-33765898 on Build GRCh38
rs961639672 Chr.21: 33766190-33766190 on Build GRCh38
rs142196969 Chr.21: 33766052-33766052 on Build GRCh38
rs968117464 Chr.21: 33765816-33765816 on Build GRCh38
rs558161532 Chr.21: 33766128-33766128 on Build GRCh38
rs775871104 Chr.21: 33765895-33765895 on Build GRCh38
rs772804164 Chr.21: 33765866-33765866 on Build GRCh38
rs149112262 Chr.21: 33766015-33766015 on Build GRCh38
rs980501940 Chr.21: 33766227-33766227 on Build GRCh38

View Full Product Details