Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33767608-33768155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITSN1
Entrez Gene ID
6453
Gene Name
intersectin 1
Gene Aliases
ITSN, SH3D1A, SH3P17
UniGene
Hs.160324
Gene Chromosome Location
Chr.21: 33642400-33899861 on Build GRCh38
Transcripts
NM_001001132.1 NM_003024.2 XM_005261026.2 XM_005261027.1 XM_005261031.1 XM_011529691.1 XM_011529692.1 XM_011529693.2 XM_017028428.1 XM_017028429.1 XM_017028430.1 XM_017028431.1 XM_017028432.1 XM_017028433.1 XM_017028434.1 XM_017028435.1 XM_017028436.1 XM_017028437.1 XM_017028438.1 XM_017028439.1 XM_017028440.1 XM_017028441.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3550432 c.934C>T p.R312* Chr.21: 33767720-33767720 on Build GRCh38
7372774 c.992A>G p.E331G Chr.21: 33767778-33767778 on Build GRCh38
3423928 c.1032G>T p.K344N Chr.21: 33767818-33767818 on Build GRCh38
3423929 c.1032G>T p.K344N Chr.21: 33767818-33767818 on Build GRCh38
1211179 c.935G>A p.R312Q Chr.21: 33767721-33767721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760758707 Chr.21: 33767823-33767823 on Build GRCh38
rs967588148 Chr.21: 33767799-33767799 on Build GRCh38
rs183371865 Chr.21: 33767890-33767890 on Build GRCh38
rs778262651 Chr.21: 33767721-33767721 on Build GRCh38
rs1019557552 Chr.21: 33767638-33767638 on Build GRCh38
rs575623686 Chr.21: 33767883-33767883 on Build GRCh38
rs756429059 Chr.21: 33768034-33768034 on Build GRCh38
rs149644806 Chr.21: 33767652-33767652 on Build GRCh38
rs992885847 Chr.21: 33768037-33768037 on Build GRCh38
rs900804070 Chr.21: 33767903-33767903 on Build GRCh38
rs755981817 Chr.21: 33767702-33767702 on Build GRCh38
rs372922212 Chr.21: 33767675-33767675 on Build GRCh38
rs756809140 Chr.21: 33767870-33767870 on Build GRCh38
rs910682181 Chr.21: 33768123-33768123 on Build GRCh38
rs763947679 Chr.21: 33767839-33767839 on Build GRCh38
rs1006596967 Chr.21: 33767888-33767888 on Build GRCh38
rs937822406 Chr.21: 33768056-33768056 on Build GRCh38
rs773608079 Chr.21: 33767753-33767753 on Build GRCh38
rs766649165 Chr.21: 33767762-33767762 on Build GRCh38
rs777127226 Chr.21: 33767734-33767734 on Build GRCh38
rs952573141 Chr.21: 33767971-33767971 on Build GRCh38
rs368797638 Chr.21: 33767735-33767735 on Build GRCh38
rs994059491 Chr.21: 33767904-33767904 on Build GRCh38
rs778657875 Chr.21: 33767871-33767871 on Build GRCh38
rs769110337 Chr.21: 33767733-33767733 on Build GRCh38
rs774258924 Chr.21: 33767764-33767764 on Build GRCh38
rs936183396 Chr.21: 33768096-33768096 on Build GRCh38
rs112589405 Chr.21: 33767731-33767731 on Build GRCh38
rs143379999 Chr.21: 33767687-33767687 on Build GRCh38
rs933671636 Chr.21: 33767936-33767936 on Build GRCh38
rs6517192 Chr.21: 33768122-33768122 on Build GRCh38
rs753714251 Chr.21: 33767845-33767845 on Build GRCh38
rs375683794 Chr.21: 33767679-33767679 on Build GRCh38
rs770842320 Chr.21: 33767663-33767664 on Build GRCh38
rs529968953 Chr.21: 33768046-33768046 on Build GRCh38
rs1019702021 Chr.21: 33767892-33767892 on Build GRCh38
rs773411625 Chr.21: 33767669-33767669 on Build GRCh38
rs79522704 Chr.21: 33767749-33767749 on Build GRCh38
rs115178190 Chr.21: 33767703-33767703 on Build GRCh38
rs756971499 Chr.21: 33767718-33767718 on Build GRCh38
rs188771331 Chr.21: 33768085-33768085 on Build GRCh38
rs141514977 Chr.21: 33768043-33768043 on Build GRCh38
rs538862072 Chr.21: 33767685-33767685 on Build GRCh38
rs752664591 Chr.21: 33767786-33767786 on Build GRCh38
rs765386856 Chr.21: 33767662-33767662 on Build GRCh38
rs908471713 Chr.21: 33767992-33767992 on Build GRCh38
rs376858445 Chr.21: 33767723-33767723 on Build GRCh38
rs763065231 Chr.21: 33767759-33767759 on Build GRCh38
rs117832917 Chr.21: 33767915-33767915 on Build GRCh38
rs576846041 Chr.21: 33768027-33768027 on Build GRCh38
rs571728817 Chr.21: 33767661-33767661 on Build GRCh38
rs759669553 Chr.21: 33767783-33767783 on Build GRCh38
rs1055899189 Chr.21: 33768087-33768087 on Build GRCh38
rs933640829 Chr.21: 33767658-33767658 on Build GRCh38
rs540886694 Chr.21: 33768042-33768042 on Build GRCh38
rs755617092 Chr.21: 33768023-33768023 on Build GRCh38
rs751711922 Chr.21: 33767682-33767682 on Build GRCh38
rs972636562 Chr.21: 33767649-33767649 on Build GRCh38
rs763776781 Chr.21: 33767838-33767838 on Build GRCh38
rs748884589 Chr.21: 33767707-33767707 on Build GRCh38
rs926173969 Chr.21: 33768068-33768068 on Build GRCh38
rs965672366 Chr.21: 33767648-33767648 on Build GRCh38
rs752478539 Chr.21: 33767693-33767693 on Build GRCh38

View Full Product Details