Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 7758084-7758634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM11A
Entrez Gene ID
54065
Gene Name
small integral membrane protein 11A
Gene Aliases
C21orf51, FAM165B, SMIM11
UniGene
Hs.656195
Gene Chromosome Location
Chr.21: 34375450-34402777 on Build GRCh38
Transcripts
NM_058182.4 XM_017028397.1 XM_017028398.1 XM_017028399.1 NM_001313692.1 XM_017028203.1 XM_017028204.1 XM_017028205.1

Gene Symbol
SMIM11B
Entrez Gene ID
102723553
Gene Name
small integral membrane protein 11B
Gene Aliases
-
UniGene
Hs.656195
Gene Chromosome Location
Chr.21: 7744951-7772253 on Build GRCh38
Transcripts
NM_058182.4 XM_017028397.1 XM_017028398.1 XM_017028399.1 NM_001313692.1 XM_017028203.1 XM_017028204.1 XM_017028205.1

SNP Detail

SNP ID Chromosome Location
rs114933254 Chr.21: 34389051-34389051 on Build GRCh38
rs956631603 Chr.21: 34389124-34389124 on Build GRCh38
rs148086708 Chr.21: 34388788-34388788 on Build GRCh38
rs555620869 Chr.21: 34389054-34389054 on Build GRCh38
rs539823135 Chr.21: 34389022-34389022 on Build GRCh38
rs946473503 Chr.21: 34388974-34388976 on Build GRCh38
rs557470807 Chr.21: 34388991-34388991 on Build GRCh38
rs370461308 Chr.21: 34388780-34388780 on Build GRCh38
rs142795800 Chr.21: 34388937-34388937 on Build GRCh38
rs75196527 Chr.21: 34389101-34389101 on Build GRCh38
rs944417730 Chr.21: 34388992-34388992 on Build GRCh38
rs562610463 Chr.21: 34389100-34389100 on Build GRCh38
rs566773071 Chr.21: 34388784-34388784 on Build GRCh38
rs113386464 Chr.21: 34389023-34389023 on Build GRCh38
rs773287998 Chr.21: 34388936-34388936 on Build GRCh38
rs905177028 Chr.21: 34389078-34389078 on Build GRCh38
rs199943330 Chr.21: 34389076-34389076 on Build GRCh38
rs1050073302 Chr.21: 34388974-34388974 on Build GRCh38
rs768538202 Chr.21: 34388919-34388919 on Build GRCh38
rs987897172 Chr.21: 34389129-34389129 on Build GRCh38
rs538016374 Chr.21: 34388939-34388939 on Build GRCh38
rs878908259 Chr.21: 34388836-34388836 on Build GRCh38
rs76497658 Chr.21: 34388775-34388775 on Build GRCh38
rs145045485 Chr.21: 34389003-34389003 on Build GRCh38
rs575610648 Chr.21: 34389020-34389020 on Build GRCh38
rs573945445 Chr.21: 34389061-34389061 on Build GRCh38
rs143841894 Chr.21: 34388743-34388743 on Build GRCh38
rs367833101 Chr.21: 34389047-34389047 on Build GRCh38
rs749035384 Chr.21: 34388871-34388871 on Build GRCh38
rs771204011 Chr.21: 34388793-34388793 on Build GRCh38
rs994362788 Chr.21: 34388640-34388640 on Build GRCh38
rs771313840 Chr.21: 34388938-34388938 on Build GRCh38
rs549213773 Chr.21: 34388873-34388873 on Build GRCh38
rs956519363 Chr.21: 34388905-34388905 on Build GRCh38
rs527610876 Chr.21: 34388665-34388665 on Build GRCh38
rs775556115 Chr.21: 34388857-34388857 on Build GRCh38
rs1003568612 Chr.21: 34389081-34389081 on Build GRCh38
rs1025731743 Chr.21: 34388738-34388738 on Build GRCh38
rs781008403 Chr.21: 34388727-34388729 on Build GRCh38
rs201503145 Chr.21: 34389101-34389101 on Build GRCh38
rs35568330 Chr.21: 34388817-34388818 on Build GRCh38

View Full Product Details