Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39661597-39662104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT5
Entrez Gene ID
10317
Gene Name
beta-1,3-galactosyltransferase 5
Gene Aliases
B3GalT-V, B3GalTx, B3T5, GLCT5, beta-1,3-GalTase 5, beta-3-Gx-T5, beta3Gal-T5
UniGene
Hs.733638
Gene Chromosome Location
Chr.21: 39612940-39662889 on Build GRCh38
Transcripts
NM_001278650.1 NM_006057.2 NM_033170.2 NM_033171.2 NM_033172.2 XM_017028227.1 XM_017028228.1 XM_017028229.1 XM_017028230.1 XM_017028231.1 XM_017028232.1 XM_017028233.1 XM_017028234.1 XM_017028235.1 XM_017028236.1 XM_017028237.1 XM_017028238.1 XM_017028239.1 XM_017028240.1 XM_017028241.1

SNP Detail

SNP ID Chromosome Location
rs1029001787 Chr.21: 39661646-39661646 on Build GRCh38
rs944531965 Chr.21: 39661986-39661986 on Build GRCh38
rs1041019648 Chr.21: 39661951-39661951 on Build GRCh38
rs527829871 Chr.21: 39661808-39661808 on Build GRCh38
rs114221867 Chr.21: 39661764-39661764 on Build GRCh38
rs550224771 Chr.21: 39661910-39661910 on Build GRCh38
rs1011060701 Chr.21: 39661835-39661835 on Build GRCh38
rs780218357 Chr.21: 39661627-39661627 on Build GRCh38
rs543319531 Chr.21: 39661676-39661676 on Build GRCh38
rs1044899129 Chr.21: 39661653-39661653 on Build GRCh38
rs4816637 Chr.21: 39661858-39661858 on Build GRCh38
rs909182 Chr.21: 39661889-39661889 on Build GRCh38
rs887123432 Chr.21: 39661688-39661688 on Build GRCh38
rs191653482 Chr.21: 39661685-39661685 on Build GRCh38
rs949163373 Chr.21: 39661851-39661851 on Build GRCh38
rs959042593 Chr.21: 39661677-39661678 on Build GRCh38
rs904707363 Chr.21: 39661980-39661980 on Build GRCh38
rs1031934229 Chr.21: 39662056-39662056 on Build GRCh38
rs966461164 Chr.21: 39661665-39661665 on Build GRCh38
rs983407518 Chr.21: 39661945-39661945 on Build GRCh38
rs574103076 Chr.21: 39661868-39661869 on Build GRCh38
rs1003216524 Chr.21: 39661868-39661868 on Build GRCh38
rs114976501 Chr.21: 39661813-39661813 on Build GRCh38
rs1041110954 Chr.21: 39662003-39662003 on Build GRCh38
rs866813331 Chr.21: 39662060-39662060 on Build GRCh38
rs565221258 Chr.21: 39661779-39661779 on Build GRCh38
rs931112137 Chr.21: 39661873-39661873 on Build GRCh38
rs996924966 Chr.21: 39661961-39661961 on Build GRCh38
rs997462543 Chr.21: 39661762-39661762 on Build GRCh38
rs188415216 Chr.21: 39662029-39662029 on Build GRCh38
rs552190522 Chr.21: 39661923-39661923 on Build GRCh38
rs1004146734 Chr.21: 39661720-39661720 on Build GRCh38
rs956360771 Chr.21: 39661825-39661825 on Build GRCh38
rs574345008 Chr.21: 39661675-39661675 on Build GRCh38
rs997073390 Chr.21: 39661636-39661636 on Build GRCh38
rs1019660481 Chr.21: 39661744-39661744 on Build GRCh38
rs565635399 Chr.21: 39661960-39661960 on Build GRCh38
rs1034737739 Chr.21: 39661871-39661871 on Build GRCh38
rs949251861 Chr.21: 39661618-39661618 on Build GRCh38
rs1048318169 Chr.21: 39661875-39661875 on Build GRCh38
rs940057832 Chr.21: 39661898-39661898 on Build GRCh38
rs769244655 Chr.21: 39661722-39661722 on Build GRCh38
rs905149883 Chr.21: 39661657-39661657 on Build GRCh38
rs894083765 Chr.21: 39661827-39661827 on Build GRCh38
rs543188784 Chr.21: 39661857-39661857 on Build GRCh38
rs554300504 Chr.21: 39661967-39661967 on Build GRCh38
rs926464394 Chr.21: 39661870-39661870 on Build GRCh38
rs534415158 Chr.21: 39661966-39661966 on Build GRCh38
rs545067597 Chr.21: 39662081-39662081 on Build GRCh38
rs368118002 Chr.21: 39662068-39662068 on Build GRCh38
rs183429605 Chr.21: 39661892-39661892 on Build GRCh38
rs531628024 Chr.21: 39661758-39661758 on Build GRCh38
rs770872324 Chr.21: 39661807-39661807 on Build GRCh38
rs908608907 Chr.21: 39661762-39661763 on Build GRCh38
rs920507845 Chr.21: 39661725-39661725 on Build GRCh38
rs917709196 Chr.21: 39661850-39661850 on Build GRCh38
rs574305632 Chr.21: 39661981-39661981 on Build GRCh38
rs561469496 Chr.21: 39661834-39661834 on Build GRCh38
rs538239275 Chr.21: 39661896-39661896 on Build GRCh38
rs969402543 Chr.21: 39661797-39661797 on Build GRCh38
rs777775668 Chr.21: 39661663-39661663 on Build GRCh38
rs556951831 Chr.21: 39662064-39662064 on Build GRCh38
rs958193774 Chr.21: 39661656-39661656 on Build GRCh38

View Full Product Details