Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89262591-89263093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP5
Entrez Gene ID
115362
Gene Name
guanylate binding protein 5
Gene Aliases
GBP-5
UniGene
Hs.513726
Gene Chromosome Location
Chr.1: 89258950-89272861 on Build GRCh38
Transcripts
NM_001134486.2 NM_052942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669810 c.1363-2A>G p.? Chr.1: 89262787-89262787 on Build GRCh38
6460764 c.1427C>G p.T476S Chr.1: 89262721-89262721 on Build GRCh38
396495 c.1427C>A p.T476N Chr.1: 89262721-89262721 on Build GRCh38
4942755 c.1439T>C p.L480P Chr.1: 89262709-89262709 on Build GRCh38
1602762 c.1379A>T p.Q460L Chr.1: 89262769-89262769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905448937 Chr.1: 89263026-89263026 on Build GRCh38
rs774373746 Chr.1: 89262718-89262718 on Build GRCh38
rs1014374368 Chr.1: 89263022-89263022 on Build GRCh38
rs373455084 Chr.1: 89263025-89263025 on Build GRCh38
rs919933220 Chr.1: 89262665-89262665 on Build GRCh38
rs867042587 Chr.1: 89262894-89262894 on Build GRCh38
rs369628117 Chr.1: 89262939-89262939 on Build GRCh38
rs766922146 Chr.1: 89262747-89262747 on Build GRCh38
rs1042731396 Chr.1: 89262866-89262866 on Build GRCh38
rs377675257 Chr.1: 89262743-89262743 on Build GRCh38
rs779147050 Chr.1: 89262690-89262691 on Build GRCh38
rs548056021 Chr.1: 89262870-89262870 on Build GRCh38
rs768565499 Chr.1: 89262832-89262832 on Build GRCh38
rs144341165 Chr.1: 89263056-89263056 on Build GRCh38
rs145775181 Chr.1: 89262754-89262754 on Build GRCh38
rs895547076 Chr.1: 89263020-89263020 on Build GRCh38
rs932548736 Chr.1: 89262944-89262944 on Build GRCh38
rs549723161 Chr.1: 89262795-89262795 on Build GRCh38
rs748318010 Chr.1: 89262837-89262837 on Build GRCh38
rs181786126 Chr.1: 89262732-89262732 on Build GRCh38
rs909678994 Chr.1: 89262974-89262974 on Build GRCh38
rs140084693 Chr.1: 89262900-89262901 on Build GRCh38
rs752232079 Chr.1: 89262765-89262765 on Build GRCh38
rs753388330 Chr.1: 89262648-89262648 on Build GRCh38
rs750518373 Chr.1: 89262968-89262968 on Build GRCh38
rs779537685 Chr.1: 89262636-89262636 on Build GRCh38
rs953766801 Chr.1: 89262927-89262927 on Build GRCh38
rs976762193 Chr.1: 89262914-89262914 on Build GRCh38
rs762091110 Chr.1: 89262638-89262638 on Build GRCh38
rs903505709 Chr.1: 89262873-89262873 on Build GRCh38
rs939773491 Chr.1: 89262977-89262977 on Build GRCh38
rs1036755743 Chr.1: 89262999-89262999 on Build GRCh38
rs1027965044 Chr.1: 89262903-89262903 on Build GRCh38
rs747865151 Chr.1: 89262671-89262671 on Build GRCh38
rs973397923 Chr.1: 89262620-89262620 on Build GRCh38
rs950385917 Chr.1: 89262963-89262963 on Build GRCh38
rs148530854 Chr.1: 89263044-89263044 on Build GRCh38
rs142498526 Chr.1: 89262745-89262745 on Build GRCh38
rs961884508 Chr.1: 89262955-89262955 on Build GRCh38
rs777704631 Chr.1: 89262789-89262789 on Build GRCh38
rs758061499 Chr.1: 89262674-89262674 on Build GRCh38
rs775677633 Chr.1: 89262699-89262699 on Build GRCh38
rs771056238 Chr.1: 89262693-89262693 on Build GRCh38
rs1002370781 Chr.1: 89263027-89263027 on Build GRCh38
rs758891023 Chr.1: 89263049-89263049 on Build GRCh38
rs931337397 Chr.1: 89262681-89262681 on Build GRCh38
rs774551885 Chr.1: 89262833-89262833 on Build GRCh38
rs1009486212 Chr.1: 89262840-89262840 on Build GRCh38
rs548599507 Chr.1: 89262724-89262724 on Build GRCh38
rs754405892 Chr.1: 89262756-89262756 on Build GRCh38
rs757235285 Chr.1: 89262796-89262796 on Build GRCh38
rs532184304 Chr.1: 89262967-89262967 on Build GRCh38
rs745827987 Chr.1: 89262825-89262825 on Build GRCh38
rs755178093 Chr.1: 89262789-89262789 on Build GRCh38
rs530157452 Chr.1: 89262660-89262660 on Build GRCh38
rs868042226 Chr.1: 89262682-89262682 on Build GRCh38
rs758125506 Chr.1: 89262767-89262767 on Build GRCh38
rs768799881 Chr.1: 89262704-89262704 on Build GRCh38
rs781223878 Chr.1: 89262814-89262814 on Build GRCh38
rs1009678219 Chr.1: 89263062-89263062 on Build GRCh38
rs370238022 Chr.1: 89262759-89262759 on Build GRCh38
rs772497791 Chr.1: 89262808-89262809 on Build GRCh38
rs969662808 Chr.1: 89262706-89262706 on Build GRCh38
rs1044651837 Chr.1: 89262986-89262986 on Build GRCh38
rs1025670059 Chr.1: 89262700-89262700 on Build GRCh38
rs377076158 Chr.1: 89262731-89262731 on Build GRCh38
rs778381367 Chr.1: 89262661-89262661 on Build GRCh38
rs192198096 Chr.1: 89262943-89262943 on Build GRCh38
rs746928691 Chr.1: 89262683-89262683 on Build GRCh38

View Full Product Details