Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29308377-29308882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L1
Entrez Gene ID
10634
Gene Name
growth arrest specific 2 like 1
Gene Aliases
GAR22
UniGene
Hs.322852
Gene Chromosome Location
Chr.22: 29306618-29312789 on Build GRCh38
Transcripts
NM_001278730.1 NM_006478.4 NM_152236.2 XM_011529824.1 XM_011529825.1 XM_017028533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669382 c.561C>T p.T187T Chr.22: 29308666-29308666 on Build GRCh38
4913440 c.375G>T p.E125D Chr.22: 29308480-29308480 on Build GRCh38
6669395 c.528C>T p.N176N Chr.22: 29308633-29308633 on Build GRCh38
5000185 c.298C>A p.R100S Chr.22: 29308403-29308403 on Build GRCh38
1415495 c.566C>A p.P189H Chr.22: 29308671-29308671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750346629 Chr.22: 29308583-29308583 on Build GRCh38
rs755932088 Chr.22: 29308594-29308594 on Build GRCh38
rs934784187 Chr.22: 29308461-29308461 on Build GRCh38
rs368533422 Chr.22: 29308685-29308685 on Build GRCh38
rs772201234 Chr.22: 29308528-29308528 on Build GRCh38
rs770295311 Chr.22: 29308717-29308717 on Build GRCh38
rs572530394 Chr.22: 29308551-29308551 on Build GRCh38
rs778008336 Chr.22: 29308697-29308697 on Build GRCh38
rs541058137 Chr.22: 29308698-29308698 on Build GRCh38
rs772163549 Chr.22: 29308427-29308427 on Build GRCh38
rs930946576 Chr.22: 29308601-29308601 on Build GRCh38
rs762725485 Chr.22: 29308658-29308658 on Build GRCh38
rs753741281 Chr.22: 29308782-29308782 on Build GRCh38
rs750388383 Chr.22: 29308673-29308673 on Build GRCh38
rs747541065 Chr.22: 29308537-29308537 on Build GRCh38
rs751556696 Chr.22: 29308466-29308466 on Build GRCh38
rs1019814796 Chr.22: 29308768-29308768 on Build GRCh38
rs529843808 Chr.22: 29308771-29308771 on Build GRCh38
rs371142942 Chr.22: 29308476-29308476 on Build GRCh38
rs774216974 Chr.22: 29308755-29308755 on Build GRCh38
rs919640330 Chr.22: 29308576-29308576 on Build GRCh38
rs779045810 Chr.22: 29308501-29308501 on Build GRCh38
rs774125414 Chr.22: 29308667-29308667 on Build GRCh38
rs970471152 Chr.22: 29308859-29308859 on Build GRCh38
rs781274105 Chr.22: 29308702-29308702 on Build GRCh38
rs777169933 Chr.22: 29308542-29308542 on Build GRCh38
rs768722703 Chr.22: 29308403-29308403 on Build GRCh38
rs759856018 Chr.22: 29308547-29308547 on Build GRCh38
rs986943182 Chr.22: 29308462-29308462 on Build GRCh38
rs754742383 Chr.22: 29308696-29308696 on Build GRCh38
rs112582307 Chr.22: 29308793-29308793 on Build GRCh38
rs765627874 Chr.22: 29308550-29308550 on Build GRCh38
rs56037813 Chr.22: 29308672-29308672 on Build GRCh38
rs908167095 Chr.22: 29308857-29308857 on Build GRCh38
rs770067614 Chr.22: 29308647-29308647 on Build GRCh38
rs771375346 Chr.22: 29308539-29308539 on Build GRCh38
rs1012193224 Chr.22: 29308455-29308455 on Build GRCh38
rs151047568 Chr.22: 29308419-29308419 on Build GRCh38
rs142321804 Chr.22: 29308456-29308456 on Build GRCh38
rs545449101 Chr.22: 29308591-29308591 on Build GRCh38
rs756025463 Chr.22: 29308683-29308683 on Build GRCh38
rs780305998 Chr.22: 29308721-29308721 on Build GRCh38
rs771498880 Chr.22: 29308766-29308766 on Build GRCh38
rs764843628 Chr.22: 29308439-29308439 on Build GRCh38
rs775741782 Chr.22: 29308433-29308438 on Build GRCh38
rs202161456 Chr.22: 29308433-29308433 on Build GRCh38
rs1011521148 Chr.22: 29308679-29308679 on Build GRCh38
rs763124341 Chr.22: 29308760-29308779 on Build GRCh38
rs772715078 Chr.22: 29308769-29308769 on Build GRCh38
rs1051889004 Chr.22: 29308827-29308827 on Build GRCh38
rs769113573 Chr.22: 29308743-29308743 on Build GRCh38
rs746371311 Chr.22: 29308642-29308642 on Build GRCh38
rs900683165 Chr.22: 29308792-29308792 on Build GRCh38
rs772878158 Chr.22: 29308764-29308764 on Build GRCh38
rs950717626 Chr.22: 29308606-29308606 on Build GRCh38
rs746365930 Chr.22: 29308715-29308715 on Build GRCh38
rs191844093 Chr.22: 29308637-29308637 on Build GRCh38
rs148265669 Chr.22: 29308607-29308607 on Build GRCh38
rs567152329 Chr.22: 29308722-29308722 on Build GRCh38
rs140711281 Chr.22: 29308534-29308534 on Build GRCh38
rs777070349 Chr.22: 29308640-29308640 on Build GRCh38
rs767613647 Chr.22: 29308578-29308578 on Build GRCh38
rs757284019 Chr.22: 29308573-29308573 on Build GRCh38
rs982460750 Chr.22: 29308849-29308849 on Build GRCh38
rs755917216 Chr.22: 29308493-29308493 on Build GRCh38
rs769281251 Chr.22: 29308634-29308634 on Build GRCh38
rs751074390 Chr.22: 29308572-29308572 on Build GRCh38
rs766730164 Chr.22: 29308435-29308435 on Build GRCh38
rs372396375 Chr.22: 29308710-29308710 on Build GRCh38
rs763701756 Chr.22: 29308570-29308570 on Build GRCh38
rs766096399 Chr.22: 29308688-29308688 on Build GRCh38
rs759252440 Chr.22: 29308437-29308437 on Build GRCh38
rs749720814 Chr.22: 29308729-29308729 on Build GRCh38
rs934316495 Chr.22: 29308412-29308412 on Build GRCh38
rs147637354 Chr.22: 29308770-29308770 on Build GRCh38
rs201596421 Chr.22: 29308652-29308652 on Build GRCh38
rs781164860 Chr.22: 29308477-29308477 on Build GRCh38
rs757912067 Chr.22: 29308440-29308440 on Build GRCh38
rs763791690 Chr.22: 29308666-29308666 on Build GRCh38
rs1020177406 Chr.22: 29308513-29308513 on Build GRCh38
rs754159174 Chr.22: 29308436-29308436 on Build GRCh38
rs748400918 Chr.22: 29308509-29308509 on Build GRCh38
rs758574943 Chr.22: 29308619-29308619 on Build GRCh38
rs914189363 Chr.22: 29308713-29308713 on Build GRCh38
rs573277869 Chr.22: 29308584-29308584 on Build GRCh38
rs147668503 Chr.22: 29308426-29308426 on Build GRCh38
rs1017779104 Chr.22: 29308472-29308472 on Build GRCh38
rs113516975 Chr.22: 29308609-29308609 on Build GRCh38
rs762607803 Chr.22: 29308553-29308553 on Build GRCh38
rs371060454 Chr.22: 29308708-29308708 on Build GRCh38
rs369870873 Chr.22: 29308628-29308628 on Build GRCh38
rs745602248 Chr.22: 29308487-29308487 on Build GRCh38
rs1027634800 Chr.22: 29308785-29308785 on Build GRCh38
rs753617015 Chr.22: 29308692-29308692 on Build GRCh38
rs964334817 Chr.22: 29308521-29308521 on Build GRCh38
rs760119921 Chr.22: 29308843-29308843 on Build GRCh38
rs757383475 Chr.22: 29308701-29308701 on Build GRCh38
rs188482420 Chr.22: 29308633-29308633 on Build GRCh38

View Full Product Details