Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31933729-31934232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf24
Entrez Gene ID
25775
Gene Name
chromosome 22 open reading frame 24
Gene Aliases
HSN44A4A
UniGene
Hs.627602
Gene Chromosome Location
Chr.22: 31933521-31945362 on Build GRCh38
Transcripts
NM_001302819.1 NM_001302820.1 NM_015372.2 NR_126507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002165 c.472T>C p.S158P Chr.22: 31934129-31934129 on Build GRCh38
7358418 c.625G>C p.E209Q Chr.22: 31934177-31934177 on Build GRCh38
5527714 c.609G>A p.R203R Chr.22: 31934193-31934193 on Build GRCh38
4002164 c.673T>C p.S225P Chr.22: 31934129-31934129 on Build GRCh38
7207306 c.475G>A p.E159K Chr.22: 31934126-31934126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747496700 Chr.22: 31934186-31934186 on Build GRCh38
rs955403763 Chr.22: 31933806-31933806 on Build GRCh38
rs915114625 Chr.22: 31933848-31933848 on Build GRCh38
rs912270369 Chr.22: 31934047-31934047 on Build GRCh38
rs555320546 Chr.22: 31934077-31934077 on Build GRCh38
rs559839229 Chr.22: 31933759-31933759 on Build GRCh38
rs968285230 Chr.22: 31933851-31933851 on Build GRCh38
rs1022112470 Chr.22: 31934074-31934074 on Build GRCh38
rs745867164 Chr.22: 31933908-31933908 on Build GRCh38
rs748877527 Chr.22: 31934160-31934160 on Build GRCh38
rs763914735 Chr.22: 31934099-31934099 on Build GRCh38
rs981414425 Chr.22: 31933983-31933983 on Build GRCh38
rs368955677 Chr.22: 31933843-31933843 on Build GRCh38
rs190337370 Chr.22: 31934005-31934005 on Build GRCh38
rs891379151 Chr.22: 31933844-31933844 on Build GRCh38
rs923823895 Chr.22: 31933865-31933865 on Build GRCh38
rs955893239 Chr.22: 31933991-31933991 on Build GRCh38
rs200151600 Chr.22: 31934066-31934066 on Build GRCh38
rs758334817 Chr.22: 31934097-31934097 on Build GRCh38
rs564939798 Chr.22: 31933891-31933891 on Build GRCh38
rs932337195 Chr.22: 31933807-31933807 on Build GRCh38
rs776978565 Chr.22: 31934187-31934187 on Build GRCh38
rs763209664 Chr.22: 31934198-31934198 on Build GRCh38
rs569737691 Chr.22: 31934009-31934009 on Build GRCh38
rs1018332811 Chr.22: 31933830-31933830 on Build GRCh38
rs994732240 Chr.22: 31934016-31934016 on Build GRCh38
rs373251129 Chr.22: 31934090-31934090 on Build GRCh38
rs558782433 Chr.22: 31934026-31934026 on Build GRCh38
rs947819534 Chr.22: 31933900-31933900 on Build GRCh38
rs566109175 Chr.22: 31934031-31934031 on Build GRCh38
rs745433608 Chr.22: 31934129-31934129 on Build GRCh38
rs764013005 Chr.22: 31934199-31934199 on Build GRCh38
rs892785458 Chr.22: 31933783-31933783 on Build GRCh38
rs978061747 Chr.22: 31933862-31933862 on Build GRCh38
rs1054159471 Chr.22: 31933762-31933762 on Build GRCh38
rs765402679 Chr.22: 31934191-31934191 on Build GRCh38
rs944280412 Chr.22: 31934058-31934058 on Build GRCh38
rs369831055 Chr.22: 31934070-31934070 on Build GRCh38
rs529861139 Chr.22: 31933987-31933987 on Build GRCh38
rs527279753 Chr.22: 31933899-31933899 on Build GRCh38
rs755611464 Chr.22: 31934139-31934139 on Build GRCh38
rs923854239 Chr.22: 31934114-31934114 on Build GRCh38
rs544110613 Chr.22: 31934120-31934120 on Build GRCh38
rs762553045 Chr.22: 31934127-31934127 on Build GRCh38
rs759500515 Chr.22: 31934078-31934078 on Build GRCh38
rs567177014 Chr.22: 31933964-31933964 on Build GRCh38
rs755694468 Chr.22: 31934129-31934131 on Build GRCh38
rs960579075 Chr.22: 31934115-31934115 on Build GRCh38
rs772442193 Chr.22: 31934162-31934162 on Build GRCh38
rs1022848287 Chr.22: 31933975-31933975 on Build GRCh38
rs773513791 Chr.22: 31934167-31934167 on Build GRCh38
rs114976638 Chr.22: 31934103-31934103 on Build GRCh38
rs182167720 Chr.22: 31934076-31934076 on Build GRCh38
rs994468389 Chr.22: 31933776-31933776 on Build GRCh38
rs1025387043 Chr.22: 31934130-31934130 on Build GRCh38
rs773716607 Chr.22: 31934068-31934068 on Build GRCh38
rs775859524 Chr.22: 31934193-31934193 on Build GRCh38
rs914335862 Chr.22: 31934025-31934025 on Build GRCh38
rs769523123 Chr.22: 31933934-31933934 on Build GRCh38
rs750150271 Chr.22: 31934127-31934129 on Build GRCh38
rs369126906 Chr.22: 31934190-31934190 on Build GRCh38
rs971716551 Chr.22: 31933978-31933978 on Build GRCh38
rs547298664 Chr.22: 31933963-31933963 on Build GRCh38
rs767409062 Chr.22: 31934092-31934095 on Build GRCh38
rs964007958 Chr.22: 31934069-31934069 on Build GRCh38
rs1056101118 Chr.22: 31933875-31933875 on Build GRCh38
rs779750860 Chr.22: 31934155-31934155 on Build GRCh38
rs992068429 Chr.22: 31934179-31934179 on Build GRCh38
rs1012676670 Chr.22: 31933966-31933966 on Build GRCh38
rs113741815 Chr.22: 31934017-31934017 on Build GRCh38
rs897362115 Chr.22: 31933931-31933931 on Build GRCh38
rs1026298833 Chr.22: 31933803-31933803 on Build GRCh38
rs757185920 Chr.22: 31934087-31934091 on Build GRCh38
rs939015598 Chr.22: 31933866-31933866 on Build GRCh38
rs970779649 Chr.22: 31934182-31934182 on Build GRCh38
rs1027097642 Chr.22: 31933942-31933942 on Build GRCh38
rs749491160 Chr.22: 31933932-31933932 on Build GRCh38

View Full Product Details