Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35262794-35263292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGXB4
Entrez Gene ID
10042
Gene Name
HMG-box containing 4
Gene Aliases
HMG2L1, HMGBCG, THC211630
UniGene
Hs.588815
Gene Chromosome Location
Chr.22: 35257274-35295807 on Build GRCh38
Transcripts
NM_001003681.2 NR_027780.1 XM_006724100.3 XM_006724101.3 XM_006724102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1033725 c.77C>A p.A26D Chr.22: 35263123-35263123 on Build GRCh38
281731 c.161C>A p.S54Y Chr.22: 35263207-35263207 on Build GRCh38
4690761 c.117delT p.L41fs*1 Chr.22: 35263163-35263163 on Build GRCh38
5366690 c.114A>C p.K38N Chr.22: 35263160-35263160 on Build GRCh38
1033727 c.119T>G p.F40C Chr.22: 35263165-35263165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753561585 Chr.22: 35263257-35263257 on Build GRCh38
rs764848286 Chr.22: 35263212-35263214 on Build GRCh38
rs374024629 Chr.22: 35263127-35263127 on Build GRCh38
rs375108602 Chr.22: 35263120-35263120 on Build GRCh38
rs374037009 Chr.22: 35263149-35263149 on Build GRCh38
rs1007474853 Chr.22: 35262968-35262968 on Build GRCh38
rs767336493 Chr.22: 35263241-35263241 on Build GRCh38
rs759138844 Chr.22: 35263011-35263011 on Build GRCh38
rs765396511 Chr.22: 35263087-35263087 on Build GRCh38
rs775566830 Chr.22: 35263150-35263150 on Build GRCh38
rs909518663 Chr.22: 35262936-35262936 on Build GRCh38
rs951183410 Chr.22: 35263199-35263199 on Build GRCh38
rs761769506 Chr.22: 35263201-35263201 on Build GRCh38
rs181553597 Chr.22: 35262842-35262842 on Build GRCh38
rs754147563 Chr.22: 35263005-35263005 on Build GRCh38
rs939523354 Chr.22: 35262830-35262830 on Build GRCh38
rs934142739 Chr.22: 35263065-35263065 on Build GRCh38
rs926369030 Chr.22: 35262885-35262885 on Build GRCh38
rs752583927 Chr.22: 35263050-35263050 on Build GRCh38
rs866275854 Chr.22: 35263091-35263091 on Build GRCh38
rs367894563 Chr.22: 35263254-35263254 on Build GRCh38
rs112851119 Chr.22: 35263211-35263211 on Build GRCh38
rs755574573 Chr.22: 35263246-35263246 on Build GRCh38
rs757128285 Chr.22: 35263102-35263102 on Build GRCh38
rs763889473 Chr.22: 35263192-35263192 on Build GRCh38
rs771528182 Chr.22: 35263137-35263137 on Build GRCh38
rs1009886623 Chr.22: 35262834-35262834 on Build GRCh38
rs765342743 Chr.22: 35263029-35263029 on Build GRCh38
rs761917915 Chr.22: 35263078-35263078 on Build GRCh38
rs981891753 Chr.22: 35262883-35262883 on Build GRCh38
rs764081484 Chr.22: 35263071-35263071 on Build GRCh38
rs755841286 Chr.22: 35263110-35263110 on Build GRCh38
rs998447064 Chr.22: 35263044-35263044 on Build GRCh38
rs532050657 Chr.22: 35262881-35262881 on Build GRCh38
rs772573363 Chr.22: 35263129-35263129 on Build GRCh38
rs983806909 Chr.22: 35262934-35262934 on Build GRCh38
rs527722937 Chr.22: 35263244-35263244 on Build GRCh38
rs983906624 Chr.22: 35263236-35263236 on Build GRCh38
rs551704821 Chr.22: 35262957-35262957 on Build GRCh38
rs9610276 Chr.22: 35262849-35262849 on Build GRCh38
rs369944150 Chr.22: 35263122-35263122 on Build GRCh38
rs780775000 Chr.22: 35263107-35263107 on Build GRCh38
rs747543978 Chr.22: 35263136-35263136 on Build GRCh38
rs369654023 Chr.22: 35262868-35262868 on Build GRCh38
rs577501174 Chr.22: 35262860-35262860 on Build GRCh38
rs184356295 Chr.22: 35263015-35263015 on Build GRCh38
rs1010973796 Chr.22: 35262882-35262882 on Build GRCh38
rs776809462 Chr.22: 35263138-35263138 on Build GRCh38
rs759510429 Chr.22: 35263144-35263144 on Build GRCh38
rs1043779197 Chr.22: 35262903-35262903 on Build GRCh38
rs959071442 Chr.22: 35262919-35262919 on Build GRCh38
rs79846609 Chr.22: 35263143-35263143 on Build GRCh38
rs763094582 Chr.22: 35263188-35263188 on Build GRCh38
rs778316700 Chr.22: 35263263-35263263 on Build GRCh38
rs779559800 Chr.22: 35263112-35263112 on Build GRCh38
rs940313761 Chr.22: 35262960-35262960 on Build GRCh38
rs778229270 Chr.22: 35263133-35263133 on Build GRCh38
rs751304115 Chr.22: 35263198-35263198 on Build GRCh38
rs750427293 Chr.22: 35263193-35263193 on Build GRCh38
rs150842119 Chr.22: 35263108-35263108 on Build GRCh38
rs547556531 Chr.22: 35263262-35263262 on Build GRCh38
rs776148345 Chr.22: 35262861-35262861 on Build GRCh38
rs758172551 Chr.22: 35263057-35263057 on Build GRCh38
rs909630056 Chr.22: 35263255-35263255 on Build GRCh38

View Full Product Details