Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36930346-36930848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2RB
Entrez Gene ID
1439
Gene Name
colony stimulating factor 2 receptor beta common subunit
Gene Aliases
CD131, CDw131, IL3RB, IL5RB, SMDP5
UniGene
Hs.592192
Gene Chromosome Location
Chr.22: 36913574-36940449 on Build GRCh38
Transcripts
NM_000395.2 XM_005261340.3 XM_011529903.2 XM_011529904.2 XM_011529905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6526520 c.870C>A p.S290S Chr.22: 36930688-36930688 on Build GRCh38
478930 c.965C>G p.S322C Chr.22: 36930783-36930783 on Build GRCh38
1033900 c.916T>C p.C306R Chr.22: 36930734-36930734 on Build GRCh38
6539640 c.810C>G p.S270R Chr.22: 36930466-36930466 on Build GRCh38
1033901 c.987G>T p.E329D Chr.22: 36930805-36930805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772126395 Chr.22: 36930446-36930446 on Build GRCh38
rs16845 Chr.22: 36930401-36930401 on Build GRCh38
rs1021865587 Chr.22: 36930746-36930746 on Build GRCh38
rs116739204 Chr.22: 36930636-36930636 on Build GRCh38
rs767755998 Chr.22: 36930370-36930370 on Build GRCh38
rs765176076 Chr.22: 36930397-36930397 on Build GRCh38
rs781530019 Chr.22: 36930715-36930715 on Build GRCh38
rs201954835 Chr.22: 36930674-36930674 on Build GRCh38
rs576486802 Chr.22: 36930710-36930710 on Build GRCh38
rs376146978 Chr.22: 36930519-36930519 on Build GRCh38
rs756696682 Chr.22: 36930492-36930492 on Build GRCh38
rs760667424 Chr.22: 36930460-36930460 on Build GRCh38
rs752076635 Chr.22: 36930795-36930795 on Build GRCh38
rs777828160 Chr.22: 36930525-36930525 on Build GRCh38
rs777489716 Chr.22: 36930419-36930419 on Build GRCh38
rs938725526 Chr.22: 36930542-36930542 on Build GRCh38
rs201262017 Chr.22: 36930643-36930643 on Build GRCh38
rs571343472 Chr.22: 36930703-36930703 on Build GRCh38
rs141959318 Chr.22: 36930439-36930439 on Build GRCh38
rs759668933 Chr.22: 36930695-36930695 on Build GRCh38
rs747307683 Chr.22: 36930526-36930526 on Build GRCh38
rs113128697 Chr.22: 36930425-36930425 on Build GRCh38
rs928518258 Chr.22: 36930756-36930756 on Build GRCh38
rs760148608 Chr.22: 36930699-36930699 on Build GRCh38
rs755409736 Chr.22: 36930513-36930513 on Build GRCh38
rs532772042 Chr.22: 36930421-36930421 on Build GRCh38
rs748517710 Chr.22: 36930444-36930444 on Build GRCh38
rs370426702 Chr.22: 36930658-36930658 on Build GRCh38
rs780151719 Chr.22: 36930823-36930823 on Build GRCh38
rs768505622 Chr.22: 36930671-36930671 on Build GRCh38
rs757623965 Chr.22: 36930713-36930713 on Build GRCh38
rs761525371 Chr.22: 36930685-36930685 on Build GRCh38
rs530551352 Chr.22: 36930571-36930571 on Build GRCh38
rs965299657 Chr.22: 36930408-36930408 on Build GRCh38
rs551025109 Chr.22: 36930422-36930422 on Build GRCh38
rs776815049 Chr.22: 36930530-36930530 on Build GRCh38
rs368230425 Chr.22: 36930779-36930779 on Build GRCh38
rs1050599256 Chr.22: 36930557-36930557 on Build GRCh38
rs759697603 Chr.22: 36930532-36930532 on Build GRCh38
rs375783135 Chr.22: 36930758-36930758 on Build GRCh38
rs779207096 Chr.22: 36930442-36930442 on Build GRCh38
rs143834108 Chr.22: 36930496-36930496 on Build GRCh38
rs772326447 Chr.22: 36930519-36930519 on Build GRCh38
rs751261357 Chr.22: 36930488-36930488 on Build GRCh38
rs547780142 Chr.22: 36930609-36930609 on Build GRCh38
rs771081852 Chr.22: 36930527-36930527 on Build GRCh38
rs771086036 Chr.22: 36930466-36930466 on Build GRCh38
rs749198467 Chr.22: 36930664-36930664 on Build GRCh38
rs773542147 Chr.22: 36930457-36930457 on Build GRCh38
rs757436084 Chr.22: 36930654-36930654 on Build GRCh38
rs555140256 Chr.22: 36930766-36930766 on Build GRCh38
rs775542545 Chr.22: 36930662-36930662 on Build GRCh38
rs984721671 Chr.22: 36930750-36930750 on Build GRCh38
rs765923544 Chr.22: 36930705-36930705 on Build GRCh38
rs1011820968 Chr.22: 36930588-36930588 on Build GRCh38
rs774127718 Chr.22: 36930677-36930677 on Build GRCh38
rs767143075 Chr.22: 36930687-36930687 on Build GRCh38
rs756336062 Chr.22: 36930820-36930820 on Build GRCh38
rs776142666 Chr.22: 36930771-36930771 on Build GRCh38
rs150251048 Chr.22: 36930759-36930759 on Build GRCh38
rs144034620 Chr.22: 36930627-36930627 on Build GRCh38
rs189973117 Chr.22: 36930587-36930587 on Build GRCh38
rs748497720 Chr.22: 36930523-36930523 on Build GRCh38
rs754790619 Chr.22: 36930395-36930395 on Build GRCh38
rs766214768 Chr.22: 36930381-36930381 on Build GRCh38
rs376898001 Chr.22: 36930561-36930561 on Build GRCh38
rs758984857 Chr.22: 36930778-36930778 on Build GRCh38
rs770131622 Chr.22: 36930424-36930424 on Build GRCh38
rs760820946 Chr.22: 36930380-36930380 on Build GRCh38
rs151112655 Chr.22: 36930707-36930707 on Build GRCh38
rs745934088 Chr.22: 36930660-36930660 on Build GRCh38
rs373556286 Chr.22: 36930751-36930751 on Build GRCh38
rs749754665 Chr.22: 36930482-36930482 on Build GRCh38
rs368637457 Chr.22: 36930385-36930385 on Build GRCh38
rs762858972 Chr.22: 36930478-36930478 on Build GRCh38
rs372288492 Chr.22: 36930799-36930799 on Build GRCh38
rs752478464 Chr.22: 36930473-36930473 on Build GRCh38
rs773097805 Chr.22: 36930376-36930376 on Build GRCh38
rs761485907 Chr.22: 36930613-36930613 on Build GRCh38
rs767940994 Chr.22: 36930812-36930812 on Build GRCh38
rs778151079 Chr.22: 36930638-36930638 on Build GRCh38
rs373990658 Chr.22: 36930550-36930550 on Build GRCh38
rs117805308 Chr.22: 36930752-36930752 on Build GRCh38
rs762241530 Chr.22: 36930800-36930800 on Build GRCh38
rs672601313 Chr.22: 36930468-36930468 on Build GRCh38
rs746137268 Chr.22: 36930721-36930721 on Build GRCh38
rs539051663 Chr.22: 36930661-36930661 on Build GRCh38
rs764516861 Chr.22: 36930709-36930709 on Build GRCh38
rs267606239 Chr.22: 36930672-36930672 on Build GRCh38
rs749965216 Chr.22: 36930509-36930509 on Build GRCh38
rs749331765 Chr.22: 36930745-36930745 on Build GRCh38
rs767079000 Chr.22: 36930614-36930614 on Build GRCh38
rs986241313 Chr.22: 36930804-36930804 on Build GRCh38
rs138918081 Chr.22: 36930760-36930760 on Build GRCh38
rs1038250280 Chr.22: 36930612-36930612 on Build GRCh38
rs1006764309 Chr.22: 36930391-36930391 on Build GRCh38
rs750845271 Chr.22: 36930813-36930813 on Build GRCh38
rs756424237 Chr.22: 36930727-36930727 on Build GRCh38
rs765298790 Chr.22: 36930469-36930469 on Build GRCh38
rs986540208 Chr.22: 36930432-36930432 on Build GRCh38
rs747969899 Chr.22: 36930757-36930757 on Build GRCh38
rs775632483 Chr.22: 36930558-36930558 on Build GRCh38
rs371290321 Chr.22: 36930484-36930484 on Build GRCh38
rs1030675779 Chr.22: 36930692-36930692 on Build GRCh38
rs375358994 Chr.22: 36930628-36930628 on Build GRCh38
rs201384090 Chr.22: 36930415-36930415 on Build GRCh38

View Full Product Details