Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38084762-38085258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2L2
Entrez Gene ID
80115
Gene Name
BAI1 associated protein 2 like 2
Gene Aliases
-
UniGene
Hs.474822
Gene Chromosome Location
Chr.22: 38084889-38110944 on Build GRCh38
Transcripts
NM_025045.5 XM_005261751.4 XM_011530379.2 XM_011530380.2 XM_011530381.2 XM_011530382.2 XM_011530383.2 XM_011530384.2 XM_011530386.2 XM_011530387.2 XM_011530388.2

SNP Detail

SNP ID Chromosome Location
rs888631937 Chr.22: 38085071-38085071 on Build GRCh38
rs954589266 Chr.22: 38085152-38085152 on Build GRCh38
rs910356747 Chr.22: 38085160-38085160 on Build GRCh38
rs528125183 Chr.22: 38084953-38084953 on Build GRCh38
rs569024255 Chr.22: 38084997-38084997 on Build GRCh38
rs917197109 Chr.22: 38084910-38084913 on Build GRCh38
rs539522624 Chr.22: 38085214-38085222 on Build GRCh38
rs534279456 Chr.22: 38085107-38085107 on Build GRCh38
rs987536761 Chr.22: 38084807-38084807 on Build GRCh38
rs961055239 Chr.22: 38085018-38085018 on Build GRCh38
rs1034281376 Chr.22: 38085187-38085187 on Build GRCh38
rs920507988 Chr.22: 38085037-38085038 on Build GRCh38
rs530338067 Chr.22: 38084805-38084805 on Build GRCh38
rs755950108 Chr.22: 38085134-38085134 on Build GRCh38
rs968550480 Chr.22: 38085162-38085162 on Build GRCh38
rs1044562090 Chr.22: 38085051-38085051 on Build GRCh38
rs550485609 Chr.22: 38084984-38084984 on Build GRCh38
rs999087514 Chr.22: 38084830-38084830 on Build GRCh38
rs960874781 Chr.22: 38085195-38085195 on Build GRCh38
rs546233096 Chr.22: 38084967-38084967 on Build GRCh38
rs916919236 Chr.22: 38085236-38085236 on Build GRCh38
rs564612550 Chr.22: 38085232-38085232 on Build GRCh38
rs1057374806 Chr.22: 38085022-38085022 on Build GRCh38
rs34319574 Chr.22: 38085233-38085233 on Build GRCh38
rs766756118 Chr.22: 38085072-38085072 on Build GRCh38
rs567002876 Chr.22: 38085098-38085098 on Build GRCh38
rs182118899 Chr.22: 38085159-38085159 on Build GRCh38
rs1036381743 Chr.22: 38084831-38084831 on Build GRCh38
rs1022589154 Chr.22: 38085172-38085172 on Build GRCh38
rs1015569023 Chr.22: 38084913-38084913 on Build GRCh38
rs371494239 Chr.22: 38084892-38084892 on Build GRCh38
rs539922417 Chr.22: 38085203-38085203 on Build GRCh38
rs774281353 Chr.22: 38085120-38085120 on Build GRCh38
rs985892922 Chr.22: 38084991-38084991 on Build GRCh38
rs763235019 Chr.22: 38085034-38085034 on Build GRCh38
rs397689926 Chr.22: 38085011-38085012 on Build GRCh38
rs910387977 Chr.22: 38084994-38084994 on Build GRCh38
rs952736860 Chr.22: 38084899-38084899 on Build GRCh38
rs947146308 Chr.22: 38084782-38084782 on Build GRCh38
rs1042689239 Chr.22: 38084815-38084815 on Build GRCh38
rs561045868 Chr.22: 38084893-38084893 on Build GRCh38
rs902834761 Chr.22: 38085007-38085007 on Build GRCh38
rs3216435 Chr.22: 38085006-38085007 on Build GRCh38
rs968275950 Chr.22: 38085182-38085183 on Build GRCh38
rs961707336 Chr.22: 38084940-38084940 on Build GRCh38
rs1036207555 Chr.22: 38084915-38084918 on Build GRCh38
rs899009665 Chr.22: 38084919-38084919 on Build GRCh38
rs747428154 Chr.22: 38085021-38085022 on Build GRCh38
rs532303676 Chr.22: 38084983-38084983 on Build GRCh38
rs556910394 Chr.22: 38085164-38085164 on Build GRCh38
rs559027683 Chr.22: 38085121-38085121 on Build GRCh38
rs570898292 Chr.22: 38084973-38084973 on Build GRCh38
rs994587968 Chr.22: 38084922-38084922 on Build GRCh38
rs753749016 Chr.22: 38085209-38085209 on Build GRCh38
rs138639815 Chr.22: 38085183-38085183 on Build GRCh38
rs377728531 Chr.22: 38084929-38084930 on Build GRCh38
rs915426376 Chr.22: 38084883-38084883 on Build GRCh38
rs902865196 Chr.22: 38084828-38084828 on Build GRCh38
rs956926232 Chr.22: 38084847-38084848 on Build GRCh38
rs113285054 Chr.22: 38085146-38085146 on Build GRCh38
rs542022197 Chr.22: 38084833-38084833 on Build GRCh38
rs578229803 Chr.22: 38085112-38085112 on Build GRCh38
rs548033400 Chr.22: 38085038-38085038 on Build GRCh38
rs11912970 Chr.22: 38085201-38085201 on Build GRCh38
rs939401471 Chr.22: 38084900-38084901 on Build GRCh38
rs1016118516 Chr.22: 38085088-38085088 on Build GRCh38
rs748672035 Chr.22: 38084966-38084966 on Build GRCh38
rs961396999 Chr.22: 38085230-38085230 on Build GRCh38
rs1013129257 Chr.22: 38084852-38084852 on Build GRCh38
rs947113653 Chr.22: 38084995-38084995 on Build GRCh38
rs896102529 Chr.22: 38085026-38085026 on Build GRCh38
rs988370882 Chr.22: 38084856-38084856 on Build GRCh38
rs146229052 Chr.22: 38085020-38085020 on Build GRCh38
rs984632958 Chr.22: 38084905-38084905 on Build GRCh38
rs572931361 Chr.22: 38085202-38085202 on Build GRCh38
rs1050670384 Chr.22: 38085118-38085118 on Build GRCh38
rs542410666 Chr.22: 38085189-38085189 on Build GRCh38
rs200918160 Chr.22: 38085010-38085011 on Build GRCh38
rs971989291 Chr.22: 38084950-38084950 on Build GRCh38

View Full Product Details