Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38097965-38098455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2L2
Entrez Gene ID
80115
Gene Name
BAI1 associated protein 2 like 2
Gene Aliases
-
UniGene
Hs.474822
Gene Chromosome Location
Chr.22: 38084889-38110944 on Build GRCh38
Transcripts
NM_025045.5 XM_005261751.4 XM_011530379.2 XM_011530380.2 XM_011530381.2 XM_011530382.2 XM_011530383.2 XM_011530384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444985 c.331G>A p.D111N Chr.22: 38098428-38098428 on Build GRCh38
6890942 c.403A>C p.K135Q Chr.22: 38098125-38098125 on Build GRCh38
7224110 c.465+4C>A p.? Chr.22: 38098059-38098059 on Build GRCh38
4104056 c.390G>T p.A130A Chr.22: 38098138-38098138 on Build GRCh38
1034098 c.372C>T p.L124L Chr.22: 38098156-38098156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769078557 Chr.22: 38098389-38098389 on Build GRCh38
rs545639069 Chr.22: 38098223-38098223 on Build GRCh38
rs757451621 Chr.22: 38098134-38098134 on Build GRCh38
rs146914325 Chr.22: 38098239-38098239 on Build GRCh38
rs2413500 Chr.22: 38098010-38098010 on Build GRCh38
rs765749399 Chr.22: 38098079-38098081 on Build GRCh38
rs781383109 Chr.22: 38098040-38098040 on Build GRCh38
rs749494215 Chr.22: 38098131-38098131 on Build GRCh38
rs774617806 Chr.22: 38098062-38098062 on Build GRCh38
rs371204360 Chr.22: 38098172-38098172 on Build GRCh38
rs764791297 Chr.22: 38098093-38098093 on Build GRCh38
rs746344585 Chr.22: 38098073-38098073 on Build GRCh38
rs761314064 Chr.22: 38098092-38098092 on Build GRCh38
rs1034824482 Chr.22: 38098373-38098373 on Build GRCh38
rs1001280964 Chr.22: 38098216-38098216 on Build GRCh38
rs925576972 Chr.22: 38098004-38098034 on Build GRCh38
rs373761427 Chr.22: 38098045-38098045 on Build GRCh38
rs975681188 Chr.22: 38098183-38098183 on Build GRCh38
rs899864303 Chr.22: 38098211-38098211 on Build GRCh38
rs147676763 Chr.22: 38097999-38098011 on Build GRCh38
rs769275796 Chr.22: 38098173-38098173 on Build GRCh38
rs1044990737 Chr.22: 38098279-38098279 on Build GRCh38
rs201911939 Chr.22: 38098025-38098025 on Build GRCh38
rs774140016 Chr.22: 38098188-38098188 on Build GRCh38
rs958366955 Chr.22: 38098363-38098363 on Build GRCh38
rs1010040957 Chr.22: 38098252-38098252 on Build GRCh38
rs780442026 Chr.22: 38098365-38098365 on Build GRCh38
rs374968109 Chr.22: 38098196-38098196 on Build GRCh38
rs998830508 Chr.22: 38098000-38098000 on Build GRCh38
rs2413498 Chr.22: 38098007-38098007 on Build GRCh38
rs771304812 Chr.22: 38098052-38098052 on Build GRCh38
rs746229242 Chr.22: 38098161-38098161 on Build GRCh38
rs181557999 Chr.22: 38098033-38098033 on Build GRCh38
rs778147192 Chr.22: 38098043-38098043 on Build GRCh38
rs766169557 Chr.22: 38098095-38098095 on Build GRCh38
rs371450984 Chr.22: 38098041-38098041 on Build GRCh38
rs772640472 Chr.22: 38098177-38098177 on Build GRCh38
rs2413496 Chr.22: 38098005-38098005 on Build GRCh38
rs2413497 Chr.22: 38098006-38098006 on Build GRCh38
rs963293226 Chr.22: 38098182-38098182 on Build GRCh38
rs772498507 Chr.22: 38098163-38098163 on Build GRCh38
rs530602449 Chr.22: 38098103-38098103 on Build GRCh38
rs759319686 Chr.22: 38098406-38098406 on Build GRCh38
rs981198399 Chr.22: 38098337-38098337 on Build GRCh38
rs746698397 Chr.22: 38097998-38097999 on Build GRCh38
rs138193993 Chr.22: 38098340-38098343 on Build GRCh38
rs779004883 Chr.22: 38098226-38098226 on Build GRCh38
rs770085261 Chr.22: 38098042-38098042 on Build GRCh38
rs761403395 Chr.22: 38098012-38098012 on Build GRCh38
rs753624045 Chr.22: 38098115-38098115 on Build GRCh38
rs751345794 Chr.22: 38098096-38098096 on Build GRCh38
rs926955302 Chr.22: 38098338-38098338 on Build GRCh38
rs773872676 Chr.22: 38098403-38098403 on Build GRCh38
rs148375299 Chr.22: 38098253-38098253 on Build GRCh38
rs367764330 Chr.22: 38098094-38098094 on Build GRCh38
rs745998475 Chr.22: 38098100-38098100 on Build GRCh38
rs548050897 Chr.22: 38098228-38098228 on Build GRCh38
rs937589890 Chr.22: 38098056-38098056 on Build GRCh38
rs1032889149 Chr.22: 38098237-38098237 on Build GRCh38
rs1054866521 Chr.22: 38098271-38098271 on Build GRCh38
rs776060117 Chr.22: 38098089-38098089 on Build GRCh38
rs760640401 Chr.22: 38098416-38098416 on Build GRCh38
rs2413501 Chr.22: 38098011-38098011 on Build GRCh38
rs376440553 Chr.22: 38098401-38098401 on Build GRCh38
rs893721236 Chr.22: 38098019-38098019 on Build GRCh38
rs752758932 Chr.22: 38098114-38098114 on Build GRCh38
rs758744849 Chr.22: 38098364-38098364 on Build GRCh38
rs369986403 Chr.22: 38098402-38098402 on Build GRCh38
rs975816624 Chr.22: 38098127-38098128 on Build GRCh38
rs1003323958 Chr.22: 38098058-38098058 on Build GRCh38
rs775455048 Chr.22: 38098414-38098414 on Build GRCh38
rs755008387 Chr.22: 38098037-38098037 on Build GRCh38
rs200330597 Chr.22: 38098155-38098155 on Build GRCh38
rs764193292 Chr.22: 38098201-38098201 on Build GRCh38
rs367875355 Chr.22: 38098198-38098198 on Build GRCh38
rs374671605 Chr.22: 38098185-38098185 on Build GRCh38
rs771971719 Chr.22: 38098412-38098412 on Build GRCh38
rs902250365 Chr.22: 38097987-38097987 on Build GRCh38
rs934727841 Chr.22: 38098255-38098255 on Build GRCh38
rs930122249 Chr.22: 38098171-38098171 on Build GRCh38
rs775968335 Chr.22: 38098166-38098166 on Build GRCh38
rs762630015 Chr.22: 38098180-38098180 on Build GRCh38
rs572221144 Chr.22: 38098195-38098195 on Build GRCh38
rs777292221 Chr.22: 38098392-38098392 on Build GRCh38
rs370486366 Chr.22: 38098274-38098274 on Build GRCh38
rs753885320 Chr.22: 38098222-38098222 on Build GRCh38
rs748763917 Chr.22: 38098395-38098395 on Build GRCh38
rs946659754 Chr.22: 38098278-38098278 on Build GRCh38
rs372491905 Chr.22: 38098004-38098016 on Build GRCh38
rs1003397840 Chr.22: 38098355-38098355 on Build GRCh38
rs2413499 Chr.22: 38098008-38098008 on Build GRCh38
rs772588519 Chr.22: 38098074-38098074 on Build GRCh38
rs553752701 Chr.22: 38098121-38098121 on Build GRCh38
rs375167081 Chr.22: 38098028-38098028 on Build GRCh38
rs764879371 Chr.22: 38098020-38098020 on Build GRCh38

View Full Product Details