Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39025339-39025834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3D
Entrez Gene ID
140564
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3D
Gene Aliases
A3D, APOBEC3DE, APOBEC3E, ARP6
UniGene
Hs.629775
Gene Chromosome Location
Chr.22: 39021113-39034328 on Build GRCh38
Transcripts
NM_152426.3 XM_005261337.3 XM_017028596.1 XM_017028643.1

Gene Symbol
APOBEC3F
Entrez Gene ID
200316
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3F
Gene Aliases
A3F, ARP8, BK150C2.4.MRNA, KA6
UniGene
Hs.659809
Gene Chromosome Location
Chr.22: 39020454-39055972 on Build GRCh38
Transcripts
NM_152426.3 XM_005261337.3 XM_017028596.1 XM_017028643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5484509 c.677C>A p.A226D Chr.22: 39025536-39025536 on Build GRCh38
6414621 c.505T>G p.W169G Chr.22: 39025571-39025571 on Build GRCh38
3363648 c.486T>C p.Y162Y Chr.22: 39025345-39025345 on Build GRCh38
259468 c.765C>T p.F255F Chr.22: 39025624-39025624 on Build GRCh38
6162252 c.577C>A p.L193M Chr.22: 39025643-39025643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373246643 Chr.22: 39025697-39025697 on Build GRCh38
rs114229069 Chr.22: 39025504-39025504 on Build GRCh38
rs776170055 Chr.22: 39025708-39025708 on Build GRCh38
rs11364441 Chr.22: 39025774-39025774 on Build GRCh38
rs752289272 Chr.22: 39025454-39025454 on Build GRCh38
rs1028632627 Chr.22: 39025763-39025763 on Build GRCh38
rs769445080 Chr.22: 39025711-39025711 on Build GRCh38
rs754429373 Chr.22: 39025448-39025448 on Build GRCh38
rs529621406 Chr.22: 39025705-39025705 on Build GRCh38
rs754916485 Chr.22: 39025675-39025675 on Build GRCh38
rs369340896 Chr.22: 39025695-39025695 on Build GRCh38
rs775676692 Chr.22: 39025488-39025488 on Build GRCh38
rs181901737 Chr.22: 39025696-39025696 on Build GRCh38
rs372459013 Chr.22: 39025677-39025677 on Build GRCh38
rs201658890 Chr.22: 39025607-39025607 on Build GRCh38
rs558890879 Chr.22: 39025728-39025728 on Build GRCh38
rs773322348 Chr.22: 39025545-39025547 on Build GRCh38
rs759622157 Chr.22: 39025391-39025391 on Build GRCh38
rs748431888 Chr.22: 39025545-39025545 on Build GRCh38
rs760685405 Chr.22: 39025663-39025664 on Build GRCh38
rs768598048 Chr.22: 39025519-39025520 on Build GRCh38
rs115210060 Chr.22: 39025507-39025507 on Build GRCh38
rs752681692 Chr.22: 39025667-39025667 on Build GRCh38
rs201835248 Chr.22: 39025378-39025378 on Build GRCh38
rs959721511 Chr.22: 39025458-39025458 on Build GRCh38
rs769992817 Chr.22: 39025396-39025396 on Build GRCh38
rs190831871 Chr.22: 39025602-39025602 on Build GRCh38
rs777417359 Chr.22: 39025526-39025526 on Build GRCh38
rs749341019 Chr.22: 39025519-39025519 on Build GRCh38
rs754274350 Chr.22: 39025435-39025435 on Build GRCh38
rs762174544 Chr.22: 39025508-39025508 on Build GRCh38
rs776857077 Chr.22: 39025544-39025544 on Build GRCh38
rs146744584 Chr.22: 39025512-39025512 on Build GRCh38
rs370274357 Chr.22: 39025532-39025532 on Build GRCh38
rs770951387 Chr.22: 39025390-39025390 on Build GRCh38
rs764940909 Chr.22: 39025654-39025654 on Build GRCh38
rs5757426 Chr.22: 39025625-39025625 on Build GRCh38
rs542231983 Chr.22: 39025762-39025762 on Build GRCh38
rs772290561 Chr.22: 39025394-39025394 on Build GRCh38
rs907553690 Chr.22: 39025491-39025491 on Build GRCh38
rs778917803 Chr.22: 39025529-39025529 on Build GRCh38
rs201373761 Chr.22: 39025587-39025587 on Build GRCh38
rs186672417 Chr.22: 39025559-39025559 on Build GRCh38
rs768625030 Chr.22: 39025543-39025543 on Build GRCh38
rs563709240 Chr.22: 39025801-39025801 on Build GRCh38
rs386821331 Chr.22: 39025745-39025746 on Build GRCh38
rs375390945 Chr.22: 39025534-39025534 on Build GRCh38
rs186308428 Chr.22: 39025745-39025745 on Build GRCh38
rs777408528 Chr.22: 39025462-39025462 on Build GRCh38
rs1049952294 Chr.22: 39025515-39025515 on Build GRCh38
rs747068569 Chr.22: 39025541-39025541 on Build GRCh38
rs763570476 Chr.22: 39025586-39025586 on Build GRCh38
rs750919260 Chr.22: 39025514-39025514 on Build GRCh38
rs12159311 Chr.22: 39025734-39025734 on Build GRCh38
rs963420093 Chr.22: 39025500-39025500 on Build GRCh38
rs550306184 Chr.22: 39025418-39025418 on Build GRCh38
rs745764932 Chr.22: 39025533-39025533 on Build GRCh38
rs965615634 Chr.22: 39025791-39025791 on Build GRCh38
rs371292817 Chr.22: 39025451-39025451 on Build GRCh38
rs374339162 Chr.22: 39025474-39025474 on Build GRCh38
rs189444557 Chr.22: 39025380-39025380 on Build GRCh38
rs200273887 Chr.22: 39025590-39025590 on Build GRCh38
rs757104391 Chr.22: 39025528-39025528 on Build GRCh38
rs201653331 Chr.22: 39025641-39025641 on Build GRCh38
rs367844222 Chr.22: 39025527-39025527 on Build GRCh38
rs770906662 Chr.22: 39025710-39025712 on Build GRCh38
rs189860500 Chr.22: 39025746-39025746 on Build GRCh38
rs772270455 Chr.22: 39025487-39025487 on Build GRCh38
rs372956016 Chr.22: 39025387-39025387 on Build GRCh38
rs764332727 Chr.22: 39025432-39025432 on Build GRCh38
rs368179608 Chr.22: 39025632-39025632 on Build GRCh38
rs953092167 Chr.22: 39025765-39025765 on Build GRCh38
rs749069463 Chr.22: 39025470-39025470 on Build GRCh38
rs111333866 Chr.22: 39025743-39025743 on Build GRCh38
rs143474193 Chr.22: 39025422-39025422 on Build GRCh38
rs139309648 Chr.22: 39025581-39025581 on Build GRCh38
rs760027760 Chr.22: 39025651-39025651 on Build GRCh38
rs746324437 Chr.22: 39025398-39025398 on Build GRCh38
rs566220681 Chr.22: 39025714-39025714 on Build GRCh38
rs755076267 Chr.22: 39025624-39025624 on Build GRCh38
rs376752851 Chr.22: 39025701-39025701 on Build GRCh38
rs368930676 Chr.22: 39025381-39025381 on Build GRCh38
rs200163974 Chr.22: 39025521-39025521 on Build GRCh38
rs766985357 Chr.22: 39025520-39025520 on Build GRCh38
rs368121641 Chr.22: 39025558-39025558 on Build GRCh38
rs142602921 Chr.22: 39025710-39025710 on Build GRCh38
rs772405688 Chr.22: 39025702-39025702 on Build GRCh38
rs947633275 Chr.22: 39025804-39025804 on Build GRCh38
rs756219183 Chr.22: 39025689-39025689 on Build GRCh38
rs770853742 Chr.22: 39025471-39025471 on Build GRCh38
rs528328922 Chr.22: 39025608-39025608 on Build GRCh38
rs1020190561 Chr.22: 39025773-39025773 on Build GRCh38
rs376455314 Chr.22: 39025443-39025443 on Build GRCh38
rs748178761 Chr.22: 39025684-39025684 on Build GRCh38
rs746506547 Chr.22: 39025649-39025649 on Build GRCh38
rs763627354 Chr.22: 39025652-39025652 on Build GRCh38
rs184060031 Chr.22: 39025377-39025377 on Build GRCh38
rs770142981 Chr.22: 39025557-39025557 on Build GRCh38
rs1001399987 Chr.22: 39025593-39025593 on Build GRCh38
rs374999207 Chr.22: 39025769-39025769 on Build GRCh38
rs898812252 Chr.22: 39025759-39025759 on Build GRCh38
rs778934213 Chr.22: 39025481-39025481 on Build GRCh38
rs140300593 Chr.22: 39025795-39025795 on Build GRCh38
rs758212292 Chr.22: 39025668-39025668 on Build GRCh38
rs375774801 Chr.22: 39025688-39025688 on Build GRCh38
rs191086739 Chr.22: 39025362-39025362 on Build GRCh38
rs201911698 Chr.22: 39025535-39025535 on Build GRCh38
rs374640232 Chr.22: 39025726-39025726 on Build GRCh38
rs201456810 Chr.22: 39025617-39025617 on Build GRCh38
rs377283514 Chr.22: 39025485-39025485 on Build GRCh38
rs765098359 Chr.22: 39025595-39025595 on Build GRCh38
rs760172009 Chr.22: 39025585-39025585 on Build GRCh38
rs766147548 Chr.22: 39025669-39025669 on Build GRCh38
rs1014970697 Chr.22: 39025620-39025620 on Build GRCh38
rs1014867116 Chr.22: 39025755-39025755 on Build GRCh38
rs755602956 Chr.22: 39025523-39025523 on Build GRCh38
rs369041729 Chr.22: 39025674-39025674 on Build GRCh38
rs574930814 Chr.22: 39025761-39025761 on Build GRCh38
rs773446740 Chr.22: 39025640-39025640 on Build GRCh38
rs375389371 Chr.22: 39025650-39025650 on Build GRCh38
rs780030323 Chr.22: 39025447-39025447 on Build GRCh38
rs937427421 Chr.22: 39025735-39025735 on Build GRCh38
rs776326105 Chr.22: 39025653-39025653 on Build GRCh38
rs777912378 Chr.22: 39025690-39025690 on Build GRCh38
rs760781145 Chr.22: 39025489-39025489 on Build GRCh38
rs763519936 Chr.22: 39025516-39025516 on Build GRCh38
rs766861928 Chr.22: 39025561-39025561 on Build GRCh38
rs1057330058 Chr.22: 39025737-39025737 on Build GRCh38
rs765696442 Chr.22: 39025439-39025439 on Build GRCh38
rs755830934 Chr.22: 39025461-39025461 on Build GRCh38

View Full Product Details