Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41607757-41608269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DESI1
Entrez Gene ID
27351
Gene Name
desumoylating isopeptidase 1
Gene Aliases
D15Wsu75e, DESI2, DJ347H13.4, DeSI-1, FAM152B, PPPDE2
UniGene
Hs.570455
Gene Chromosome Location
Chr.22: 41598028-41621077 on Build GRCh38
Transcripts
NM_015704.2 XM_005261571.4

SNP Detail

SNP ID Chromosome Location
rs1018913386 Chr.22: 41608162-41608162 on Build GRCh38
rs754329956 Chr.22: 41607965-41607965 on Build GRCh38
rs1011087713 Chr.22: 41608173-41608173 on Build GRCh38
rs537155501 Chr.22: 41607982-41607982 on Build GRCh38
rs976850413 Chr.22: 41608187-41608187 on Build GRCh38
rs945805066 Chr.22: 41607906-41607906 on Build GRCh38
rs578249741 Chr.22: 41608151-41608151 on Build GRCh38
rs183060986 Chr.22: 41607816-41607816 on Build GRCh38
rs772416975 Chr.22: 41607814-41607814 on Build GRCh38
rs757708714 Chr.22: 41607990-41607990 on Build GRCh38
rs770575676 Chr.22: 41607890-41607890 on Build GRCh38
rs757397375 Chr.22: 41607791-41607791 on Build GRCh38
rs1014767336 Chr.22: 41607993-41607993 on Build GRCh38
rs1056142490 Chr.22: 41607970-41607971 on Build GRCh38
rs764171116 Chr.22: 41607911-41607911 on Build GRCh38
rs748319687 Chr.22: 41607905-41607906 on Build GRCh38
rs750823288 Chr.22: 41608089-41608089 on Build GRCh38
rs1026982200 Chr.22: 41608100-41608100 on Build GRCh38
rs976900532 Chr.22: 41607872-41607872 on Build GRCh38
rs556733401 Chr.22: 41607827-41607827 on Build GRCh38
rs985716314 Chr.22: 41608139-41608139 on Build GRCh38
rs1022525948 Chr.22: 41608189-41608189 on Build GRCh38
rs998274757 Chr.22: 41608110-41608110 on Build GRCh38
rs555794416 Chr.22: 41608008-41608008 on Build GRCh38
rs779380772 Chr.22: 41607808-41607808 on Build GRCh38
rs767293350 Chr.22: 41607903-41607903 on Build GRCh38
rs904221738 Chr.22: 41607955-41607955 on Build GRCh38
rs765708532 Chr.22: 41608042-41608042 on Build GRCh38
rs547373625 Chr.22: 41608081-41608081 on Build GRCh38
rs553433020 Chr.22: 41608145-41608145 on Build GRCh38
rs1049880583 Chr.22: 41608171-41608171 on Build GRCh38
rs1037018528 Chr.22: 41608057-41608057 on Build GRCh38
rs200027450 Chr.22: 41607888-41607888 on Build GRCh38
rs191530366 Chr.22: 41608086-41608086 on Build GRCh38
rs774808128 Chr.22: 41607870-41607872 on Build GRCh38
rs759575200 Chr.22: 41607901-41607901 on Build GRCh38
rs190470555 Chr.22: 41607787-41607787 on Build GRCh38
rs186935063 Chr.22: 41607905-41607905 on Build GRCh38
rs967006114 Chr.22: 41608212-41608212 on Build GRCh38
rs769297078 Chr.22: 41607830-41607830 on Build GRCh38
rs939985631 Chr.22: 41608051-41608051 on Build GRCh38
rs772981321 Chr.22: 41607831-41607831 on Build GRCh38
rs761520301 Chr.22: 41608221-41608221 on Build GRCh38
rs112538887 Chr.22: 41608172-41608172 on Build GRCh38
rs367865815 Chr.22: 41607893-41607893 on Build GRCh38
rs1042670837 Chr.22: 41607927-41607927 on Build GRCh38
rs1048924359 Chr.22: 41607978-41607978 on Build GRCh38
rs573970482 Chr.22: 41608074-41608074 on Build GRCh38
rs746390290 Chr.22: 41607811-41607811 on Build GRCh38
rs372766561 Chr.22: 41607869-41607869 on Build GRCh38

View Full Product Details