Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45546953-45547383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
UniGene
Hs.24601
Gene Chromosome Location
Chr.22: 45502839-45601134 on Build GRCh38
Transcripts
NM_001996.3 NM_006485.3 NM_006486.2 NM_006487.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274974 c.1322-9G>T p.? Chr.22: 45547076-45547076 on Build GRCh38
4806931 c.1440A>C p.E480D Chr.22: 45547203-45547203 on Build GRCh38
5461240 c.1417G>A p.D473N Chr.22: 45547180-45547180 on Build GRCh38
4887864 c.1369G>A p.V457I Chr.22: 45547132-45547132 on Build GRCh38
6446836 c.1386G>C p.Q462H Chr.22: 45547149-45547149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756810396 Chr.22: 45547240-45547240 on Build GRCh38
rs770754324 Chr.22: 45547106-45547106 on Build GRCh38
rs764808881 Chr.22: 45547239-45547239 on Build GRCh38
rs571138392 Chr.22: 45547208-45547208 on Build GRCh38
rs765389132 Chr.22: 45547102-45547102 on Build GRCh38
rs774139312 Chr.22: 45547163-45547163 on Build GRCh38
rs767833538 Chr.22: 45547069-45547069 on Build GRCh38
rs975993931 Chr.22: 45547334-45547334 on Build GRCh38
rs111619546 Chr.22: 45547205-45547205 on Build GRCh38
rs142391879 Chr.22: 45547274-45547274 on Build GRCh38
rs113098769 Chr.22: 45547082-45547082 on Build GRCh38
rs748749795 Chr.22: 45547159-45547159 on Build GRCh38
rs574640165 Chr.22: 45547269-45547269 on Build GRCh38
rs752326620 Chr.22: 45547244-45547244 on Build GRCh38
rs1024708622 Chr.22: 45547014-45547014 on Build GRCh38
rs753878099 Chr.22: 45547090-45547090 on Build GRCh38
rs531215150 Chr.22: 45547335-45547335 on Build GRCh38
rs35445684 Chr.22: 45547134-45547134 on Build GRCh38
rs776048831 Chr.22: 45547070-45547070 on Build GRCh38
rs986047984 Chr.22: 45547068-45547068 on Build GRCh38
rs866515808 Chr.22: 45547024-45547024 on Build GRCh38
rs201120975 Chr.22: 45547209-45547209 on Build GRCh38
rs748669790 Chr.22: 45547194-45547194 on Build GRCh38
rs764463728 Chr.22: 45547184-45547184 on Build GRCh38
rs9626169 Chr.22: 45547314-45547314 on Build GRCh38
rs1050533573 Chr.22: 45547013-45547013 on Build GRCh38
rs778613166 Chr.22: 45547241-45547241 on Build GRCh38
rs760586019 Chr.22: 45547096-45547097 on Build GRCh38
rs1054154314 Chr.22: 45546987-45546987 on Build GRCh38
rs116684373 Chr.22: 45547235-45547235 on Build GRCh38
rs376076899 Chr.22: 45547254-45547254 on Build GRCh38
rs145367437 Chr.22: 45547055-45547055 on Build GRCh38
rs774159876 Chr.22: 45547061-45547061 on Build GRCh38
rs772457843 Chr.22: 45547179-45547179 on Build GRCh38
rs201622466 Chr.22: 45547045-45547045 on Build GRCh38
rs374070196 Chr.22: 45547054-45547054 on Build GRCh38
rs978142318 Chr.22: 45547283-45547283 on Build GRCh38
rs1000794640 Chr.22: 45546981-45546981 on Build GRCh38
rs764205817 Chr.22: 45547089-45547089 on Build GRCh38
rs958900683 Chr.22: 45547095-45547095 on Build GRCh38
rs752383299 Chr.22: 45547223-45547223 on Build GRCh38
rs777139607 Chr.22: 45547195-45547195 on Build GRCh38
rs576733050 Chr.22: 45546995-45546997 on Build GRCh38
rs978026091 Chr.22: 45547092-45547092 on Build GRCh38
rs947974782 Chr.22: 45547349-45547349 on Build GRCh38
rs534127321 Chr.22: 45547212-45547212 on Build GRCh38
rs377280819 Chr.22: 45547216-45547216 on Build GRCh38
rs115375707 Chr.22: 45547071-45547071 on Build GRCh38
rs139939070 Chr.22: 45547131-45547131 on Build GRCh38
rs748103377 Chr.22: 45547041-45547041 on Build GRCh38
rs369758080 Chr.22: 45547051-45547051 on Build GRCh38
rs529863673 Chr.22: 45547168-45547168 on Build GRCh38
rs776440284 Chr.22: 45547213-45547214 on Build GRCh38
rs372113593 Chr.22: 45547137-45547137 on Build GRCh38
rs374018571 Chr.22: 45547229-45547229 on Build GRCh38
rs1027212854 Chr.22: 45547239-45547239 on Build GRCh38
rs370504340 Chr.22: 45547242-45547242 on Build GRCh38
rs374918773 Chr.22: 45547138-45547138 on Build GRCh38
rs142798380 Chr.22: 45547213-45547213 on Build GRCh38
rs942106591 Chr.22: 45547356-45547356 on Build GRCh38
rs759910515 Chr.22: 45547067-45547067 on Build GRCh38
rs367929911 Chr.22: 45547132-45547132 on Build GRCh38
rs185931186 Chr.22: 45547207-45547207 on Build GRCh38
rs924058406 Chr.22: 45547304-45547304 on Build GRCh38
rs775754823 Chr.22: 45547180-45547180 on Build GRCh38
rs747283699 Chr.22: 45547245-45547245 on Build GRCh38
rs554914133 Chr.22: 45547268-45547268 on Build GRCh38
rs894973422 Chr.22: 45546991-45546991 on Build GRCh38
rs781118191 Chr.22: 45547039-45547039 on Build GRCh38
rs375196311 Chr.22: 45547100-45547100 on Build GRCh38
rs970485679 Chr.22: 45547017-45547017 on Build GRCh38
rs761814877 Chr.22: 45547200-45547200 on Build GRCh38
rs967843988 Chr.22: 45547274-45547275 on Build GRCh38
rs538180785 Chr.22: 45547261-45547261 on Build GRCh38
rs985972627 Chr.22: 45547347-45547347 on Build GRCh38
rs761136393 Chr.22: 45547183-45547183 on Build GRCh38
rs770569480 Chr.22: 45547160-45547160 on Build GRCh38
rs739270 Chr.22: 45547323-45547323 on Build GRCh38
rs777119256 Chr.22: 45547256-45547256 on Build GRCh38
rs916336229 Chr.22: 45547119-45547119 on Build GRCh38
rs755950048 Chr.22: 45547226-45547226 on Build GRCh38
rs989905704 Chr.22: 45547328-45547328 on Build GRCh38
rs200010110 Chr.22: 45547044-45547044 on Build GRCh38
rs1046384609 Chr.22: 45547355-45547355 on Build GRCh38
rs189561938 Chr.22: 45547227-45547227 on Build GRCh38
rs1012523095 Chr.22: 45547001-45547001 on Build GRCh38
rs777786177 Chr.22: 45547140-45547140 on Build GRCh38
rs759324313 Chr.22: 45547228-45547228 on Build GRCh38
rs898817787 Chr.22: 45547228-45547228 on Build GRCh38
rs5765464 Chr.22: 45547012-45547012 on Build GRCh38
rs974343262 Chr.22: 45547052-45547052 on Build GRCh38
rs956606137 Chr.22: 45547324-45547324 on Build GRCh38
rs979615596 Chr.22: 45547301-45547301 on Build GRCh38
rs931766796 Chr.22: 45547341-45547341 on Build GRCh38
rs750649269 Chr.22: 45547130-45547130 on Build GRCh38

View Full Product Details