Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15644683-15645177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTD
Entrez Gene ID
686
Gene Name
biotinidase
Gene Aliases
-
UniGene
Hs.444197
Gene Chromosome Location
Chr.3: 15601352-15653709 on Build GRCh38
Transcripts
XM_006713314.3 XM_011534041.2 XM_017007088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1318710 c.1063G>A p.D355N Chr.3: 15644919-15644919 on Build GRCh38
4114805 c.1168G>T p.A390S Chr.3: 15645024-15645024 on Build GRCh38
6096722 c.1294A>G p.T432A Chr.3: 15645150-15645150 on Build GRCh38
4114804 c.1090A>T p.I364F Chr.3: 15644946-15644946 on Build GRCh38
6033410 c.1101C>T p.G367G Chr.3: 15644957-15644957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397514434 Chr.3: 15645014-15645014 on Build GRCh38
rs756863026 Chr.3: 15645017-15645017 on Build GRCh38
rs375239909 Chr.3: 15645028-15645028 on Build GRCh38
rs766139591 Chr.3: 15644881-15644881 on Build GRCh38
rs35034250 Chr.3: 15645027-15645027 on Build GRCh38
rs397514413 Chr.3: 15645124-15645124 on Build GRCh38
rs201476469 Chr.3: 15645115-15645115 on Build GRCh38
rs753466926 Chr.3: 15644913-15644913 on Build GRCh38
rs760612966 Chr.3: 15644982-15644982 on Build GRCh38
rs1012667588 Chr.3: 15644959-15644959 on Build GRCh38
rs891263072 Chr.3: 15644984-15644984 on Build GRCh38
rs886058116 Chr.3: 15645030-15645030 on Build GRCh38
rs397514394 Chr.3: 15644788-15644788 on Build GRCh38
rs748172820 Chr.3: 15645069-15645069 on Build GRCh38
rs912713223 Chr.3: 15645084-15645084 on Build GRCh38
rs746332288 Chr.3: 15644822-15644822 on Build GRCh38
rs672601248 Chr.3: 15645083-15645097 on Build GRCh38
rs887049660 Chr.3: 15645110-15645110 on Build GRCh38
rs764811197 Chr.3: 15645106-15645106 on Build GRCh38
rs397514407 Chr.3: 15645095-15645095 on Build GRCh38
rs397514412 Chr.3: 15645123-15645123 on Build GRCh38
rs397514404 Chr.3: 15645095-15645106 on Build GRCh38
rs397514334 Chr.3: 15644721-15644721 on Build GRCh38
rs980570750 Chr.3: 15644821-15644821 on Build GRCh38
rs397514406 Chr.3: 15645070-15645070 on Build GRCh38
rs397514401 Chr.3: 15645013-15645013 on Build GRCh38
rs532041656 Chr.3: 15644769-15644769 on Build GRCh38
rs397514335 Chr.3: 15645127-15645127 on Build GRCh38
rs931268644 Chr.3: 15645097-15645097 on Build GRCh38
rs780655352 Chr.3: 15644932-15644932 on Build GRCh38
rs886058115 Chr.3: 15644959-15644959 on Build GRCh38
rs140890299 Chr.3: 15644963-15644963 on Build GRCh38
rs587783006 Chr.3: 15644754-15644754 on Build GRCh38
rs750006399 Chr.3: 15645107-15645107 on Build GRCh38
rs397514392 Chr.3: 15644752-15644752 on Build GRCh38
rs764376897 Chr.3: 15644720-15644720 on Build GRCh38
rs1057517256 Chr.3: 15645027-15645028 on Build GRCh38
rs939163397 Chr.3: 15645099-15645099 on Build GRCh38
rs145379955 Chr.3: 15644741-15644741 on Build GRCh38
rs754158925 Chr.3: 15644724-15644724 on Build GRCh38
rs201023772 Chr.3: 15645061-15645061 on Build GRCh38
rs1057516252 Chr.3: 15644985-15644985 on Build GRCh38
rs747059644 Chr.3: 15644833-15644833 on Build GRCh38
rs397514409 Chr.3: 15645105-15645105 on Build GRCh38
rs104893686 Chr.3: 15645063-15645063 on Build GRCh38
rs915150723 Chr.3: 15644783-15644783 on Build GRCh38
rs748194807 Chr.3: 15644911-15644911 on Build GRCh38
rs747259439 Chr.3: 15644863-15644863 on Build GRCh38
rs377651057 Chr.3: 15644791-15644791 on Build GRCh38
rs1031728921 Chr.3: 15645001-15645001 on Build GRCh38
rs747548016 Chr.3: 15644941-15644941 on Build GRCh38
rs397514396 Chr.3: 15644790-15644790 on Build GRCh38
rs117806438 Chr.3: 15644888-15644888 on Build GRCh38
rs374141881 Chr.3: 15645093-15645093 on Build GRCh38
rs750109648 Chr.3: 15645020-15645020 on Build GRCh38
rs397514408 Chr.3: 15645108-15645108 on Build GRCh38
rs757604137 Chr.3: 15644778-15644778 on Build GRCh38
rs773137513 Chr.3: 15644848-15644857 on Build GRCh38
rs771812661 Chr.3: 15645078-15645078 on Build GRCh38
rs753278397 Chr.3: 15645008-15645008 on Build GRCh38
rs397507176 Chr.3: 15644824-15644824 on Build GRCh38
rs768786956 Chr.3: 15644951-15644951 on Build GRCh38
rs144084212 Chr.3: 15644763-15644763 on Build GRCh38
rs777799585 Chr.3: 15645075-15645075 on Build GRCh38
rs751942955 Chr.3: 15645120-15645120 on Build GRCh38
rs760508628 Chr.3: 15645091-15645091 on Build GRCh38
rs397514414 Chr.3: 15645131-15645131 on Build GRCh38
rs148193489 Chr.3: 15644965-15644965 on Build GRCh38
rs778024234 Chr.3: 15644768-15644768 on Build GRCh38
rs768876766 Chr.3: 15644844-15644844 on Build GRCh38
rs770633850 Chr.3: 15645141-15645141 on Build GRCh38
rs763540316 Chr.3: 15644992-15644992 on Build GRCh38
rs760900130 Chr.3: 15644714-15644714 on Build GRCh38
rs185225945 Chr.3: 15644742-15644742 on Build GRCh38
rs776950685 Chr.3: 15644845-15644845 on Build GRCh38
rs148764524 Chr.3: 15644918-15644918 on Build GRCh38
rs557738433 Chr.3: 15645025-15645025 on Build GRCh38
rs397514435 Chr.3: 15645096-15645107 on Build GRCh38
rs755461739 Chr.3: 15645126-15645126 on Build GRCh38
rs202165098 Chr.3: 15644876-15644876 on Build GRCh38
rs746182429 Chr.3: 15644922-15644922 on Build GRCh38
rs368320941 Chr.3: 15645098-15645098 on Build GRCh38
rs774003531 Chr.3: 15644883-15644883 on Build GRCh38
rs775283569 Chr.3: 15645083-15645083 on Build GRCh38
rs749162799 Chr.3: 15644857-15644858 on Build GRCh38
rs397514397 Chr.3: 15644905-15644905 on Build GRCh38
rs775369290 Chr.3: 15644967-15644967 on Build GRCh38
rs372039874 Chr.3: 15645104-15645104 on Build GRCh38
rs975896326 Chr.3: 15645086-15645086 on Build GRCh38
rs944166413 Chr.3: 15645085-15645085 on Build GRCh38
rs749528636 Chr.3: 15645077-15645077 on Build GRCh38
rs769573111 Chr.3: 15644722-15644723 on Build GRCh38
rs776176463 Chr.3: 15644831-15644831 on Build GRCh38
rs764770052 Chr.3: 15645018-15645018 on Build GRCh38
rs371365798 Chr.3: 15644930-15644930 on Build GRCh38
rs751445487 Chr.3: 15644734-15644734 on Build GRCh38
rs1004027979 Chr.3: 15644951-15644952 on Build GRCh38
rs200327983 Chr.3: 15644902-15644902 on Build GRCh38
rs397514411 Chr.3: 15645120-15645121 on Build GRCh38
rs397514386 Chr.3: 15644789-15644789 on Build GRCh38
rs397514403 Chr.3: 15645047-15645048 on Build GRCh38
rs397514398 Chr.3: 15644908-15644908 on Build GRCh38
rs921284450 Chr.3: 15645092-15645092 on Build GRCh38
rs397514400 Chr.3: 15644962-15644962 on Build GRCh38
rs397514395 Chr.3: 15644789-15644789 on Build GRCh38
rs946578169 Chr.3: 15644806-15644806 on Build GRCh38
rs398123142 Chr.3: 15644760-15644760 on Build GRCh38
rs147188293 Chr.3: 15645060-15645060 on Build GRCh38
rs968754573 Chr.3: 15644995-15644995 on Build GRCh38
rs35976361 Chr.3: 15644736-15644736 on Build GRCh38
rs397514433 Chr.3: 15644857-15644857 on Build GRCh38
rs752682410 Chr.3: 15644890-15644890 on Build GRCh38
rs748963622 Chr.3: 15644774-15644774 on Build GRCh38
rs35145938 Chr.3: 15645140-15645140 on Build GRCh38
rs397514405 Chr.3: 15645067-15645067 on Build GRCh38
rs397514391 Chr.3: 15644743-15644743 on Build GRCh38
rs142421934 Chr.3: 15644957-15644957 on Build GRCh38
rs144414845 Chr.3: 15644884-15644884 on Build GRCh38
rs769903360 Chr.3: 15644958-15644958 on Build GRCh38
rs956882463 Chr.3: 15645074-15645074 on Build GRCh38
rs535651486 Chr.3: 15644917-15644917 on Build GRCh38
rs397514399 Chr.3: 15644952-15644952 on Build GRCh38
rs397514410 Chr.3: 15645109-15645109 on Build GRCh38
rs397514393 Chr.3: 15644785-15644785 on Build GRCh38

View Full Product Details