Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15645456-15645962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTD
Entrez Gene ID
686
Gene Name
biotinidase
Gene Aliases
-
UniGene
Hs.444197
Gene Chromosome Location
Chr.3: 15601352-15653709 on Build GRCh38
Transcripts
XM_006713314.3 XM_011534041.2 XM_017007088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5447543 c.1632G>T p.*544Y Chr.3: 15645488-15645488 on Build GRCh38
2152804 c.1612C>T p.R538C Chr.3: 15645468-15645468 on Build GRCh38
1186208 c.1625G>T p.R542M Chr.3: 15645481-15645481 on Build GRCh38
297740 c.1613G>A p.R538H Chr.3: 15645469-15645469 on Build GRCh38
3005392 c.1614C>T p.R538R Chr.3: 15645470-15645470 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1034718749 Chr.3: 15645856-15645856 on Build GRCh38
rs905884941 Chr.3: 15645601-15645601 on Build GRCh38
rs761431603 Chr.3: 15645541-15645541 on Build GRCh38
rs997397379 Chr.3: 15645725-15645725 on Build GRCh38
rs973865557 Chr.3: 15645739-15645739 on Build GRCh38
rs112280794 Chr.3: 15645668-15645668 on Build GRCh38
rs373183298 Chr.3: 15645836-15645837 on Build GRCh38
rs549506770 Chr.3: 15645782-15645782 on Build GRCh38
rs530872564 Chr.3: 15645647-15645647 on Build GRCh38
rs960653182 Chr.3: 15645682-15645682 on Build GRCh38
rs908548821 Chr.3: 15645799-15645799 on Build GRCh38
rs72312983 Chr.3: 15645854-15645855 on Build GRCh38
rs397514432 Chr.3: 15645483-15645483 on Build GRCh38
rs1001475875 Chr.3: 15645609-15645609 on Build GRCh38
rs992758252 Chr.3: 15645841-15645841 on Build GRCh38
rs985124440 Chr.3: 15645807-15645807 on Build GRCh38
rs187175217 Chr.3: 15645836-15645836 on Build GRCh38
rs78601074 Chr.3: 15645699-15645699 on Build GRCh38
rs200147547 Chr.3: 15645531-15645531 on Build GRCh38
rs565011248 Chr.3: 15645598-15645598 on Build GRCh38
rs944311784 Chr.3: 15645914-15645914 on Build GRCh38
rs529324919 Chr.3: 15645764-15645764 on Build GRCh38
rs960652511 Chr.3: 15645859-15645859 on Build GRCh38
rs891458576 Chr.3: 15645617-15645617 on Build GRCh38
rs763867431 Chr.3: 15645913-15645913 on Build GRCh38
rs533707218 Chr.3: 15645860-15645860 on Build GRCh38
rs146136265 Chr.3: 15645485-15645485 on Build GRCh38
rs374047871 Chr.3: 15645512-15645512 on Build GRCh38
rs189885639 Chr.3: 15645798-15645798 on Build GRCh38
rs1027802587 Chr.3: 15645728-15645728 on Build GRCh38
rs62240764 Chr.3: 15645833-15645833 on Build GRCh38
rs773652007 Chr.3: 15645496-15645496 on Build GRCh38
rs75091068 Chr.3: 15645521-15645521 on Build GRCh38
rs763033233 Chr.3: 15645503-15645503 on Build GRCh38
rs755607927 Chr.3: 15645854-15645854 on Build GRCh38
rs543421200 Chr.3: 15645678-15645678 on Build GRCh38
rs199510824 Chr.3: 15645832-15645832 on Build GRCh38
rs948646179 Chr.3: 15645884-15645884 on Build GRCh38
rs1015433194 Chr.3: 15645698-15645698 on Build GRCh38
rs886058118 Chr.3: 15645545-15645545 on Build GRCh38
rs76633683 Chr.3: 15645522-15645522 on Build GRCh38
rs1050035768 Chr.3: 15645484-15645484 on Build GRCh38
rs1011665542 Chr.3: 15645545-15645545 on Build GRCh38
rs1004621476 Chr.3: 15645854-15645854 on Build GRCh38
rs917147984 Chr.3: 15645843-15645843 on Build GRCh38
rs552027846 Chr.3: 15645845-15645845 on Build GRCh38
rs766374135 Chr.3: 15645511-15645511 on Build GRCh38
rs950471658 Chr.3: 15645837-15645837 on Build GRCh38
rs1009776386 Chr.3: 15645629-15645629 on Build GRCh38
rs1013869612 Chr.3: 15645687-15645687 on Build GRCh38

View Full Product Details