Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40400681-40401086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD3
Entrez Gene ID
956
Gene Name
ectonucleoside triphosphate diphosphohydrolase 3
Gene Aliases
CD39L3, HB6, NTPDase-3
UniGene
Hs.441145
Gene Chromosome Location
Chr.3: 40387156-40428626 on Build GRCh38
Transcripts
NM_001248.3 NM_001291960.1 NM_001291961.1 XM_011534266.2 NR_040100.1

Gene Symbol
ENTPD3-AS1
Entrez Gene ID
285266
Gene Name
ENTPD3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.619663
Gene Chromosome Location
Chr.3: 40390951-40453308 on Build GRCh38
Transcripts
NM_001248.3 NM_001291960.1 NM_001291961.1 XM_011534266.2 NR_040100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934143 c.181C>T p.L61L Chr.3: 40400906-40400906 on Build GRCh38
6654291 c.257T>A p.V86D Chr.3: 40400982-40400982 on Build GRCh38
1753208 c.172G>C p.G58R Chr.3: 40400897-40400897 on Build GRCh38
1044545 c.258C>T p.V86V Chr.3: 40400983-40400983 on Build GRCh38
1642254 c.249C>T p.T83T Chr.3: 40400974-40400974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374284055 Chr.3: 40400824-40400824 on Build GRCh38
rs375029066 Chr.3: 40400968-40400968 on Build GRCh38
rs764033756 Chr.3: 40400921-40400921 on Build GRCh38
rs1005506281 Chr.3: 40400726-40400726 on Build GRCh38
rs746211554 Chr.3: 40400938-40400938 on Build GRCh38
rs1009197999 Chr.3: 40401001-40401001 on Build GRCh38
rs927584244 Chr.3: 40400933-40400933 on Build GRCh38
rs572919132 Chr.3: 40400905-40400905 on Build GRCh38
rs561278527 Chr.3: 40400976-40400976 on Build GRCh38
rs546672562 Chr.3: 40400724-40400724 on Build GRCh38
rs200025674 Chr.3: 40400858-40400858 on Build GRCh38
rs940352803 Chr.3: 40400964-40400964 on Build GRCh38
rs9875245 Chr.3: 40400789-40400789 on Build GRCh38
rs750632406 Chr.3: 40401000-40401000 on Build GRCh38
rs566652238 Chr.3: 40400763-40400763 on Build GRCh38
rs769321937 Chr.3: 40400965-40400965 on Build GRCh38
rs376474213 Chr.3: 40401011-40401011 on Build GRCh38
rs777732815 Chr.3: 40400939-40400939 on Build GRCh38
rs191889161 Chr.3: 40400945-40400945 on Build GRCh38
rs747378414 Chr.3: 40401021-40401021 on Build GRCh38
rs201464840 Chr.3: 40400974-40400974 on Build GRCh38
rs754941697 Chr.3: 40400917-40400917 on Build GRCh38
rs879108898 Chr.3: 40400919-40400919 on Build GRCh38
rs755432694 Chr.3: 40401031-40401031 on Build GRCh38
rs779088777 Chr.3: 40400852-40400852 on Build GRCh38
rs371722176 Chr.3: 40401042-40401042 on Build GRCh38
rs202138813 Chr.3: 40400892-40400892 on Build GRCh38
rs1035076701 Chr.3: 40400804-40400804 on Build GRCh38
rs1008835573 Chr.3: 40400854-40400854 on Build GRCh38
rs898952370 Chr.3: 40400797-40400797 on Build GRCh38
rs766285710 Chr.3: 40400915-40400915 on Build GRCh38
rs1046456479 Chr.3: 40400757-40400757 on Build GRCh38
rs147399746 Chr.3: 40400967-40400967 on Build GRCh38
rs557340787 Chr.3: 40400833-40400833 on Build GRCh38
rs769388780 Chr.3: 40401046-40401046 on Build GRCh38
rs748892001 Chr.3: 40401059-40401059 on Build GRCh38
rs140869368 Chr.3: 40400975-40400975 on Build GRCh38
rs767495462 Chr.3: 40400980-40400980 on Build GRCh38
rs537355480 Chr.3: 40400799-40400799 on Build GRCh38
rs775781869 Chr.3: 40400870-40400871 on Build GRCh38
rs1050663221 Chr.3: 40400999-40400999 on Build GRCh38
rs530209145 Chr.3: 40400986-40400986 on Build GRCh38
rs751541001 Chr.3: 40400916-40400916 on Build GRCh38
rs777299047 Chr.3: 40401047-40401047 on Build GRCh38
rs753848660 Chr.3: 40400985-40400985 on Build GRCh38
rs756159226 Chr.3: 40400951-40400951 on Build GRCh38
rs994627122 Chr.3: 40400813-40400813 on Build GRCh38
rs1028855333 Chr.3: 40400815-40400815 on Build GRCh38
rs916765127 Chr.3: 40400925-40400925 on Build GRCh38
rs774065613 Chr.3: 40401063-40401063 on Build GRCh38
rs779253485 Chr.3: 40400936-40400936 on Build GRCh38
rs371608385 Chr.3: 40400951-40400951 on Build GRCh38
rs200251476 Chr.3: 40400969-40400969 on Build GRCh38
rs948279492 Chr.3: 40400928-40400928 on Build GRCh38
rs769215133 Chr.3: 40400944-40400944 on Build GRCh38
rs772691990 Chr.3: 40400859-40400859 on Build GRCh38
rs140576342 Chr.3: 40400930-40400930 on Build GRCh38
rs751499090 Chr.3: 40400941-40400941 on Build GRCh38
rs546810984 Chr.3: 40401012-40401012 on Build GRCh38
rs770586333 Chr.3: 40401062-40401062 on Build GRCh38
rs374234278 Chr.3: 40400875-40400875 on Build GRCh38
rs149445548 Chr.3: 40400914-40400914 on Build GRCh38
rs989632285 Chr.3: 40400922-40400922 on Build GRCh38
rs948357202 Chr.3: 40400727-40400727 on Build GRCh38
rs369251258 Chr.3: 40400992-40400992 on Build GRCh38
rs753967078 Chr.3: 40400927-40400927 on Build GRCh38
rs903142713 Chr.3: 40401014-40401014 on Build GRCh38
rs772992073 Chr.3: 40400909-40400909 on Build GRCh38
rs747592761 Chr.3: 40401043-40401043 on Build GRCh38
rs758712621 Chr.3: 40401009-40401009 on Build GRCh38
rs761310856 Chr.3: 40400894-40400894 on Build GRCh38
rs1024088992 Chr.3: 40400853-40400853 on Build GRCh38
rs747075884 Chr.3: 40400929-40400930 on Build GRCh38
rs968257067 Chr.3: 40400869-40400869 on Build GRCh38
rs545361567 Chr.3: 40400788-40400788 on Build GRCh38

View Full Product Details