Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48180580-48181093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25A
Entrez Gene ID
993
Gene Name
cell division cycle 25A
Gene Aliases
CDC25A2
UniGene
Hs.437705
Gene Chromosome Location
Chr.3: 48157178-48188658 on Build GRCh38
Transcripts
NM_001789.2 NM_201567.1 XM_006713434.2 XM_006713435.2 XM_011534316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6624150 c.525G>A p.R175R Chr.3: 48180745-48180745 on Build GRCh38
13940 c.450G>A p.K150K Chr.3: 48180820-48180820 on Build GRCh38
94045 c.439G>A p.E147K Chr.3: 48180831-48180831 on Build GRCh38
5594377 c.549+2T>C p.? Chr.3: 48180719-48180719 on Build GRCh38
3594858 c.501G>C p.E167D Chr.3: 48180769-48180769 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1027151795 Chr.3: 48180683-48180683 on Build GRCh38
rs758692144 Chr.3: 48180635-48180635 on Build GRCh38
rs553480894 Chr.3: 48180745-48180745 on Build GRCh38
rs542157524 Chr.3: 48180796-48180796 on Build GRCh38
rs774470623 Chr.3: 48180791-48180791 on Build GRCh38
rs778364050 Chr.3: 48180783-48180783 on Build GRCh38
rs778374927 Chr.3: 48180690-48180690 on Build GRCh38
rs931629325 Chr.3: 48180639-48180639 on Build GRCh38
rs371529299 Chr.3: 48180699-48180699 on Build GRCh38
rs758047372 Chr.3: 48180758-48180758 on Build GRCh38
rs149337385 Chr.3: 48180648-48180648 on Build GRCh38
rs762890600 Chr.3: 48180725-48180725 on Build GRCh38
rs953031305 Chr.3: 48180895-48180895 on Build GRCh38
rs1027226794 Chr.3: 48181006-48181006 on Build GRCh38
rs756655625 Chr.3: 48180752-48180752 on Build GRCh38
rs905879740 Chr.3: 48180772-48180772 on Build GRCh38
rs749272262 Chr.3: 48180780-48180780 on Build GRCh38
rs377264850 Chr.3: 48180712-48180712 on Build GRCh38
rs770572616 Chr.3: 48180797-48180797 on Build GRCh38
rs752459968 Chr.3: 48180734-48180734 on Build GRCh38
rs778482044 Chr.3: 48180753-48180753 on Build GRCh38
rs780485559 Chr.3: 48180642-48180642 on Build GRCh38
rs993813128 Chr.3: 48180988-48180988 on Build GRCh38
rs761798691 Chr.3: 48180826-48180826 on Build GRCh38
rs763881238 Chr.3: 48180810-48180810 on Build GRCh38
rs375690864 Chr.3: 48180836-48180836 on Build GRCh38
rs369740191 Chr.3: 48180855-48180855 on Build GRCh38
rs756815985 Chr.3: 48180688-48180688 on Build GRCh38
rs760406123 Chr.3: 48180801-48180801 on Build GRCh38
rs1045991219 Chr.3: 48180971-48180971 on Build GRCh38
rs1012391502 Chr.3: 48181047-48181047 on Build GRCh38
rs774367168 Chr.3: 48180728-48180728 on Build GRCh38
rs760496535 Chr.3: 48180735-48180735 on Build GRCh38
rs139739590 Chr.3: 48180959-48180959 on Build GRCh38
rs574823236 Chr.3: 48180757-48180757 on Build GRCh38
rs778275944 Chr.3: 48180867-48180867 on Build GRCh38
rs372655408 Chr.3: 48180800-48180800 on Build GRCh38
rs765385956 Chr.3: 48180730-48180730 on Build GRCh38
rs771698687 Chr.3: 48180949-48180949 on Build GRCh38
rs753788423 Chr.3: 48180750-48180750 on Build GRCh38
rs780780665 Chr.3: 48181025-48181026 on Build GRCh38
rs377100437 Chr.3: 48180865-48180865 on Build GRCh38
rs941530325 Chr.3: 48180668-48180668 on Build GRCh38
rs755127153 Chr.3: 48180654-48180654 on Build GRCh38
rs771688958 Chr.3: 48180702-48180702 on Build GRCh38
rs369006632 Chr.3: 48180874-48180874 on Build GRCh38
rs3731507 Chr.3: 48180863-48180863 on Build GRCh38
rs745728538 Chr.3: 48180899-48180899 on Build GRCh38
rs6771386 Chr.3: 48180726-48180726 on Build GRCh38
rs770391848 Chr.3: 48180714-48180714 on Build GRCh38
rs757877366 Chr.3: 48180838-48180838 on Build GRCh38
rs753644381 Chr.3: 48180823-48180823 on Build GRCh38
rs867769160 Chr.3: 48180981-48180981 on Build GRCh38
rs531022248 Chr.3: 48180951-48180951 on Build GRCh38
rs748482211 Chr.3: 48180764-48180764 on Build GRCh38
rs370053913 Chr.3: 48180733-48180733 on Build GRCh38
rs373023348 Chr.3: 48180680-48180680 on Build GRCh38
rs1046395415 Chr.3: 48180732-48180732 on Build GRCh38
rs748759350 Chr.3: 48180670-48180670 on Build GRCh38
rs779324041 Chr.3: 48180889-48180889 on Build GRCh38
rs1007917782 Chr.3: 48180625-48180625 on Build GRCh38
rs984425421 Chr.3: 48180902-48180902 on Build GRCh38
rs773886369 Chr.3: 48180718-48180718 on Build GRCh38
rs771441675 Chr.3: 48180882-48180882 on Build GRCh38
rs766815798 Chr.3: 48180750-48180751 on Build GRCh38
rs6763010 Chr.3: 48180968-48180968 on Build GRCh38
rs771404791 Chr.3: 48180788-48180788 on Build GRCh38
rs966262914 Chr.3: 48180850-48180850 on Build GRCh38
rs763971281 Chr.3: 48180739-48180739 on Build GRCh38
rs144492976 Chr.3: 48181067-48181067 on Build GRCh38
rs202079033 Chr.3: 48180669-48180669 on Build GRCh38
rs779890422 Chr.3: 48180763-48180763 on Build GRCh38
rs888693023 Chr.3: 48181033-48181033 on Build GRCh38
rs908977790 Chr.3: 48180940-48180940 on Build GRCh38

View Full Product Details