Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48450357-48450898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRIP
Entrez Gene ID
84126
Gene Name
ATR interacting protein
Gene Aliases
-
UniGene
Hs.694840
Gene Chromosome Location
Chr.3: 48446710-48465655 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2

SNP Detail

SNP ID Chromosome Location
rs752340543 Chr.3: 48450576-48450576 on Build GRCh38
rs75695438 Chr.3: 48450689-48450689 on Build GRCh38
rs1006729459 Chr.3: 48450823-48450823 on Build GRCh38
rs991789513 Chr.3: 48450561-48450561 on Build GRCh38
rs1026517945 Chr.3: 48450627-48450627 on Build GRCh38
rs578158086 Chr.3: 48450481-48450481 on Build GRCh38
rs766393286 Chr.3: 48450467-48450467 on Build GRCh38
rs893775783 Chr.3: 48450380-48450380 on Build GRCh38
rs192316999 Chr.3: 48450815-48450815 on Build GRCh38
rs768956145 Chr.3: 48450526-48450526 on Build GRCh38
rs1051034982 Chr.3: 48450808-48450808 on Build GRCh38
rs962315824 Chr.3: 48450568-48450568 on Build GRCh38
rs959071855 Chr.3: 48450546-48450546 on Build GRCh38
rs982402033 Chr.3: 48450674-48450674 on Build GRCh38
rs185809694 Chr.3: 48450843-48450843 on Build GRCh38
rs868318764 Chr.3: 48450749-48450749 on Build GRCh38
rs991272728 Chr.3: 48450710-48450710 on Build GRCh38
rs1013223063 Chr.3: 48450875-48450875 on Build GRCh38
rs766997108 Chr.3: 48450534-48450534 on Build GRCh38
rs879082726 Chr.3: 48450590-48450592 on Build GRCh38
rs950998013 Chr.3: 48450662-48450662 on Build GRCh38
rs150345328 Chr.3: 48450740-48450740 on Build GRCh38
rs561466435 Chr.3: 48450743-48450743 on Build GRCh38
rs923836283 Chr.3: 48450789-48450789 on Build GRCh38
rs977927663 Chr.3: 48450585-48450585 on Build GRCh38
rs902464923 Chr.3: 48450393-48450393 on Build GRCh38
rs995745348 Chr.3: 48450491-48450491 on Build GRCh38
rs1042325219 Chr.3: 48450390-48450390 on Build GRCh38
rs538250510 Chr.3: 48450436-48450436 on Build GRCh38
rs752675830 Chr.3: 48450518-48450518 on Build GRCh38
rs763116772 Chr.3: 48450433-48450433 on Build GRCh38
rs762445797 Chr.3: 48450849-48450849 on Build GRCh38
rs962916164 Chr.3: 48450485-48450485 on Build GRCh38
rs534776780 Chr.3: 48450405-48450405 on Build GRCh38
rs1029935080 Chr.3: 48450470-48450470 on Build GRCh38
rs13080046 Chr.3: 48450826-48450826 on Build GRCh38
rs925130701 Chr.3: 48450590-48450596 on Build GRCh38
rs1051549709 Chr.3: 48450734-48450734 on Build GRCh38
rs572580928 Chr.3: 48450707-48450707 on Build GRCh38
rs878961508 Chr.3: 48450590-48450590 on Build GRCh38
rs1033300424 Chr.3: 48450512-48450512 on Build GRCh38
rs545237292 Chr.3: 48450538-48450538 on Build GRCh38
rs946702092 Chr.3: 48450388-48450388 on Build GRCh38
rs759086429 Chr.3: 48450514-48450514 on Build GRCh38
rs753687308 Chr.3: 48450738-48450738 on Build GRCh38
rs866210291 Chr.3: 48450492-48450492 on Build GRCh38
rs1004358677 Chr.3: 48450454-48450454 on Build GRCh38
rs577938560 Chr.3: 48450801-48450801 on Build GRCh38
rs891205529 Chr.3: 48450432-48450432 on Build GRCh38
rs756559976 Chr.3: 48450589-48450590 on Build GRCh38
rs556646454 Chr.3: 48450440-48450440 on Build GRCh38
rs895410929 Chr.3: 48450871-48450871 on Build GRCh38
rs567652194 Chr.3: 48450412-48450412 on Build GRCh38
rs531170020 Chr.3: 48450787-48450787 on Build GRCh38
rs573223048 Chr.3: 48450868-48450868 on Build GRCh38
rs966874811 Chr.3: 48450580-48450580 on Build GRCh38
rs959572610 Chr.3: 48450700-48450700 on Build GRCh38
rs1039002277 Chr.3: 48450679-48450679 on Build GRCh38
rs972415029 Chr.3: 48450593-48450593 on Build GRCh38
rs1016647998 Chr.3: 48450566-48450566 on Build GRCh38
rs766409015 Chr.3: 48450479-48450479 on Build GRCh38
rs1042440178 Chr.3: 48450768-48450768 on Build GRCh38
rs996924474 Chr.3: 48450399-48450399 on Build GRCh38

View Full Product Details