Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49015552-49016060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DALRD3
Entrez Gene ID
55152
Gene Name
DALR anticodon binding domain containing 3
Gene Aliases
-
UniGene
Hs.654815
Gene Chromosome Location
Chr.3: 49015399-49021386 on Build GRCh38
Transcripts
NM_001009996.2 NM_001276405.1 NM_018114.5 XM_005265269.3 XM_006713219.2 XM_011533891.1 XM_011533892.1 XM_011533893.1 XM_017006723.1 NM_001320546.1 NM_001320547.1 NM_018031.4

Gene Symbol
WDR6
Entrez Gene ID
11180
Gene Name
WD repeat domain 6
Gene Aliases
-
UniGene
Hs.654815
Gene Chromosome Location
Chr.3: 49007062-49015953 on Build GRCh38
Transcripts
NM_001009996.2 NM_001276405.1 NM_018114.5 XM_005265269.3 XM_006713219.2 XM_011533891.1 XM_011533892.1 XM_011533893.1 XM_017006723.1 NM_001320546.1 NM_001320547.1 NM_018031.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7171230 c.1060C>T p.L354L Chr.3: 49015659-49015659 on Build GRCh38
7171232 c.1018C>G p.R340G Chr.3: 49015701-49015701 on Build GRCh38
1617742 c.922A>G p.I308V Chr.3: 49015993-49015993 on Build GRCh38
4526095 c.871G>A p.D291N Chr.3: 49016044-49016044 on Build GRCh38
6861647 c.1492A>C p.N498H Chr.3: 49015824-49015824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373593638 Chr.3: 49016004-49016004 on Build GRCh38
rs768216071 Chr.3: 49015816-49015816 on Build GRCh38
rs760755516 Chr.3: 49015867-49015867 on Build GRCh38
rs567655530 Chr.3: 49015837-49015837 on Build GRCh38
rs753246937 Chr.3: 49015666-49015667 on Build GRCh38
rs374525061 Chr.3: 49015627-49015627 on Build GRCh38
rs369996113 Chr.3: 49015983-49015983 on Build GRCh38
rs772724633 Chr.3: 49015936-49015936 on Build GRCh38
rs764281318 Chr.3: 49015942-49015942 on Build GRCh38
rs921368659 Chr.3: 49015779-49015779 on Build GRCh38
rs751618056 Chr.3: 49015628-49015628 on Build GRCh38
rs775720727 Chr.3: 49015866-49015866 on Build GRCh38
rs753954360 Chr.3: 49015774-49015774 on Build GRCh38
rs757396708 Chr.3: 49015885-49015885 on Build GRCh38
rs745853227 Chr.3: 49015793-49015793 on Build GRCh38
rs771608468 Chr.3: 49015706-49015706 on Build GRCh38
rs758346722 Chr.3: 49015902-49015902 on Build GRCh38
rs754784023 Chr.3: 49015964-49015964 on Build GRCh38
rs752817985 Chr.3: 49015766-49015766 on Build GRCh38
rs780782761 Chr.3: 49015898-49015898 on Build GRCh38
rs376211800 Chr.3: 49015879-49015879 on Build GRCh38
rs781393149 Chr.3: 49015631-49015631 on Build GRCh38
rs1002462876 Chr.3: 49015701-49015701 on Build GRCh38
rs199567076 Chr.3: 49015905-49015905 on Build GRCh38
rs754861347 Chr.3: 49015965-49015965 on Build GRCh38
rs369611449 Chr.3: 49015804-49015804 on Build GRCh38
rs532886583 Chr.3: 49015680-49015680 on Build GRCh38
rs370677021 Chr.3: 49015737-49015737 on Build GRCh38
rs1001770694 Chr.3: 49015972-49015972 on Build GRCh38
rs373458866 Chr.3: 49015994-49015994 on Build GRCh38
rs1020896977 Chr.3: 49015961-49015961 on Build GRCh38
rs767760752 Chr.3: 49015765-49015765 on Build GRCh38
rs765387162 Chr.3: 49015949-49015949 on Build GRCh38
rs763374494 Chr.3: 49015623-49015623 on Build GRCh38
rs761752333 Chr.3: 49016032-49016032 on Build GRCh38
rs762100811 Chr.3: 49015944-49015944 on Build GRCh38
rs897536397 Chr.3: 49015634-49015634 on Build GRCh38
rs761345422 Chr.3: 49015821-49015821 on Build GRCh38
rs138712639 Chr.3: 49015987-49015987 on Build GRCh38
rs756405689 Chr.3: 49015769-49015769 on Build GRCh38
rs949067306 Chr.3: 49015898-49015898 on Build GRCh38
rs966690564 Chr.3: 49016011-49016011 on Build GRCh38
rs754560245 Chr.3: 49015909-49015909 on Build GRCh38
rs150379594 Chr.3: 49015700-49015700 on Build GRCh38
rs780547603 Chr.3: 49015912-49015912 on Build GRCh38
rs367970354 Chr.3: 49015776-49015776 on Build GRCh38
rs776558997 Chr.3: 49015943-49015943 on Build GRCh38
rs758988047 Chr.3: 49015589-49015589 on Build GRCh38
rs746812813 Chr.3: 49015941-49015941 on Build GRCh38
rs760100962 Chr.3: 49015762-49015762 on Build GRCh38
rs752155553 Chr.3: 49015751-49015751 on Build GRCh38
rs549158920 Chr.3: 49015794-49015794 on Build GRCh38
rs868347018 Chr.3: 49015810-49015810 on Build GRCh38
rs774957928 Chr.3: 49015719-49015719 on Build GRCh38
rs769784149 Chr.3: 49015931-49015932 on Build GRCh38
rs571238030 Chr.3: 49015918-49015918 on Build GRCh38
rs371125542 Chr.3: 49015617-49015617 on Build GRCh38
rs538215497 Chr.3: 49015926-49015926 on Build GRCh38
rs369575864 Chr.3: 49015578-49015578 on Build GRCh38
rs753020281 Chr.3: 49015647-49015647 on Build GRCh38
rs376382703 Chr.3: 49015608-49015608 on Build GRCh38
rs1032135266 Chr.3: 49015716-49015716 on Build GRCh38
rs746463962 Chr.3: 49015809-49015809 on Build GRCh38
rs988108461 Chr.3: 49015727-49015727 on Build GRCh38
rs910086884 Chr.3: 49015829-49015829 on Build GRCh38
rs776310312 Chr.3: 49015820-49015820 on Build GRCh38
rs747541669 Chr.3: 49015984-49015984 on Build GRCh38
rs374823694 Chr.3: 49015738-49015738 on Build GRCh38
rs772604029 Chr.3: 49015995-49015995 on Build GRCh38
rs150836979 Chr.3: 49015775-49015775 on Build GRCh38
rs769037729 Chr.3: 49016017-49016017 on Build GRCh38
rs538237469 Chr.3: 49015968-49015968 on Build GRCh38
rs758150380 Chr.3: 49015960-49015960 on Build GRCh38
rs768067749 Chr.3: 49015858-49015858 on Build GRCh38
rs777643550 Chr.3: 49015946-49015947 on Build GRCh38
rs904942019 Chr.3: 49015917-49015917 on Build GRCh38
rs149038250 Chr.3: 49015828-49015828 on Build GRCh38
rs775974240 Chr.3: 49016003-49016003 on Build GRCh38
rs958201618 Chr.3: 49015726-49015726 on Build GRCh38
rs756389338 Chr.3: 49015670-49015670 on Build GRCh38
rs746172211 Chr.3: 49015935-49015935 on Build GRCh38
rs764314278 Chr.3: 49015979-49015980 on Build GRCh38
rs760184519 Chr.3: 49015850-49015850 on Build GRCh38
rs7100 Chr.3: 49015786-49015786 on Build GRCh38
rs769323842 Chr.3: 49015921-49015921 on Build GRCh38
rs143367268 Chr.3: 49015629-49015629 on Build GRCh38
rs775082689 Chr.3: 49015602-49015602 on Build GRCh38
rs777514739 Chr.3: 49015993-49015993 on Build GRCh38
rs776098843 Chr.3: 49015940-49015940 on Build GRCh38
rs139247938 Chr.3: 49016031-49016031 on Build GRCh38
rs750153587 Chr.3: 49015889-49015889 on Build GRCh38
rs954179101 Chr.3: 49016008-49016008 on Build GRCh38
rs140081609 Chr.3: 49015671-49015671 on Build GRCh38
rs757036832 Chr.3: 49015785-49015785 on Build GRCh38
rs749040850 Chr.3: 49015678-49015678 on Build GRCh38
rs199790792 Chr.3: 49015685-49015685 on Build GRCh38
rs767177804 Chr.3: 49015593-49015593 on Build GRCh38
rs369378004 Chr.3: 49015888-49015888 on Build GRCh38
rs770034304 Chr.3: 49016035-49016035 on Build GRCh38
rs886376570 Chr.3: 49015919-49015919 on Build GRCh38
rs766831505 Chr.3: 49015740-49015740 on Build GRCh38
rs201468616 Chr.3: 49015603-49015603 on Build GRCh38
rs750241321 Chr.3: 49015955-49015955 on Build GRCh38
rs200525644 Chr.3: 49015875-49015875 on Build GRCh38
rs773466982 Chr.3: 49016037-49016037 on Build GRCh38
rs762619897 Chr.3: 49015722-49015722 on Build GRCh38
rs375809849 Chr.3: 49015962-49015962 on Build GRCh38
rs768288881 Chr.3: 49015729-49015729 on Build GRCh38
rs142301323 Chr.3: 49015758-49015758 on Build GRCh38
rs781052741 Chr.3: 49016015-49016015 on Build GRCh38
rs747982898 Chr.3: 49015899-49015900 on Build GRCh38
rs754412035 Chr.3: 49015880-49015881 on Build GRCh38
rs1032497812 Chr.3: 49015934-49015934 on Build GRCh38

View Full Product Details