Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50353527-50354022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561D2
Entrez Gene ID
11068
Gene Name
cytochrome b561 family member D2
Gene Aliases
101F6, TSP10, XXcos-LUCA11.4
UniGene
Hs.149443
Gene Chromosome Location
Chr.3: 50350695-50368197 on Build GRCh38
Transcripts
NM_001291284.1 NM_007022.4 NR_111912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1485333 c.557C>T p.S186F Chr.3: 50353632-50353632 on Build GRCh38
7181439 c.557C>A p.S186Y Chr.3: 50353632-50353632 on Build GRCh38
4394688 c.572C>T p.A191V Chr.3: 50353647-50353647 on Build GRCh38
7226545 c.511A>G p.S171G Chr.3: 50353586-50353586 on Build GRCh38
1717013 c.606C>G p.T202T Chr.3: 50353681-50353681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373995167 Chr.3: 50353664-50353664 on Build GRCh38
rs1045075836 Chr.3: 50353977-50353977 on Build GRCh38
rs143409947 Chr.3: 50353643-50353643 on Build GRCh38
rs193294004 Chr.3: 50353874-50353874 on Build GRCh38
rs587725716 Chr.3: 50353644-50353644 on Build GRCh38
rs373904011 Chr.3: 50353781-50353781 on Build GRCh38
rs587676191 Chr.3: 50353839-50353839 on Build GRCh38
rs1017093863 Chr.3: 50353951-50353951 on Build GRCh38
rs745756036 Chr.3: 50353608-50353608 on Build GRCh38
rs763197657 Chr.3: 50353583-50353583 on Build GRCh38
rs1001573807 Chr.3: 50353900-50353900 on Build GRCh38
rs779861254 Chr.3: 50353723-50353723 on Build GRCh38
rs145546128 Chr.3: 50353552-50353552 on Build GRCh38
rs768116906 Chr.3: 50353780-50353780 on Build GRCh38
rs771871967 Chr.3: 50353618-50353618 on Build GRCh38
rs767968239 Chr.3: 50353982-50353982 on Build GRCh38
rs749930129 Chr.3: 50353667-50353667 on Build GRCh38
rs34123887 Chr.3: 50353849-50353850 on Build GRCh38
rs983916506 Chr.3: 50353762-50353762 on Build GRCh38
rs138212854 Chr.3: 50353626-50353626 on Build GRCh38
rs369073135 Chr.3: 50353793-50353793 on Build GRCh38
rs921184570 Chr.3: 50353749-50353749 on Build GRCh38
rs768061858 Chr.3: 50353658-50353658 on Build GRCh38
rs149406359 Chr.3: 50353718-50353718 on Build GRCh38
rs954021616 Chr.3: 50353755-50353755 on Build GRCh38
rs774767633 Chr.3: 50353732-50353732 on Build GRCh38
rs114924982 Chr.3: 50353736-50353736 on Build GRCh38
rs587611473 Chr.3: 50353810-50353810 on Build GRCh38
rs906063470 Chr.3: 50353638-50353638 on Build GRCh38
rs769201143 Chr.3: 50353752-50353752 on Build GRCh38
rs918823108 Chr.3: 50353991-50353991 on Build GRCh38
rs768263665 Chr.3: 50353641-50353641 on Build GRCh38
rs772183823 Chr.3: 50353630-50353631 on Build GRCh38
rs1036771931 Chr.3: 50353834-50353834 on Build GRCh38
rs772774269 Chr.3: 50353765-50353765 on Build GRCh38
rs749307449 Chr.3: 50353571-50353571 on Build GRCh38
rs141223231 Chr.3: 50353650-50353650 on Build GRCh38
rs760075681 Chr.3: 50353654-50353654 on Build GRCh38
rs1000120147 Chr.3: 50353767-50353767 on Build GRCh38
rs896085332 Chr.3: 50353609-50353609 on Build GRCh38
rs111559356 Chr.3: 50353652-50353652 on Build GRCh38
rs1010211171 Chr.3: 50353695-50353695 on Build GRCh38
rs1014951076 Chr.3: 50353611-50353611 on Build GRCh38
rs182965524 Chr.3: 50353586-50353586 on Build GRCh38
rs757293150 Chr.3: 50353672-50353672 on Build GRCh38
rs776177904 Chr.3: 50353731-50353731 on Build GRCh38
rs201850283 Chr.3: 50353624-50353624 on Build GRCh38
rs962818115 Chr.3: 50353961-50353961 on Build GRCh38
rs201691431 Chr.3: 50353661-50353661 on Build GRCh38
rs774189500 Chr.3: 50353587-50353587 on Build GRCh38
rs1000320674 Chr.3: 50353983-50353983 on Build GRCh38
rs768390021 Chr.3: 50353729-50353729 on Build GRCh38
rs778997198 Chr.3: 50353687-50353687 on Build GRCh38
rs587630193 Chr.3: 50353975-50353975 on Build GRCh38
rs35935807 Chr.3: 50353756-50353756 on Build GRCh38
rs147173262 Chr.3: 50353697-50353697 on Build GRCh38
rs970138039 Chr.3: 50353877-50353877 on Build GRCh38
rs760161984 Chr.3: 50353770-50353770 on Build GRCh38
rs962974045 Chr.3: 50353594-50353594 on Build GRCh38
rs375562029 Chr.3: 50353724-50353724 on Build GRCh38

View Full Product Details