Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 70954622-70955119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP1
Entrez Gene ID
27086
Gene Name
forkhead box P1
Gene Aliases
12CC4, HSPC215, MFH, QRF1, hFKH1B
UniGene
Hs.59368
Gene Chromosome Location
Chr.3: 70954714-71583989 on Build GRCh38
Transcripts
NM_001244808.1 NM_001244810.1 NM_001244812.1 NM_001244813.1 NM_001244814.1 NM_001244815.1 NM_001244816.1 NM_032682.5

SNP Detail

SNP ID Chromosome Location
rs189112133 Chr.3: 70954732-70954732 on Build GRCh38
rs1018407190 Chr.3: 70954860-70954860 on Build GRCh38
rs150318535 Chr.3: 70954706-70954706 on Build GRCh38
rs201218600 Chr.3: 70954874-70954878 on Build GRCh38
rs373934071 Chr.3: 70954762-70954762 on Build GRCh38
rs949376715 Chr.3: 70954993-70954993 on Build GRCh38
rs896244619 Chr.3: 70954839-70954839 on Build GRCh38
rs905252896 Chr.3: 70955011-70955011 on Build GRCh38
rs1019981905 Chr.3: 70954771-70954771 on Build GRCh38
rs376510763 Chr.3: 70954655-70954655 on Build GRCh38
rs550705154 Chr.3: 70955019-70955019 on Build GRCh38
rs777251817 Chr.3: 70954949-70954949 on Build GRCh38
rs748084009 Chr.3: 70954817-70954817 on Build GRCh38
rs920076759 Chr.3: 70954914-70954914 on Build GRCh38
rs966776858 Chr.3: 70954650-70954650 on Build GRCh38
rs950766538 Chr.3: 70955065-70955065 on Build GRCh38
rs919293157 Chr.3: 70954725-70954725 on Build GRCh38
rs57959417 Chr.3: 70954898-70954899 on Build GRCh38
rs936719364 Chr.3: 70955071-70955071 on Build GRCh38
rs568087080 Chr.3: 70954822-70954822 on Build GRCh38
rs370863323 Chr.3: 70954873-70954874 on Build GRCh38
rs564352000 Chr.3: 70954693-70954694 on Build GRCh38
rs1045514422 Chr.3: 70954721-70954721 on Build GRCh38
rs533313728 Chr.3: 70954812-70954812 on Build GRCh38
rs193042825 Chr.3: 70954836-70954836 on Build GRCh38
rs754750250 Chr.3: 70954710-70954710 on Build GRCh38
rs1033664565 Chr.3: 70954662-70954662 on Build GRCh38
rs748736809 Chr.3: 70954973-70954973 on Build GRCh38
rs917965969 Chr.3: 70954977-70954977 on Build GRCh38
rs974190063 Chr.3: 70954904-70954904 on Build GRCh38
rs184043647 Chr.3: 70954700-70954700 on Build GRCh38
rs1013329420 Chr.3: 70954895-70954898 on Build GRCh38
rs1001338007 Chr.3: 70954740-70954740 on Build GRCh38
rs549792004 Chr.3: 70954819-70954819 on Build GRCh38
rs763925659 Chr.3: 70954660-70954661 on Build GRCh38
rs369025750 Chr.3: 70954667-70954667 on Build GRCh38
rs1017746934 Chr.3: 70954975-70954975 on Build GRCh38
rs983433825 Chr.3: 70954950-70954950 on Build GRCh38
rs910652418 Chr.3: 70954703-70954703 on Build GRCh38
rs1045166638 Chr.3: 70955005-70955005 on Build GRCh38
rs1032945277 Chr.3: 70954741-70954741 on Build GRCh38
rs550105200 Chr.3: 70954711-70954711 on Build GRCh38
rs964194934 Chr.3: 70954883-70954883 on Build GRCh38
rs893081679 Chr.3: 70954755-70954755 on Build GRCh38
rs528075960 Chr.3: 70954720-70954720 on Build GRCh38
rs557812253 Chr.3: 70954791-70954792 on Build GRCh38
rs902204059 Chr.3: 70954920-70954920 on Build GRCh38
rs769785355 Chr.3: 70954917-70954917 on Build GRCh38
rs985167118 Chr.3: 70955069-70955069 on Build GRCh38
rs1033283131 Chr.3: 70954940-70954940 on Build GRCh38
rs1020540059 Chr.3: 70954649-70954649 on Build GRCh38
rs1000918077 Chr.3: 70954923-70954923 on Build GRCh38
rs919323714 Chr.3: 70955064-70955069 on Build GRCh38
rs1038597936 Chr.3: 70954654-70954654 on Build GRCh38
rs1054317834 Chr.3: 70955079-70955079 on Build GRCh38
rs1010143415 Chr.3: 70954761-70954761 on Build GRCh38
rs559109218 Chr.3: 70954878-70954879 on Build GRCh38

View Full Product Details