Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 74262493-74263008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN3
Entrez Gene ID
5067
Gene Name
contactin 3
Gene Aliases
BIG-1, PANG, PCS
UniGene
Hs.12723
Gene Chromosome Location
Chr.3: 74262568-74614448 on Build GRCh38
Transcripts
NM_020872.2 XM_005264757.3 XM_011533768.2 XM_017006507.1 XM_017006508.1 XM_017006509.1

SNP Detail

SNP ID Chromosome Location
rs906413868 Chr.3: 74262613-74262613 on Build GRCh38
rs984112214 Chr.3: 74262547-74262547 on Build GRCh38
rs964492433 Chr.3: 74262908-74262908 on Build GRCh38
rs945124131 Chr.3: 74262895-74262895 on Build GRCh38
rs150697852 Chr.3: 74262708-74262708 on Build GRCh38
rs920838463 Chr.3: 74262522-74262522 on Build GRCh38
rs926174448 Chr.3: 74262861-74262861 on Build GRCh38
rs998311127 Chr.3: 74262794-74262794 on Build GRCh38
rs1029818453 Chr.3: 74262818-74262818 on Build GRCh38
rs867173134 Chr.3: 74262747-74262747 on Build GRCh38
rs989515912 Chr.3: 74262886-74262886 on Build GRCh38
rs1053473806 Chr.3: 74262543-74262543 on Build GRCh38
rs1055478447 Chr.3: 74262665-74262665 on Build GRCh38
rs562150481 Chr.3: 74262931-74262931 on Build GRCh38
rs980273066 Chr.3: 74262967-74262967 on Build GRCh38
rs1002106869 Chr.3: 74262620-74262620 on Build GRCh38
rs562470326 Chr.3: 74262610-74262610 on Build GRCh38
rs985660437 Chr.3: 74262849-74262849 on Build GRCh38
rs1015612551 Chr.3: 74262748-74262748 on Build GRCh38
rs116186614 Chr.3: 74262732-74262732 on Build GRCh38
rs146699550 Chr.3: 74262944-74262944 on Build GRCh38
rs192962821 Chr.3: 74262952-74262952 on Build GRCh38
rs72572779 Chr.3: 74262922-74262925 on Build GRCh38
rs1005928951 Chr.3: 74262734-74262734 on Build GRCh38
rs182251992 Chr.3: 74262888-74262888 on Build GRCh38
rs927378023 Chr.3: 74262969-74262969 on Build GRCh38
rs917523473 Chr.3: 74262965-74262965 on Build GRCh38
rs747994320 Chr.3: 74262562-74262562 on Build GRCh38
rs954385950 Chr.3: 74262848-74262848 on Build GRCh38
rs867632354 Chr.3: 74262791-74262791 on Build GRCh38
rs577484224 Chr.3: 74262634-74262634 on Build GRCh38
rs971524446 Chr.3: 74262966-74262966 on Build GRCh38
rs772955758 Chr.3: 74262918-74262918 on Build GRCh38
rs1046205774 Chr.3: 74262557-74262557 on Build GRCh38
rs994909391 Chr.3: 74262910-74262910 on Build GRCh38
rs545890500 Chr.3: 74262686-74262686 on Build GRCh38
rs1017354422 Chr.3: 74262900-74262900 on Build GRCh38
rs1054498674 Chr.3: 74262782-74262782 on Build GRCh38
rs370774689 Chr.3: 74262772-74262772 on Build GRCh38
rs565853707 Chr.3: 74262757-74262757 on Build GRCh38
rs376340669 Chr.3: 74262641-74262641 on Build GRCh38
rs537959125 Chr.3: 74262955-74262955 on Build GRCh38
rs186893792 Chr.3: 74262898-74262898 on Build GRCh38
rs1009450246 Chr.3: 74262831-74262831 on Build GRCh38
rs752052556 Chr.3: 74262959-74262959 on Build GRCh38
rs950237220 Chr.3: 74262635-74262635 on Build GRCh38
rs957646849 Chr.3: 74262883-74262883 on Build GRCh38
rs143254641 Chr.3: 74262546-74262546 on Build GRCh38
rs890272176 Chr.3: 74262897-74262897 on Build GRCh38

View Full Product Details