Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107645684-107646185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
UniGene
Hs.124366
Gene Chromosome Location
Chr.3: 107522936-107811329 on Build GRCh38
Transcripts
NM_001142568.2 NM_001276286.1 NM_020235.6 XM_005247642.3 XM_005247643.3 XM_005247644.3 XM_011512999.2 XM_011513000.1 XM_011513001.1 XM_011513002.2 XM_011513003.2 XM_011513004.2 XM_011513005.2 XM_011513006.2 XM_011513007.2 XM_011513010.2 XM_017006881.1 XM_017006882.1 XM_017006883.1

SNP Detail

SNP ID Chromosome Location
rs887718712 Chr.3: 107646078-107646078 on Build GRCh38
rs192168764 Chr.3: 107646146-107646146 on Build GRCh38
rs1063166 Chr.3: 107645866-107645866 on Build GRCh38
rs1033777244 Chr.3: 107646071-107646071 on Build GRCh38
rs939116484 Chr.3: 107645934-107645937 on Build GRCh38
rs114713747 Chr.3: 107645944-107645944 on Build GRCh38
rs1007543861 Chr.3: 107646133-107646133 on Build GRCh38
rs1018034379 Chr.3: 107646141-107646141 on Build GRCh38
rs1001843368 Chr.3: 107646038-107646038 on Build GRCh38
rs966570066 Chr.3: 107646142-107646142 on Build GRCh38
rs913724623 Chr.3: 107646112-107646112 on Build GRCh38
rs992341718 Chr.3: 107646106-107646106 on Build GRCh38
rs926365000 Chr.3: 107645837-107645837 on Build GRCh38
rs371968086 Chr.3: 107645789-107645789 on Build GRCh38
rs897355652 Chr.3: 107645966-107645966 on Build GRCh38
rs925268318 Chr.3: 107645719-107645719 on Build GRCh38
rs995705969 Chr.3: 107645976-107645976 on Build GRCh38
rs139074419 Chr.3: 107645813-107645813 on Build GRCh38
rs1049006901 Chr.3: 107646069-107646069 on Build GRCh38
rs1045159128 Chr.3: 107645814-107645814 on Build GRCh38
rs375202311 Chr.3: 107645996-107645996 on Build GRCh38
rs778946249 Chr.3: 107645874-107645874 on Build GRCh38
rs1008744943 Chr.3: 107645852-107645852 on Build GRCh38
rs891179331 Chr.3: 107645821-107645821 on Build GRCh38
rs993366947 Chr.3: 107646023-107646023 on Build GRCh38
rs932709827 Chr.3: 107645737-107645737 on Build GRCh38
rs970717763 Chr.3: 107646024-107646024 on Build GRCh38
rs539029151 Chr.3: 107646004-107646004 on Build GRCh38
rs576516305 Chr.3: 107645993-107645993 on Build GRCh38
rs914859173 Chr.3: 107645788-107645788 on Build GRCh38
rs925154002 Chr.3: 107646152-107646152 on Build GRCh38
rs371938991 Chr.3: 107645942-107645942 on Build GRCh38
rs78628756 Chr.3: 107645712-107645712 on Build GRCh38
rs960464482 Chr.3: 107646104-107646104 on Build GRCh38
rs897672389 Chr.3: 107645999-107645999 on Build GRCh38
rs535177704 Chr.3: 107646116-107646116 on Build GRCh38

View Full Product Details