Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107710212-107710735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
UniGene
Hs.124366
Gene Chromosome Location
Chr.3: 107522936-107811329 on Build GRCh38
Transcripts
NM_001142568.2 NM_001276286.1 NM_020235.6 XM_005247642.3 XM_005247643.3 XM_005247644.3 XM_011512999.2 XM_011513000.1 XM_011513001.1 XM_011513002.2 XM_011513003.2 XM_011513004.2 XM_011513005.2 XM_011513006.2 XM_011513007.2 XM_011513010.2 XM_017006881.1 XM_017006882.1 XM_017006883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1647515 c.133G>T p.E45* Chr.3: 107710593-107710593 on Build GRCh38
1246008 c.150G>T p.E50D Chr.3: 107710610-107710610 on Build GRCh38
445259 c.41G>C p.G14A Chr.3: 107710501-107710501 on Build GRCh38
1246007 c.150G>T p.E50D Chr.3: 107710610-107710610 on Build GRCh38
1417560 c.142G>A p.E48K Chr.3: 107710602-107710602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188715043 Chr.3: 107710398-107710398 on Build GRCh38
rs768390204 Chr.3: 107710499-107710501 on Build GRCh38
rs564577147 Chr.3: 107710457-107710457 on Build GRCh38
rs996767449 Chr.3: 107710601-107710601 on Build GRCh38
rs775339035 Chr.3: 107710428-107710429 on Build GRCh38
rs781275532 Chr.3: 107710671-107710671 on Build GRCh38
rs759895170 Chr.3: 107710641-107710641 on Build GRCh38
rs766899282 Chr.3: 107710246-107710246 on Build GRCh38
rs1000814939 Chr.3: 107710524-107710524 on Build GRCh38
rs760926955 Chr.3: 107710602-107710602 on Build GRCh38
rs751612100 Chr.3: 107710447-107710447 on Build GRCh38
rs766765527 Chr.3: 107710492-107710492 on Build GRCh38
rs750781598 Chr.3: 107710403-107710403 on Build GRCh38
rs528150696 Chr.3: 107710485-107710485 on Build GRCh38
rs776457058 Chr.3: 107710566-107710568 on Build GRCh38
rs754358353 Chr.3: 107710420-107710420 on Build GRCh38
rs970511396 Chr.3: 107710317-107710317 on Build GRCh38
rs746723627 Chr.3: 107710452-107710452 on Build GRCh38
rs948566041 Chr.3: 107710362-107710362 on Build GRCh38
rs1015170329 Chr.3: 107710704-107710704 on Build GRCh38
rs191938995 Chr.3: 107710709-107710709 on Build GRCh38
rs371275290 Chr.3: 107710654-107710654 on Build GRCh38
rs760866987 Chr.3: 107710486-107710486 on Build GRCh38
rs140186288 Chr.3: 107710461-107710461 on Build GRCh38
rs201416014 Chr.3: 107710518-107710518 on Build GRCh38
rs745424772 Chr.3: 107710418-107710418 on Build GRCh38
rs760637156 Chr.3: 107710427-107710428 on Build GRCh38
rs769814455 Chr.3: 107710568-107710568 on Build GRCh38
rs778368771 Chr.3: 107710435-107710435 on Build GRCh38
rs61731591 Chr.3: 107710474-107710474 on Build GRCh38
rs201168714 Chr.3: 107710591-107710591 on Build GRCh38
rs562041683 Chr.3: 107710355-107710355 on Build GRCh38
rs35485772 Chr.3: 107710252-107710253 on Build GRCh38
rs755667941 Chr.3: 107710395-107710395 on Build GRCh38
rs907744517 Chr.3: 107710356-107710356 on Build GRCh38
rs938583689 Chr.3: 107710458-107710458 on Build GRCh38
rs754201392 Chr.3: 107710495-107710495 on Build GRCh38
rs908238596 Chr.3: 107710305-107710305 on Build GRCh38
rs773501127 Chr.3: 107710480-107710480 on Build GRCh38
rs890609230 Chr.3: 107710638-107710638 on Build GRCh38
rs755426100 Chr.3: 107710516-107710516 on Build GRCh38
rs745673837 Chr.3: 107710566-107710566 on Build GRCh38
rs763517682 Chr.3: 107710656-107710656 on Build GRCh38
rs941936327 Chr.3: 107710358-107710358 on Build GRCh38
rs143853371 Chr.3: 107710468-107710468 on Build GRCh38
rs961016115 Chr.3: 107710314-107710314 on Build GRCh38
rs372538007 Chr.3: 107710632-107710632 on Build GRCh38
rs777321618 Chr.3: 107710528-107710528 on Build GRCh38
rs775577388 Chr.3: 107710578-107710578 on Build GRCh38
rs369694155 Chr.3: 107710607-107710607 on Build GRCh38
rs766740901 Chr.3: 107710419-107710419 on Build GRCh38
rs781020323 Chr.3: 107710565-107710565 on Build GRCh38
rs777884102 Chr.3: 107710303-107710303 on Build GRCh38
rs146886977 Chr.3: 107710701-107710701 on Build GRCh38
rs756898704 Chr.3: 107710560-107710560 on Build GRCh38
rs756508701 Chr.3: 107710411-107710411 on Build GRCh38
rs758447683 Chr.3: 107710297-107710297 on Build GRCh38
rs748596495 Chr.3: 107710595-107710595 on Build GRCh38
rs773234461 Chr.3: 107710589-107710589 on Build GRCh38
rs771621646 Chr.3: 107710423-107710424 on Build GRCh38
rs770744932 Chr.3: 107710454-107710454 on Build GRCh38
rs765761541 Chr.3: 107710519-107710519 on Build GRCh38
rs1024692818 Chr.3: 107710252-107710252 on Build GRCh38
rs1056060103 Chr.3: 107710544-107710544 on Build GRCh38
rs963247171 Chr.3: 107710708-107710708 on Build GRCh38
rs757917284 Chr.3: 107710527-107710527 on Build GRCh38
rs750097550 Chr.3: 107710272-107710272 on Build GRCh38
rs763627098 Chr.3: 107710668-107710668 on Build GRCh38
rs748183985 Chr.3: 107710580-107710580 on Build GRCh38
rs763133432 Chr.3: 107710587-107710589 on Build GRCh38
rs377696604 Chr.3: 107710643-107710643 on Build GRCh38
rs546841118 Chr.3: 107710470-107710470 on Build GRCh38
rs746667796 Chr.3: 107710541-107710541 on Build GRCh38
rs900556346 Chr.3: 107710600-107710600 on Build GRCh38
rs371658392 Chr.3: 107710456-107710456 on Build GRCh38
rs766579067 Chr.3: 107710619-107710623 on Build GRCh38
rs973859327 Chr.3: 107710329-107710329 on Build GRCh38
rs772268205 Chr.3: 107710581-107710581 on Build GRCh38
rs747675892 Chr.3: 107710444-107710444 on Build GRCh38
rs954961276 Chr.3: 107710282-107710282 on Build GRCh38
rs755475698 Chr.3: 107710421-107710421 on Build GRCh38
rs1043131804 Chr.3: 107710531-107710531 on Build GRCh38
rs919665751 Chr.3: 107710350-107710350 on Build GRCh38

View Full Product Details