Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107732848-107733364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
UniGene
Hs.124366
Gene Chromosome Location
Chr.3: 107522936-107811329 on Build GRCh38
Transcripts
NM_001142568.2 NM_001276286.1 NM_020235.6 XM_005247642.3 XM_005247643.3 XM_005247644.3 XM_011512999.2 XM_011513000.1 XM_011513001.1 XM_011513002.2 XM_011513003.2 XM_011513004.2 XM_011513005.2 XM_011513006.2 XM_011513007.2 XM_011513010.2 XM_017006881.1 XM_017006882.1 XM_017006883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5619755 c.605C>T p.P202L Chr.3: 107732959-107732959 on Build GRCh38
5034431 c.602-7T>C p.? Chr.3: 107732949-107732949 on Build GRCh38
1593160 c.653C>T p.A218V Chr.3: 107733007-107733007 on Build GRCh38
6610237 c.663A>G p.T221T Chr.3: 107733017-107733017 on Build GRCh38
5034430 c.602-7T>C p.? Chr.3: 107732949-107732949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937444456 Chr.3: 107733215-107733215 on Build GRCh38
rs780411771 Chr.3: 107733212-107733212 on Build GRCh38
rs749560484 Chr.3: 107733254-107733254 on Build GRCh38
rs761198102 Chr.3: 107732975-107732975 on Build GRCh38
rs1043482989 Chr.3: 107733180-107733180 on Build GRCh38
rs113174336 Chr.3: 107732951-107732951 on Build GRCh38
rs376024169 Chr.3: 107733020-107733020 on Build GRCh38
rs78981234 Chr.3: 107733121-107733121 on Build GRCh38
rs184108371 Chr.3: 107733093-107733093 on Build GRCh38
rs762007949 Chr.3: 107732944-107732944 on Build GRCh38
rs558986312 Chr.3: 107733311-107733311 on Build GRCh38
rs73850141 Chr.3: 107732910-107732910 on Build GRCh38
rs767894559 Chr.3: 107732946-107732946 on Build GRCh38
rs142423142 Chr.3: 107732923-107732923 on Build GRCh38
rs774614412 Chr.3: 107733069-107733069 on Build GRCh38
rs370772925 Chr.3: 107733044-107733044 on Build GRCh38
rs923205617 Chr.3: 107732888-107732888 on Build GRCh38
rs528309793 Chr.3: 107733042-107733042 on Build GRCh38
rs147992493 Chr.3: 107732996-107732996 on Build GRCh38
rs758032758 Chr.3: 107733051-107733051 on Build GRCh38
rs1057208138 Chr.3: 107733083-107733083 on Build GRCh38
rs866142052 Chr.3: 107733046-107733046 on Build GRCh38
rs751120839 Chr.3: 107733233-107733233 on Build GRCh38
rs1031977466 Chr.3: 107733207-107733207 on Build GRCh38
rs745941544 Chr.3: 107733115-107733115 on Build GRCh38
rs757106654 Chr.3: 107733238-107733238 on Build GRCh38
rs905931562 Chr.3: 107733193-107733193 on Build GRCh38
rs1053041693 Chr.3: 107732904-107732904 on Build GRCh38
rs781291035 Chr.3: 107733061-107733061 on Build GRCh38
rs985195327 Chr.3: 107733301-107733301 on Build GRCh38
rs953685859 Chr.3: 107733216-107733216 on Build GRCh38
rs974590928 Chr.3: 107733091-107733091 on Build GRCh38
rs777573609 Chr.3: 107733053-107733053 on Build GRCh38
rs1036848250 Chr.3: 107732929-107732929 on Build GRCh38
rs764881073 Chr.3: 107732914-107732914 on Build GRCh38
rs768852246 Chr.3: 107733067-107733067 on Build GRCh38
rs199605174 Chr.3: 107733188-107733189 on Build GRCh38
rs551054070 Chr.3: 107733094-107733094 on Build GRCh38
rs770083343 Chr.3: 107733181-107733181 on Build GRCh38
rs777588781 Chr.3: 107733018-107733018 on Build GRCh38
rs4855764 Chr.3: 107733060-107733060 on Build GRCh38
rs947364898 Chr.3: 107733154-107733154 on Build GRCh38
rs879170943 Chr.3: 107733278-107733278 on Build GRCh38
rs62262030 Chr.3: 107732924-107732924 on Build GRCh38
rs370402912 Chr.3: 107732894-107732894 on Build GRCh38
rs778458217 Chr.3: 107733041-107733041 on Build GRCh38
rs539791674 Chr.3: 107733230-107733230 on Build GRCh38
rs77686363 Chr.3: 107733197-107733197 on Build GRCh38
rs143483812 Chr.3: 107733066-107733066 on Build GRCh38
rs563137476 Chr.3: 107732885-107732885 on Build GRCh38
rs746809616 Chr.3: 107733056-107733056 on Build GRCh38
rs1048463544 Chr.3: 107733047-107733047 on Build GRCh38
rs753283314 Chr.3: 107733016-107733016 on Build GRCh38
rs933247592 Chr.3: 107732900-107732900 on Build GRCh38

View Full Product Details