Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107744512-107745062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
UniGene
Hs.124366
Gene Chromosome Location
Chr.3: 107522936-107811329 on Build GRCh38
Transcripts
NM_001142568.2 NM_001276286.1 NM_020235.6 XM_005247642.3 XM_005247643.3 XM_005247644.3 XM_011512999.2 XM_011513000.1 XM_011513001.1 XM_011513002.2 XM_011513003.2 XM_011513004.2 XM_011513005.2 XM_011513006.2 XM_011513007.2 XM_011513010.2 XM_017006881.1 XM_017006882.1 XM_017006883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4112027 c.681C>T p.G227G Chr.3: 107744641-107744641 on Build GRCh38
6478870 c.712G>C p.E238Q Chr.3: 107744672-107744672 on Build GRCh38
2916570 c.748G>T p.E250* Chr.3: 107744708-107744708 on Build GRCh38
7230334 c.746C>A p.A249D Chr.3: 107744706-107744706 on Build GRCh38
6478871 c.712G>C p.E238Q Chr.3: 107744672-107744672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023700526 Chr.3: 107744672-107744672 on Build GRCh38
rs940413725 Chr.3: 107745022-107745022 on Build GRCh38
rs547081918 Chr.3: 107744716-107744716 on Build GRCh38
rs759983490 Chr.3: 107744731-107744731 on Build GRCh38
rs368275155 Chr.3: 107744549-107744549 on Build GRCh38
rs930544057 Chr.3: 107744785-107744785 on Build GRCh38
rs375591352 Chr.3: 107744708-107744708 on Build GRCh38
rs975669661 Chr.3: 107744937-107744937 on Build GRCh38
rs773589007 Chr.3: 107744615-107744615 on Build GRCh38
rs1035100824 Chr.3: 107744834-107744834 on Build GRCh38
rs1052312478 Chr.3: 107744623-107744623 on Build GRCh38
rs777293431 Chr.3: 107744645-107744645 on Build GRCh38
rs769830148 Chr.3: 107744609-107744610 on Build GRCh38
rs556590873 Chr.3: 107744949-107744949 on Build GRCh38
rs762074686 Chr.3: 107744581-107744581 on Build GRCh38
rs745770579 Chr.3: 107744674-107744674 on Build GRCh38
rs772165529 Chr.3: 107744558-107744558 on Build GRCh38
rs777027040 Chr.3: 107744725-107744725 on Build GRCh38
rs1045063390 Chr.3: 107744807-107744807 on Build GRCh38
rs987128157 Chr.3: 107745006-107745006 on Build GRCh38
rs762382089 Chr.3: 107744755-107744755 on Build GRCh38
rs1027950096 Chr.3: 107744801-107744805 on Build GRCh38
rs766817767 Chr.3: 107744630-107744630 on Build GRCh38
rs1003485295 Chr.3: 107744819-107744819 on Build GRCh38
rs760956546 Chr.3: 107744622-107744622 on Build GRCh38
rs1035116219 Chr.3: 107745013-107745013 on Build GRCh38
rs3838316 Chr.3: 107744610-107744610 on Build GRCh38
rs145458765 Chr.3: 107745009-107745009 on Build GRCh38
rs976531705 Chr.3: 107744856-107744856 on Build GRCh38
rs866976865 Chr.3: 107744588-107744588 on Build GRCh38
rs758027302 Chr.3: 107744644-107744644 on Build GRCh38
rs549020538 Chr.3: 107744671-107744671 on Build GRCh38
rs199730916 Chr.3: 107744775-107744776 on Build GRCh38
rs771250632 Chr.3: 107744719-107744719 on Build GRCh38
rs1003560745 Chr.3: 107744564-107744564 on Build GRCh38
rs79692584 Chr.3: 107744610-107744610 on Build GRCh38
rs897995828 Chr.3: 107745014-107745014 on Build GRCh38
rs995285188 Chr.3: 107744545-107744545 on Build GRCh38
rs1003642317 Chr.3: 107744747-107744747 on Build GRCh38
rs754335242 Chr.3: 107744632-107744632 on Build GRCh38
rs910903178 Chr.3: 107744826-107744826 on Build GRCh38
rs34291651 Chr.3: 107744799-107744800 on Build GRCh38
rs189435984 Chr.3: 107744766-107744766 on Build GRCh38
rs199846860 Chr.3: 107744707-107744707 on Build GRCh38
rs35443281 Chr.3: 107744862-107744863 on Build GRCh38
rs368875249 Chr.3: 107744718-107744718 on Build GRCh38
rs905225774 Chr.3: 107744817-107744817 on Build GRCh38
rs118142407 Chr.3: 107744977-107744977 on Build GRCh38
rs559330742 Chr.3: 107744999-107744999 on Build GRCh38
rs372483002 Chr.3: 107744664-107744664 on Build GRCh38
rs765763534 Chr.3: 107744734-107744734 on Build GRCh38
rs1022425575 Chr.3: 107744914-107744914 on Build GRCh38
rs536365277 Chr.3: 107744902-107744902 on Build GRCh38
rs553556530 Chr.3: 107744868-107744868 on Build GRCh38
rs1028563419 Chr.3: 107744966-107744966 on Build GRCh38
rs139927413 Chr.3: 107744898-107744898 on Build GRCh38
rs781201426 Chr.3: 107744673-107744673 on Build GRCh38
rs560335528 Chr.3: 107744642-107744642 on Build GRCh38
rs140692011 Chr.3: 107744679-107744679 on Build GRCh38
rs940555169 Chr.3: 107744728-107744728 on Build GRCh38
rs568783699 Chr.3: 107744678-107744678 on Build GRCh38
rs756714329 Chr.3: 107744741-107744743 on Build GRCh38
rs772208165 Chr.3: 107744613-107744613 on Build GRCh38
rs113140162 Chr.3: 107744774-107744774 on Build GRCh38
rs528981784 Chr.3: 107744667-107744667 on Build GRCh38
rs541698680 Chr.3: 107745029-107745029 on Build GRCh38
rs968715491 Chr.3: 107744923-107744923 on Build GRCh38
rs972315851 Chr.3: 107745024-107745024 on Build GRCh38
rs922394869 Chr.3: 107744862-107744862 on Build GRCh38
rs905209149 Chr.3: 107744978-107744978 on Build GRCh38
rs183760015 Chr.3: 107744635-107744635 on Build GRCh38
rs1036197295 Chr.3: 107744762-107744762 on Build GRCh38
rs1051536545 Chr.3: 107744721-107744721 on Build GRCh38
rs147829800 Chr.3: 107744853-107744853 on Build GRCh38
rs988320996 Chr.3: 107744636-107744636 on Build GRCh38
rs545402484 Chr.3: 107744985-107744985 on Build GRCh38
rs774995663 Chr.3: 107744748-107744748 on Build GRCh38

View Full Product Details