Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120173154-120173650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR156
Entrez Gene ID
165829
Gene Name
G protein-coupled receptor 156
Gene Aliases
GABABL, PGR28
UniGene
Hs.333358
Gene Chromosome Location
Chr.3: 120165481-120285122 on Build GRCh38
Transcripts
NM_001168271.1 NM_153002.2 XM_005247165.2 XM_011512484.2 XM_011512487.1 XM_017005795.1 XM_017005796.1 XM_017005797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297390 c.1023C>A p.T341T Chr.3: 120173381-120173381 on Build GRCh38
3120307 c.1053G>A p.P351P Chr.3: 120173351-120173351 on Build GRCh38
3914644 c.1077G>A p.E359E Chr.3: 120173327-120173327 on Build GRCh38
5729072 c.1023C>T p.T341T Chr.3: 120173381-120173381 on Build GRCh38
4983111 c.958G>C p.E320Q Chr.3: 120173446-120173446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047491270 Chr.3: 120173217-120173217 on Build GRCh38
rs1026402244 Chr.3: 120173484-120173484 on Build GRCh38
rs199542673 Chr.3: 120173408-120173408 on Build GRCh38
rs551086411 Chr.3: 120173486-120173486 on Build GRCh38
rs530427940 Chr.3: 120173351-120173351 on Build GRCh38
rs758922045 Chr.3: 120173581-120173581 on Build GRCh38
rs77362550 Chr.3: 120173582-120173582 on Build GRCh38
rs750799205 Chr.3: 120173515-120173515 on Build GRCh38
rs763229644 Chr.3: 120173261-120173261 on Build GRCh38
rs150638525 Chr.3: 120173368-120173368 on Build GRCh38
rs13064815 Chr.3: 120173274-120173274 on Build GRCh38
rs201936267 Chr.3: 120173298-120173298 on Build GRCh38
rs781543342 Chr.3: 120173272-120173272 on Build GRCh38
rs754391459 Chr.3: 120173390-120173390 on Build GRCh38
rs139943256 Chr.3: 120173385-120173385 on Build GRCh38
rs376630204 Chr.3: 120173268-120173268 on Build GRCh38
rs772180525 Chr.3: 120173406-120173406 on Build GRCh38
rs147227010 Chr.3: 120173249-120173249 on Build GRCh38
rs773160623 Chr.3: 120173337-120173337 on Build GRCh38
rs750801860 Chr.3: 120173371-120173371 on Build GRCh38
rs143351042 Chr.3: 120173396-120173396 on Build GRCh38
rs970494517 Chr.3: 120173397-120173397 on Build GRCh38
rs200413838 Chr.3: 120173473-120173473 on Build GRCh38
rs755745407 Chr.3: 120173284-120173284 on Build GRCh38
rs377694599 Chr.3: 120173346-120173346 on Build GRCh38
rs989448532 Chr.3: 120173616-120173616 on Build GRCh38
rs765875116 Chr.3: 120173398-120173398 on Build GRCh38
rs562535511 Chr.3: 120173352-120173352 on Build GRCh38
rs779594164 Chr.3: 120173287-120173287 on Build GRCh38
rs765413337 Chr.3: 120173348-120173348 on Build GRCh38
rs111236071 Chr.3: 120173395-120173395 on Build GRCh38
rs746846305 Chr.3: 120173414-120173414 on Build GRCh38
rs753190665 Chr.3: 120173455-120173455 on Build GRCh38
rs188976459 Chr.3: 120173258-120173258 on Build GRCh38
rs762999873 Chr.3: 120173357-120173357 on Build GRCh38
rs57059149 Chr.3: 120173425-120173425 on Build GRCh38
rs960154581 Chr.3: 120173361-120173361 on Build GRCh38
rs763239925 Chr.3: 120173435-120173435 on Build GRCh38
rs764170009 Chr.3: 120173363-120173363 on Build GRCh38
rs970556070 Chr.3: 120173236-120173236 on Build GRCh38
rs141895234 Chr.3: 120173365-120173365 on Build GRCh38
rs201161551 Chr.3: 120173267-120173267 on Build GRCh38
rs774491348 Chr.3: 120173317-120173317 on Build GRCh38
rs767823327 Chr.3: 120173369-120173369 on Build GRCh38
rs920414978 Chr.3: 120173184-120173184 on Build GRCh38
rs779642936 Chr.3: 120173378-120173378 on Build GRCh38
rs769932964 Chr.3: 120173206-120173206 on Build GRCh38
rs748046980 Chr.3: 120173400-120173400 on Build GRCh38
rs770043908 Chr.3: 120173422-120173422 on Build GRCh38
rs748112296 Chr.3: 120173324-120173324 on Build GRCh38
rs755761419 Chr.3: 120173377-120173377 on Build GRCh38
rs531495176 Chr.3: 120173490-120173491 on Build GRCh38
rs1003798453 Chr.3: 120173537-120173537 on Build GRCh38
rs959467836 Chr.3: 120173585-120173585 on Build GRCh38
rs561396784 Chr.3: 120173335-120173335 on Build GRCh38
rs528168471 Chr.3: 120173442-120173442 on Build GRCh38
rs748792521 Chr.3: 120173293-120173293 on Build GRCh38
rs911590552 Chr.3: 120173577-120173577 on Build GRCh38
rs183816015 Chr.3: 120173235-120173235 on Build GRCh38
rs981926585 Chr.3: 120173246-120173246 on Build GRCh38

View Full Product Details