Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 125971749-125972293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG1L
Entrez Gene ID
200810
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like
Gene Aliases
ALG1L1
UniGene
Hs.591299
Gene Chromosome Location
Chr.3: 125929272-125993832 on Build GRCh38
Transcripts
XM_017005844.1 XM_017005846.1 NM_001012337.2 NM_001308313.1 XM_005247138.3 XM_006713513.3

Gene Symbol
ROPN1B
Entrez Gene ID
152015
Gene Name
rhophilin associated tail protein 1B
Gene Aliases
-
UniGene
Hs.663128
Gene Chromosome Location
Chr.3: 125969144-125983454 on Build GRCh38
Transcripts
XM_017005844.1 XM_017005846.1 NM_001012337.2 NM_001308313.1 XM_005247138.3 XM_006713513.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3364974 c.70G>T p.A24S Chr.3: 125972124-125972124 on Build GRCh38
4583888 c.48A>G p.K16K Chr.3: 125972102-125972102 on Build GRCh38
295892 c.83C>A p.A28E Chr.3: 125972137-125972137 on Build GRCh38
5678562 c.45G>C p.P15P Chr.3: 125972099-125972099 on Build GRCh38
5420198 c.70_71insC p.A25fs*18 Chr.3: 125972124-125972125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537546998 Chr.3: 125972219-125972219 on Build GRCh38
rs145882895 Chr.3: 125971912-125971912 on Build GRCh38
rs377693137 Chr.3: 125971953-125971953 on Build GRCh38
rs758171322 Chr.3: 125972168-125972168 on Build GRCh38
rs948116331 Chr.3: 125972173-125972173 on Build GRCh38
rs574524081 Chr.3: 125972230-125972230 on Build GRCh38
rs368172951 Chr.3: 125972241-125972241 on Build GRCh38
rs560889493 Chr.3: 125971948-125971948 on Build GRCh38
rs371106382 Chr.3: 125972208-125972208 on Build GRCh38
rs754952885 Chr.3: 125972164-125972164 on Build GRCh38
rs767032568 Chr.3: 125972160-125972161 on Build GRCh38
rs145292095 Chr.3: 125972200-125972200 on Build GRCh38
rs754467615 Chr.3: 125972102-125972102 on Build GRCh38
rs184659715 Chr.3: 125972105-125972105 on Build GRCh38
rs906558247 Chr.3: 125971953-125971956 on Build GRCh38
rs534381110 Chr.3: 125971802-125971802 on Build GRCh38
rs144905673 Chr.3: 125972161-125972161 on Build GRCh38
rs767698838 Chr.3: 125972146-125972146 on Build GRCh38
rs751116724 Chr.3: 125972236-125972236 on Build GRCh38
rs374826502 Chr.3: 125972076-125972076 on Build GRCh38
rs771587475 Chr.3: 125971872-125971872 on Build GRCh38
rs149819719 Chr.3: 125971796-125971796 on Build GRCh38
rs543339048 Chr.3: 125972238-125972238 on Build GRCh38
rs76431673 Chr.3: 125972042-125972042 on Build GRCh38
rs753416048 Chr.3: 125972098-125972098 on Build GRCh38
rs200722264 Chr.3: 125972143-125972143 on Build GRCh38
rs116779889 Chr.3: 125972229-125972229 on Build GRCh38
rs772367820 Chr.3: 125972207-125972207 on Build GRCh38
rs143434891 Chr.3: 125972169-125972169 on Build GRCh38
rs778462153 Chr.3: 125972069-125972069 on Build GRCh38
rs758118210 Chr.3: 125972112-125972112 on Build GRCh38
rs777122175 Chr.3: 125972035-125972035 on Build GRCh38
rs531957608 Chr.3: 125971899-125971899 on Build GRCh38
rs376269226 Chr.3: 125972016-125972016 on Build GRCh38
rs939361272 Chr.3: 125972198-125972198 on Build GRCh38
rs200039132 Chr.3: 125972080-125972080 on Build GRCh38
rs201357418 Chr.3: 125972127-125972127 on Build GRCh38
rs760529425 Chr.3: 125972209-125972209 on Build GRCh38
rs201333048 Chr.3: 125972085-125972085 on Build GRCh38
rs752221407 Chr.3: 125972166-125972166 on Build GRCh38
rs762894825 Chr.3: 125972077-125972077 on Build GRCh38
rs574337764 Chr.3: 125971957-125971957 on Build GRCh38
rs530628677 Chr.3: 125972178-125972179 on Build GRCh38
rs368481108 Chr.3: 125972124-125972125 on Build GRCh38
rs769150938 Chr.3: 125972191-125972191 on Build GRCh38
rs761614386 Chr.3: 125972084-125972084 on Build GRCh38
rs759163496 Chr.3: 125972035-125972037 on Build GRCh38
rs755531803 Chr.3: 125972242-125972242 on Build GRCh38
rs528729954 Chr.3: 125972020-125972020 on Build GRCh38
rs766141327 Chr.3: 125972211-125972211 on Build GRCh38
rs559977062 Chr.3: 125971996-125971996 on Build GRCh38
rs545546571 Chr.3: 125972255-125972255 on Build GRCh38
rs146423466 Chr.3: 125972243-125972243 on Build GRCh38
rs1029450577 Chr.3: 125972233-125972233 on Build GRCh38
rs771985077 Chr.3: 125972265-125972265 on Build GRCh38
rs374099307 Chr.3: 125972045-125972045 on Build GRCh38
rs920679437 Chr.3: 125971874-125971874 on Build GRCh38
rs892598560 Chr.3: 125972039-125972039 on Build GRCh38
rs759585437 Chr.3: 125972217-125972217 on Build GRCh38
rs776563955 Chr.3: 125972212-125972212 on Build GRCh38
rs188973581 Chr.3: 125972171-125972171 on Build GRCh38
rs764331603 Chr.3: 125972078-125972078 on Build GRCh38
rs756120052 Chr.3: 125972087-125972087 on Build GRCh38
rs538353898 Chr.3: 125971810-125971810 on Build GRCh38
rs546537691 Chr.3: 125972144-125972144 on Build GRCh38
rs554237276 Chr.3: 125972246-125972246 on Build GRCh38
rs756922168 Chr.3: 125972237-125972237 on Build GRCh38
rs778717496 Chr.3: 125972103-125972103 on Build GRCh38
rs749855283 Chr.3: 125972189-125972189 on Build GRCh38
rs377668832 Chr.3: 125972071-125972071 on Build GRCh38
rs752276278 Chr.3: 125972226-125972226 on Build GRCh38
rs749375037 Chr.3: 125972133-125972133 on Build GRCh38
rs773110520 Chr.3: 125972150-125972150 on Build GRCh38
rs778664625 Chr.3: 125972008-125972008 on Build GRCh38
rs750183822 Chr.3: 125972086-125972086 on Build GRCh38
rs544160933 Chr.3: 125971929-125971929 on Build GRCh38
rs900023675 Chr.3: 125972221-125972221 on Build GRCh38
rs543353168 Chr.3: 125971984-125971984 on Build GRCh38
rs760344612 Chr.3: 125972151-125972151 on Build GRCh38
rs780625374 Chr.3: 125972175-125972175 on Build GRCh38
rs775822176 Chr.3: 125972131-125972131 on Build GRCh38
rs867630625 Chr.3: 125972156-125972156 on Build GRCh38
rs536133990 Chr.3: 125972128-125972128 on Build GRCh38
rs566310881 Chr.3: 125972165-125972165 on Build GRCh38
rs575117499 Chr.3: 125971928-125971928 on Build GRCh38
rs756081764 Chr.3: 125972263-125972264 on Build GRCh38
rs749806922 Chr.3: 125972194-125972194 on Build GRCh38

View Full Product Details