Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126541539-126542072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf22
Entrez Gene ID
152065
Gene Name
chromosome 3 open reading frame 22
Gene Aliases
-
UniGene
Hs.178210
Gene Chromosome Location
Chr.3: 126524439-126558965 on Build GRCh38
Transcripts
NR_130715.1 XM_017005753.1 NM_152889.2

Gene Symbol
CHST13
Entrez Gene ID
166012
Gene Name
carbohydrate sulfotransferase 13
Gene Aliases
C4ST3
UniGene
Hs.292375
Gene Chromosome Location
Chr.3: 126524288-126543291 on Build GRCh38
Transcripts
NR_130715.1 XM_017005753.1 NM_152889.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3695786 c.434C>T p.A145V Chr.3: 126541986-126541986 on Build GRCh38
3726415 c.496G>A p.A166T Chr.3: 126542048-126542048 on Build GRCh38
6313224 c.300G>T p.L100L Chr.3: 126541852-126541852 on Build GRCh38
6096337 c.473_474CC>AA p.A158E Chr.3: 126542025-126542026 on Build GRCh38
7361273 c.496G>T p.A166S Chr.3: 126542048-126542048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755784570 Chr.3: 126541876-126541876 on Build GRCh38
rs767401839 Chr.3: 126542026-126542026 on Build GRCh38
rs983484242 Chr.3: 126541571-126541571 on Build GRCh38
rs997701897 Chr.3: 126541769-126541769 on Build GRCh38
rs907567570 Chr.3: 126541577-126541577 on Build GRCh38
rs775760466 Chr.3: 126541943-126541943 on Build GRCh38
rs138078497 Chr.3: 126541842-126541842 on Build GRCh38
rs949224583 Chr.3: 126541641-126541641 on Build GRCh38
rs528621656 Chr.3: 126542013-126542013 on Build GRCh38
rs933756121 Chr.3: 126541946-126541946 on Build GRCh38
rs368220564 Chr.3: 126541839-126541839 on Build GRCh38
rs777682935 Chr.3: 126541763-126541763 on Build GRCh38
rs963741993 Chr.3: 126541797-126541797 on Build GRCh38
rs865862437 Chr.3: 126541607-126541607 on Build GRCh38
rs752028882 Chr.3: 126542045-126542045 on Build GRCh38
rs763186865 Chr.3: 126542004-126542004 on Build GRCh38
rs58576050 Chr.3: 126542014-126542014 on Build GRCh38
rs751471294 Chr.3: 126541864-126541864 on Build GRCh38
rs1016514217 Chr.3: 126541794-126541794 on Build GRCh38
rs1001820778 Chr.3: 126542019-126542019 on Build GRCh38
rs779610571 Chr.3: 126541971-126541971 on Build GRCh38
rs1056829843 Chr.3: 126541566-126541566 on Build GRCh38
rs552342903 Chr.3: 126541955-126541955 on Build GRCh38
rs770408862 Chr.3: 126541767-126541767 on Build GRCh38
rs760487755 Chr.3: 126542027-126542027 on Build GRCh38
rs780594261 Chr.3: 126541867-126541867 on Build GRCh38
rs774580925 Chr.3: 126541796-126541796 on Build GRCh38
rs34311016 Chr.3: 126541988-126541988 on Build GRCh38
rs970685842 Chr.3: 126542018-126542018 on Build GRCh38
rs779745607 Chr.3: 126541957-126541957 on Build GRCh38
rs764069151 Chr.3: 126541856-126541856 on Build GRCh38
rs573241475 Chr.3: 126541738-126541738 on Build GRCh38
rs765437703 Chr.3: 126541939-126541939 on Build GRCh38
rs1050229893 Chr.3: 126541632-126541632 on Build GRCh38
rs754626828 Chr.3: 126541978-126541978 on Build GRCh38
rs576908435 Chr.3: 126541710-126541710 on Build GRCh38
rs927454191 Chr.3: 126541762-126541762 on Build GRCh38
rs766596402 Chr.3: 126541818-126541818 on Build GRCh38
rs916630454 Chr.3: 126541639-126541639 on Build GRCh38
rs866549861 Chr.3: 126541837-126541837 on Build GRCh38
rs1019645443 Chr.3: 126541704-126541704 on Build GRCh38
rs780867298 Chr.3: 126541778-126541778 on Build GRCh38
rs149468240 Chr.3: 126541873-126541873 on Build GRCh38
rs374324912 Chr.3: 126541830-126541830 on Build GRCh38
rs776579439 Chr.3: 126541937-126541937 on Build GRCh38
rs369350672 Chr.3: 126542046-126542046 on Build GRCh38
rs752479972 Chr.3: 126541757-126541757 on Build GRCh38
rs886646066 Chr.3: 126541975-126541975 on Build GRCh38
rs931893240 Chr.3: 126541893-126541893 on Build GRCh38
rs12495696 Chr.3: 126541787-126541787 on Build GRCh38
rs745388619 Chr.3: 126541910-126541910 on Build GRCh38
rs374527763 Chr.3: 126541981-126541981 on Build GRCh38
rs372238580 Chr.3: 126541921-126541921 on Build GRCh38
rs766755977 Chr.3: 126541733-126541733 on Build GRCh38
rs1050699958 Chr.3: 126541894-126541894 on Build GRCh38
rs200175838 Chr.3: 126541986-126541986 on Build GRCh38
rs772759411 Chr.3: 126541803-126541803 on Build GRCh38
rs752501660 Chr.3: 126541846-126541846 on Build GRCh38
rs559156519 Chr.3: 126541744-126541744 on Build GRCh38
rs763909971 Chr.3: 126541951-126541951 on Build GRCh38
rs568699770 Chr.3: 126542038-126542038 on Build GRCh38
rs554585502 Chr.3: 126541602-126541602 on Build GRCh38
rs940318349 Chr.3: 126541608-126541608 on Build GRCh38
rs748539742 Chr.3: 126541802-126541802 on Build GRCh38
rs868823115 Chr.3: 126542024-126542024 on Build GRCh38
rs1039478444 Chr.3: 126541748-126541748 on Build GRCh38
rs377327226 Chr.3: 126541686-126541686 on Build GRCh38
rs989840123 Chr.3: 126541922-126541922 on Build GRCh38
rs1010238518 Chr.3: 126541714-126541714 on Build GRCh38
rs1021589837 Chr.3: 126542002-126542002 on Build GRCh38
rs778705094 Chr.3: 126542042-126542042 on Build GRCh38
rs757148719 Chr.3: 126541865-126541865 on Build GRCh38
rs1030473591 Chr.3: 126541770-126541770 on Build GRCh38
rs753975861 Chr.3: 126541743-126541743 on Build GRCh38
rs990741756 Chr.3: 126541821-126541821 on Build GRCh38
rs536191113 Chr.3: 126541568-126541568 on Build GRCh38
rs981888051 Chr.3: 126541647-126541647 on Build GRCh38
rs1039361311 Chr.3: 126541932-126541932 on Build GRCh38
rs909232711 Chr.3: 126541576-126541576 on Build GRCh38
rs968965509 Chr.3: 126541724-126541724 on Build GRCh38
rs748978705 Chr.3: 126541974-126541974 on Build GRCh38
rs751310243 Chr.3: 126541954-126541954 on Build GRCh38
rs576121588 Chr.3: 126541622-126541622 on Build GRCh38
rs769888325 Chr.3: 126541995-126541995 on Build GRCh38
rs1038058598 Chr.3: 126541616-126541616 on Build GRCh38
rs779741509 Chr.3: 126541895-126541895 on Build GRCh38
rs920509443 Chr.3: 126541859-126541859 on Build GRCh38
rs765934770 Chr.3: 126542035-126542035 on Build GRCh38
rs777485794 Chr.3: 126541721-126541721 on Build GRCh38
rs757864875 Chr.3: 126541684-126541684 on Build GRCh38
rs749955300 Chr.3: 126541970-126541970 on Build GRCh38
rs370653372 Chr.3: 126541812-126541812 on Build GRCh38
rs6766712 Chr.3: 126541765-126541765 on Build GRCh38
rs942375618 Chr.3: 126541747-126541747 on Build GRCh38
rs769441945 Chr.3: 126541789-126541789 on Build GRCh38
rs758338397 Chr.3: 126541758-126541758 on Build GRCh38
rs923874023 Chr.3: 126541668-126541668 on Build GRCh38
rs771505966 Chr.3: 126541936-126541936 on Build GRCh38
rs760875652 Chr.3: 126541817-126541817 on Build GRCh38
rs574367973 Chr.3: 126541746-126541746 on Build GRCh38
rs762780763 Chr.3: 126541948-126541948 on Build GRCh38
rs541211080 Chr.3: 126541720-126541720 on Build GRCh38
rs753340766 Chr.3: 126542036-126542036 on Build GRCh38
rs778742193 Chr.3: 126541831-126541831 on Build GRCh38
rs757930551 Chr.3: 126541928-126541928 on Build GRCh38
rs371981274 Chr.3: 126542047-126542047 on Build GRCh38
rs367830517 Chr.3: 126541987-126541987 on Build GRCh38
rs536705182 Chr.3: 126541637-126541637 on Build GRCh38
rs745985936 Chr.3: 126541990-126541990 on Build GRCh38
rs200849667 Chr.3: 126542048-126542048 on Build GRCh38
rs768812040 Chr.3: 126542011-126542011 on Build GRCh38
rs951281206 Chr.3: 126541911-126541911 on Build GRCh38
rs778294672 Chr.3: 126541979-126541979 on Build GRCh38
rs143024190 Chr.3: 126541929-126541929 on Build GRCh38
rs748508077 Chr.3: 126541916-126541916 on Build GRCh38
rs899074449 Chr.3: 126541618-126541618 on Build GRCh38

View Full Product Details