Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126543006-126543510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf22
Entrez Gene ID
152065
Gene Name
chromosome 3 open reading frame 22
Gene Aliases
-
UniGene
Hs.178210
Gene Chromosome Location
Chr.3: 126524439-126558965 on Build GRCh38
Transcripts
NR_130715.1 XM_017005753.1 NM_152889.2

Gene Symbol
CHST13
Entrez Gene ID
166012
Gene Name
carbohydrate sulfotransferase 13
Gene Aliases
C4ST3
UniGene
Hs.292375
Gene Chromosome Location
Chr.3: 126524288-126543291 on Build GRCh38
Transcripts
NR_130715.1 XM_017005753.1 NM_152889.2

SNP Detail

SNP ID Chromosome Location
rs761029387 Chr.3: 126543025-126543025 on Build GRCh38
rs965948915 Chr.3: 126543119-126543119 on Build GRCh38
rs898064895 Chr.3: 126543281-126543281 on Build GRCh38
rs968734894 Chr.3: 126543300-126543300 on Build GRCh38
rs138379461 Chr.3: 126543069-126543069 on Build GRCh38
rs1051189887 Chr.3: 126543406-126543406 on Build GRCh38
rs988443609 Chr.3: 126543064-126543064 on Build GRCh38
rs1018544052 Chr.3: 126543114-126543114 on Build GRCh38
rs1022691262 Chr.3: 126543043-126543043 on Build GRCh38
rs1049658056 Chr.3: 126543402-126543402 on Build GRCh38
rs888892673 Chr.3: 126543441-126543441 on Build GRCh38
rs946740875 Chr.3: 126543083-126543083 on Build GRCh38
rs981141327 Chr.3: 126543061-126543061 on Build GRCh38
rs373912257 Chr.3: 126543153-126543153 on Build GRCh38
rs757638508 Chr.3: 126543100-126543100 on Build GRCh38
rs988675562 Chr.3: 126543285-126543285 on Build GRCh38
rs1051179772 Chr.3: 126543145-126543145 on Build GRCh38
rs995024311 Chr.3: 126543365-126543365 on Build GRCh38
rs202219023 Chr.3: 126543161-126543162 on Build GRCh38
rs979638637 Chr.3: 126543171-126543171 on Build GRCh38
rs917283825 Chr.3: 126543238-126543238 on Build GRCh38
rs913803825 Chr.3: 126543149-126543149 on Build GRCh38
rs921306299 Chr.3: 126543374-126543374 on Build GRCh38
rs1027441082 Chr.3: 126543029-126543029 on Build GRCh38
rs529446361 Chr.3: 126543250-126543250 on Build GRCh38
rs926062142 Chr.3: 126543172-126543172 on Build GRCh38
rs898385570 Chr.3: 126543213-126543213 on Build GRCh38
rs1054097 Chr.3: 126543249-126543249 on Build GRCh38
rs932746707 Chr.3: 126543125-126543125 on Build GRCh38
rs939510463 Chr.3: 126543063-126543063 on Build GRCh38
rs1007185364 Chr.3: 126543103-126543103 on Build GRCh38
rs914177485 Chr.3: 126543291-126543292 on Build GRCh38
rs969748508 Chr.3: 126543051-126543051 on Build GRCh38
rs1043723291 Chr.3: 126543115-126543115 on Build GRCh38
rs13323871 Chr.3: 126543444-126543444 on Build GRCh38
rs898312645 Chr.3: 126543478-126543478 on Build GRCh38
rs779381961 Chr.3: 126543402-126543404 on Build GRCh38
rs544804103 Chr.3: 126543252-126543252 on Build GRCh38
rs979663277 Chr.3: 126543362-126543362 on Build GRCh38
rs937398729 Chr.3: 126543179-126543179 on Build GRCh38
rs540681869 Chr.3: 126543165-126543165 on Build GRCh38
rs764509492 Chr.3: 126543038-126543038 on Build GRCh38
rs368679004 Chr.3: 126543222-126543222 on Build GRCh38
rs995149089 Chr.3: 126543217-126543217 on Build GRCh38
rs573164016 Chr.3: 126543143-126543143 on Build GRCh38
rs558106415 Chr.3: 126543123-126543123 on Build GRCh38
rs377744268 Chr.3: 126543208-126543208 on Build GRCh38
rs912599886 Chr.3: 126543410-126543410 on Build GRCh38
rs1055816604 Chr.3: 126543191-126543191 on Build GRCh38
rs960911965 Chr.3: 126543273-126543273 on Build GRCh38
rs545808856 Chr.3: 126543118-126543118 on Build GRCh38

View Full Product Details